Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65309408-65309946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC6
Entrez Gene ID
9829
Gene Name
DnaJ heat shock protein family (Hsp40) member C6
Gene Aliases
DJC6, PARK19
UniGene
Hs.647643
Gene Chromosome Location
Chr.1: 65264694-65415869 on Build GRCh38
Transcripts
NM_001256864.1 NM_001256865.1 NM_014787.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936872 c.23G>A p.R8Q Chr.1: 65309768-65309768 on Build GRCh38
6835183 c.151G>C p.A51P Chr.1: 65309896-65309896 on Build GRCh38
3985197 c.129C>T p.A43A Chr.1: 65309874-65309874 on Build GRCh38
5637993 c.104T>G p.V35G Chr.1: 65309849-65309849 on Build GRCh38
5992644 c.122T>C p.V41A Chr.1: 65309867-65309867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748705237 Chr.1: 65309895-65309895 on Build GRCh38
rs528656788 Chr.1: 65309817-65309817 on Build GRCh38
rs772515803 Chr.1: 65309606-65309606 on Build GRCh38
rs748410720 Chr.1: 65309594-65309594 on Build GRCh38
rs931797277 Chr.1: 65309882-65309882 on Build GRCh38
rs562640978 Chr.1: 65309638-65309638 on Build GRCh38
rs761277268 Chr.1: 65309790-65309790 on Build GRCh38
rs747115530 Chr.1: 65309915-65309915 on Build GRCh38
rs1023388534 Chr.1: 65309717-65309717 on Build GRCh38
rs545097873 Chr.1: 65309674-65309674 on Build GRCh38
rs893661339 Chr.1: 65309469-65309469 on Build GRCh38
rs935166785 Chr.1: 65309467-65309467 on Build GRCh38
rs944049034 Chr.1: 65309551-65309551 on Build GRCh38
rs985548488 Chr.1: 65309886-65309886 on Build GRCh38
rs953016668 Chr.1: 65309580-65309580 on Build GRCh38
rs982478224 Chr.1: 65309436-65309436 on Build GRCh38
rs953542132 Chr.1: 65309503-65309503 on Build GRCh38
rs918718181 Chr.1: 65309862-65309862 on Build GRCh38
rs971851819 Chr.1: 65309860-65309860 on Build GRCh38
rs754115488 Chr.1: 65309864-65309864 on Build GRCh38
rs564672952 Chr.1: 65309678-65309678 on Build GRCh38
rs1028921511 Chr.1: 65309484-65309484 on Build GRCh38
rs772276142 Chr.1: 65309901-65309901 on Build GRCh38
rs867834466 Chr.1: 65309783-65309783 on Build GRCh38
rs778059320 Chr.1: 65309908-65309908 on Build GRCh38
rs7540105 Chr.1: 65309552-65309552 on Build GRCh38
rs779540362 Chr.1: 65309889-65309889 on Build GRCh38
rs533079013 Chr.1: 65309673-65309673 on Build GRCh38
rs962076080 Chr.1: 65309767-65309767 on Build GRCh38
rs911273317 Chr.1: 65309888-65309888 on Build GRCh38
rs751543013 Chr.1: 65309757-65309757 on Build GRCh38
rs970364944 Chr.1: 65309432-65309432 on Build GRCh38
rs989287585 Chr.1: 65309494-65309494 on Build GRCh38
rs750060801 Chr.1: 65309831-65309831 on Build GRCh38
rs981764572 Chr.1: 65309830-65309830 on Build GRCh38
rs956622990 Chr.1: 65309846-65309846 on Build GRCh38
rs373500242 Chr.1: 65309648-65309648 on Build GRCh38
rs989729855 Chr.1: 65309853-65309853 on Build GRCh38
rs1030812222 Chr.1: 65309449-65309449 on Build GRCh38
rs947446037 Chr.1: 65309540-65309540 on Build GRCh38
rs12753869 Chr.1: 65309887-65309887 on Build GRCh38
rs916872480 Chr.1: 65309909-65309909 on Build GRCh38
rs1018676240 Chr.1: 65309597-65309597 on Build GRCh38
rs532042007 Chr.1: 65309647-65309648 on Build GRCh38
rs755667643 Chr.1: 65309841-65309841 on Build GRCh38
rs779764117 Chr.1: 65309849-65309849 on Build GRCh38
rs547312338 Chr.1: 65309899-65309899 on Build GRCh38
rs1029157803 Chr.1: 65309499-65309499 on Build GRCh38
rs774601067 Chr.1: 65309450-65309450 on Build GRCh38
rs573619153 Chr.1: 65309541-65309541 on Build GRCh38
rs962129498 Chr.1: 65309826-65309826 on Build GRCh38
rs915104442 Chr.1: 65309872-65309872 on Build GRCh38
rs1016282360 Chr.1: 65309662-65309662 on Build GRCh38
rs868404743 Chr.1: 65309542-65309542 on Build GRCh38
rs1010788397 Chr.1: 65309474-65309474 on Build GRCh38
rs7528227 Chr.1: 65309504-65309504 on Build GRCh38
rs1049786636 Chr.1: 65309545-65309545 on Build GRCh38
rs914674350 Chr.1: 65309527-65309527 on Build GRCh38
rs889806990 Chr.1: 65309547-65309547 on Build GRCh38

View Full Product Details