Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145867005-145866506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD35
Entrez Gene ID
148741
Gene Name
ankyrin repeat domain 35
Gene Aliases
-
UniGene
Hs.710624
Gene Chromosome Location
Chr.1: 145866560-145885866 on Build GRCh38
Transcripts
NM_001280799.1 NM_144698.4

View Full Product Details