Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147186166-147186712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1L
Entrez Gene ID
9557
Gene Name
chromodomain helicase DNA binding protein 1 like
Gene Aliases
ALC1, CHDL
UniGene
Hs.191164
Gene Chromosome Location
Chr.1: 147172804-147295766 on Build GRCh38
Transcripts
XM_017002858.1 NM_001144829.2 NM_001144830.2 NM_001461.3 XM_005272946.4 XM_005272947.4 XM_005272948.4 XM_006711245.3 XM_011509350.2 XM_011509351.2 XM_017000801.1 XM_017000802.1

Gene Symbol
FMO5
Entrez Gene ID
2330
Gene Name
flavin containing monooxygenase 5
Gene Aliases
-
UniGene
Hs.642706
Gene Chromosome Location
Chr.1: 147183963-147225798 on Build GRCh38
Transcripts
XM_017002858.1 NM_001144829.2 NM_001144830.2 NM_001461.3 XM_005272946.4 XM_005272947.4 XM_005272948.4 XM_006711245.3 XM_011509350.2 XM_011509351.2 XM_017000801.1 XM_017000802.1

SNP Detail

SNP ID Chromosome Location
rs1013597758 Chr.1: 147186367-147186367 on Build GRCh38
rs989921299 Chr.1: 147186572-147186572 on Build GRCh38
rs587760946 Chr.1: 147186416-147186416 on Build GRCh38
rs142076274 Chr.1: 147186659-147186659 on Build GRCh38
rs35637998 Chr.1: 147186470-147186471 on Build GRCh38
rs1049282674 Chr.1: 147186326-147186326 on Build GRCh38
rs587719919 Chr.1: 147186418-147186418 on Build GRCh38
rs998569092 Chr.1: 147186532-147186532 on Build GRCh38
rs775464968 Chr.1: 147186508-147186510 on Build GRCh38
rs370230703 Chr.1: 147186573-147186573 on Build GRCh38
rs759422792 Chr.1: 147186277-147186277 on Build GRCh38
rs587737883 Chr.1: 147186601-147186601 on Build GRCh38
rs587690790 Chr.1: 147186638-147186638 on Build GRCh38
rs953931375 Chr.1: 147186604-147186604 on Build GRCh38
rs781950911 Chr.1: 147186420-147186420 on Build GRCh38
rs190673591 Chr.1: 147186620-147186620 on Build GRCh38
rs587642250 Chr.1: 147186417-147186417 on Build GRCh38
rs587655004 Chr.1: 147186374-147186374 on Build GRCh38
rs987127357 Chr.1: 147186632-147186632 on Build GRCh38
rs185815053 Chr.1: 147186577-147186577 on Build GRCh38
rs587748234 Chr.1: 147186650-147186650 on Build GRCh38
rs765198160 Chr.1: 147186400-147186400 on Build GRCh38
rs587689774 Chr.1: 147186415-147186415 on Build GRCh38
rs762836487 Chr.1: 147186687-147186687 on Build GRCh38
rs373649430 Chr.1: 147186608-147186608 on Build GRCh38
rs782089643 Chr.1: 147186539-147186539 on Build GRCh38
rs994992277 Chr.1: 147186288-147186288 on Build GRCh38
rs11387382 Chr.1: 147186462-147186463 on Build GRCh38
rs1023939515 Chr.1: 147186380-147186380 on Build GRCh38
rs957145990 Chr.1: 147186549-147186549 on Build GRCh38
rs587753891 Chr.1: 147186342-147186342 on Build GRCh38
rs1036852736 Chr.1: 147186257-147186257 on Build GRCh38
rs1803590 Chr.1: 147186543-147186543 on Build GRCh38
rs921111477 Chr.1: 147186580-147186580 on Build GRCh38

View Full Product Details