Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137435082-137435575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD8
Entrez Gene ID
377841
Gene Name
ectonucleoside triphosphate diphosphohydrolase 8
Gene Aliases
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492
UniGene
Hs.512562
Gene Chromosome Location
Chr.9: 137434364-137441710 on Build GRCh38
Transcripts
NM_001033113.1 NM_198585.2 XM_011518665.2 XM_011518666.2 XM_011518668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654353 c.1186-1G>T p.? Chr.9: 137435106-137435106 on Build GRCh38
1461408 c.1087C>T p.R363W Chr.9: 137435302-137435302 on Build GRCh38
6654355 c.1083G>A p.W361* Chr.9: 137435306-137435306 on Build GRCh38
1756071 c.1189C>T p.R397C Chr.9: 137435311-137435311 on Build GRCh38
3906065 c.1145T>G p.F382C Chr.9: 137435244-137435244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026223260 Chr.9: 137435544-137435544 on Build GRCh38
rs567186934 Chr.9: 137435134-137435134 on Build GRCh38
rs769589401 Chr.9: 137435135-137435135 on Build GRCh38
rs551339544 Chr.9: 137435518-137435520 on Build GRCh38
rs760176355 Chr.9: 137435227-137435227 on Build GRCh38
rs757918921 Chr.9: 137435105-137435105 on Build GRCh38
rs954761233 Chr.9: 137435482-137435482 on Build GRCh38
rs140353416 Chr.9: 137435257-137435257 on Build GRCh38
rs770767416 Chr.9: 137435174-137435175 on Build GRCh38
rs530607873 Chr.9: 137435400-137435400 on Build GRCh38
rs372285284 Chr.9: 137435167-137435167 on Build GRCh38
rs28479537 Chr.9: 137435106-137435106 on Build GRCh38
rs761246221 Chr.9: 137435246-137435246 on Build GRCh38
rs143518789 Chr.9: 137435241-137435241 on Build GRCh38
rs779873409 Chr.9: 137435190-137435190 on Build GRCh38
rs200307848 Chr.9: 137435373-137435373 on Build GRCh38
rs968782795 Chr.9: 137435242-137435242 on Build GRCh38
rs753529973 Chr.9: 137435262-137435262 on Build GRCh38
rs371472118 Chr.9: 137435173-137435173 on Build GRCh38
rs73565586 Chr.9: 137435440-137435440 on Build GRCh38
rs28589413 Chr.9: 137435166-137435166 on Build GRCh38
rs535828564 Chr.9: 137435428-137435428 on Build GRCh38
rs567378953 Chr.9: 137435409-137435409 on Build GRCh38
rs746883282 Chr.9: 137435168-137435169 on Build GRCh38
rs750429363 Chr.9: 137435184-137435184 on Build GRCh38
rs933590089 Chr.9: 137435407-137435407 on Build GRCh38
rs770383661 Chr.9: 137435299-137435299 on Build GRCh38
rs760203966 Chr.9: 137435336-137435336 on Build GRCh38
rs751804841 Chr.9: 137435355-137435355 on Build GRCh38
rs912396611 Chr.9: 137435496-137435496 on Build GRCh38
rs1042588470 Chr.9: 137435417-137435417 on Build GRCh38
rs552966559 Chr.9: 137435185-137435185 on Build GRCh38
rs892877869 Chr.9: 137435498-137435498 on Build GRCh38
rs756780703 Chr.9: 137435179-137435179 on Build GRCh38
rs61491031 Chr.9: 137435218-137435218 on Build GRCh38
rs759911377 Chr.9: 137435202-137435202 on Build GRCh38
rs112686769 Chr.9: 137435533-137435533 on Build GRCh38
rs765134519 Chr.9: 137435345-137435345 on Build GRCh38
rs756522593 Chr.9: 137435152-137435153 on Build GRCh38
rs766946980 Chr.9: 137435159-137435159 on Build GRCh38
rs1002805472 Chr.9: 137435534-137435534 on Build GRCh38
rs1033109003 Chr.9: 137435162-137435162 on Build GRCh38
rs758462119 Chr.9: 137435167-137435177 on Build GRCh38
rs757590818 Chr.9: 137435358-137435358 on Build GRCh38
rs981317208 Chr.9: 137435331-137435331 on Build GRCh38
rs761502158 Chr.9: 137435311-137435311 on Build GRCh38
rs781049080 Chr.9: 137435181-137435181 on Build GRCh38
rs558313761 Chr.9: 137435506-137435506 on Build GRCh38
rs61570143 Chr.9: 137435249-137435249 on Build GRCh38
rs898145311 Chr.9: 137435326-137435326 on Build GRCh38
rs575311659 Chr.9: 137435240-137435240 on Build GRCh38
rs767073234 Chr.9: 137435248-137435248 on Build GRCh38
rs781139036 Chr.9: 137435119-137435119 on Build GRCh38
rs747652502 Chr.9: 137435165-137435166 on Build GRCh38
rs181397246 Chr.9: 137435521-137435521 on Build GRCh38
rs747298693 Chr.9: 137435272-137435272 on Build GRCh38
rs1054651239 Chr.9: 137435451-137435451 on Build GRCh38
rs775683857 Chr.9: 137435383-137435383 on Build GRCh38
rs1029419862 Chr.9: 137435448-137435448 on Build GRCh38
rs772148614 Chr.9: 137435207-137435207 on Build GRCh38
rs1044474353 Chr.9: 137435384-137435385 on Build GRCh38
rs376575168 Chr.9: 137435175-137435175 on Build GRCh38
rs560582680 Chr.9: 137435332-137435332 on Build GRCh38
rs538571885 Chr.9: 137435474-137435474 on Build GRCh38
rs759910540 Chr.9: 137435165-137435165 on Build GRCh38
rs766611901 Chr.9: 137435161-137435162 on Build GRCh38
rs370001647 Chr.9: 137435178-137435178 on Build GRCh38
rs28558849 Chr.9: 137435545-137435545 on Build GRCh38
rs202080880 Chr.9: 137435258-137435258 on Build GRCh38
rs919059384 Chr.9: 137435212-137435212 on Build GRCh38
rs1036987754 Chr.9: 137435319-137435319 on Build GRCh38
rs969380977 Chr.9: 137435433-137435435 on Build GRCh38
rs758394530 Chr.9: 137435508-137435508 on Build GRCh38
rs749872587 Chr.9: 137435390-137435390 on Build GRCh38
rs771117470 Chr.9: 137435275-137435275 on Build GRCh38
rs757181046 Chr.9: 137435118-137435118 on Build GRCh38
rs987574796 Chr.9: 137435483-137435483 on Build GRCh38
rs746343980 Chr.9: 137435293-137435293 on Build GRCh38
rs879454321 Chr.9: 137435392-137435392 on Build GRCh38
rs537189881 Chr.9: 137435100-137435100 on Build GRCh38
rs943372749 Chr.9: 137435247-137435247 on Build GRCh38
rs1023338307 Chr.9: 137435310-137435310 on Build GRCh38
rs931237686 Chr.9: 137435525-137435525 on Build GRCh38
rs1012580613 Chr.9: 137435304-137435304 on Build GRCh38
rs372533240 Chr.9: 137435147-137435147 on Build GRCh38
rs769068055 Chr.9: 137435302-137435302 on Build GRCh38
rs781593375 Chr.9: 137435363-137435363 on Build GRCh38
rs147993952 Chr.9: 137435197-137435197 on Build GRCh38
rs777293937 Chr.9: 137435161-137435161 on Build GRCh38
rs778619489 Chr.9: 137435164-137435174 on Build GRCh38
rs139960415 Chr.9: 137435234-137435234 on Build GRCh38
rs981264669 Chr.9: 137435186-137435186 on Build GRCh38
rs563105010 Chr.9: 137435399-137435399 on Build GRCh38
rs754526387 Chr.9: 137435163-137435164 on Build GRCh38
rs574712118 Chr.9: 137435268-137435268 on Build GRCh38
rs112894145 Chr.9: 137435549-137435549 on Build GRCh38
rs78674871 Chr.9: 137435301-137435301 on Build GRCh38
rs943405928 Chr.9: 137435385-137435385 on Build GRCh38
rs550662960 Chr.9: 137435406-137435406 on Build GRCh38
rs777880303 Chr.9: 137435169-137435177 on Build GRCh38
rs939868658 Chr.9: 137435499-137435499 on Build GRCh38
rs546587842 Chr.9: 137435429-137435429 on Build GRCh38
rs771135023 Chr.9: 137435103-137435103 on Build GRCh38
rs753823470 Chr.9: 137435344-137435344 on Build GRCh38
rs754819736 Chr.9: 137435267-137435267 on Build GRCh38
rs773862600 Chr.9: 137435307-137435307 on Build GRCh38
rs745893259 Chr.9: 137435367-137435367 on Build GRCh38
rs200839415 Chr.9: 137435239-137435239 on Build GRCh38
rs950503997 Chr.9: 137435362-137435362 on Build GRCh38
rs752399141 Chr.9: 137435104-137435104 on Build GRCh38
rs773205956 Chr.9: 137435156-137435156 on Build GRCh38
rs112534145 Chr.9: 137435109-137435109 on Build GRCh38
rs144921257 Chr.9: 137435219-137435219 on Build GRCh38
rs766172009 Chr.9: 137435338-137435338 on Build GRCh38
rs764401210 Chr.9: 137435500-137435500 on Build GRCh38
rs760843487 Chr.9: 137435158-137435158 on Build GRCh38
rs138995854 Chr.9: 137435211-137435211 on Build GRCh38
rs371232160 Chr.9: 137435121-137435121 on Build GRCh38
rs762557874 Chr.9: 137435174-137435174 on Build GRCh38
rs746939533 Chr.9: 137435110-137435110 on Build GRCh38
rs572829352 Chr.9: 137435393-137435393 on Build GRCh38
rs772670458 Chr.9: 137435335-137435335 on Build GRCh38

View Full Product Details