Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137436738-137437229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD8
Entrez Gene ID
377841
Gene Name
ectonucleoside triphosphate diphosphohydrolase 8
Gene Aliases
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492
UniGene
Hs.512562
Gene Chromosome Location
Chr.9: 137434364-137441710 on Build GRCh38
Transcripts
NM_001033113.1 NM_198585.2 XM_011518665.2 XM_011518666.2 XM_011518668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3906070 c.499G>A p.A167T Chr.9: 137436925-137436925 on Build GRCh38
4681074 c.415G>A p.A139T Chr.9: 137437009-137437009 on Build GRCh38
3906073 c.331C>A p.P111T Chr.9: 137437223-137437223 on Build GRCh38
6654351 c.337G>A p.A113T Chr.9: 137437217-137437217 on Build GRCh38
1686108 c.519G>A p.W173* Chr.9: 137436905-137436905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751196020 Chr.9: 137436856-137436856 on Build GRCh38
rs781375797 Chr.9: 137436949-137436949 on Build GRCh38
rs748704829 Chr.9: 137437030-137437030 on Build GRCh38
rs556579838 Chr.9: 137436929-137436929 on Build GRCh38
rs201535559 Chr.9: 137437025-137437025 on Build GRCh38
rs776640416 Chr.9: 137436843-137436843 on Build GRCh38
rs372119964 Chr.9: 137436861-137436861 on Build GRCh38
rs755690701 Chr.9: 137436871-137436871 on Build GRCh38
rs753227484 Chr.9: 137436984-137436984 on Build GRCh38
rs145244915 Chr.9: 137437032-137437032 on Build GRCh38
rs773975060 Chr.9: 137436884-137436884 on Build GRCh38
rs183326706 Chr.9: 137437027-137437027 on Build GRCh38
rs767535576 Chr.9: 137436884-137436885 on Build GRCh38
rs751299947 Chr.9: 137437076-137437076 on Build GRCh38
rs769916036 Chr.9: 137436911-137436911 on Build GRCh38
rs370214919 Chr.9: 137436970-137436970 on Build GRCh38
rs779009511 Chr.9: 137437177-137437177 on Build GRCh38
rs754991362 Chr.9: 137437153-137437153 on Build GRCh38
rs376423061 Chr.9: 137436886-137436886 on Build GRCh38
rs760383211 Chr.9: 137437208-137437208 on Build GRCh38
rs371071887 Chr.9: 137437073-137437073 on Build GRCh38
rs762131755 Chr.9: 137437049-137437049 on Build GRCh38
rs767214714 Chr.9: 137436919-137436919 on Build GRCh38
rs747829056 Chr.9: 137436944-137436944 on Build GRCh38
rs776046508 Chr.9: 137436912-137436912 on Build GRCh38
rs754517266 Chr.9: 137436990-137436990 on Build GRCh38
rs754820881 Chr.9: 137436932-137436932 on Build GRCh38
rs771382589 Chr.9: 137437195-137437195 on Build GRCh38
rs988484839 Chr.9: 137436804-137436804 on Build GRCh38
rs962107524 Chr.9: 137437082-137437082 on Build GRCh38
rs937360096 Chr.9: 137437148-137437148 on Build GRCh38
rs375091607 Chr.9: 137437066-137437066 on Build GRCh38
rs1056538791 Chr.9: 137436868-137436868 on Build GRCh38
rs578205858 Chr.9: 137436879-137436879 on Build GRCh38
rs766653328 Chr.9: 137436842-137436842 on Build GRCh38
rs781015130 Chr.9: 137436864-137436864 on Build GRCh38
rs749262276 Chr.9: 137437026-137437026 on Build GRCh38
rs769956706 Chr.9: 137437024-137437024 on Build GRCh38
rs375691923 Chr.9: 137436847-137436847 on Build GRCh38
rs1017168574 Chr.9: 137436759-137436759 on Build GRCh38
rs940054366 Chr.9: 137437099-137437099 on Build GRCh38
rs535800241 Chr.9: 137436918-137436918 on Build GRCh38
rs926835553 Chr.9: 137436820-137436820 on Build GRCh38
rs771081051 Chr.9: 137436889-137436889 on Build GRCh38
rs371537336 Chr.9: 137437130-137437130 on Build GRCh38
rs139821464 Chr.9: 137436923-137436923 on Build GRCh38
rs768053348 Chr.9: 137436977-137436977 on Build GRCh38
rs758951971 Chr.9: 137437043-137437043 on Build GRCh38
rs201022191 Chr.9: 137436791-137436791 on Build GRCh38
rs1049941219 Chr.9: 137437142-137437142 on Build GRCh38
rs769838323 Chr.9: 137436956-137436956 on Build GRCh38
rs898688029 Chr.9: 137436774-137436774 on Build GRCh38
rs760846135 Chr.9: 137436818-137436818 on Build GRCh38
rs191083989 Chr.9: 137436849-137436849 on Build GRCh38
rs202059855 Chr.9: 137436915-137436915 on Build GRCh38
rs915157202 Chr.9: 137436833-137436833 on Build GRCh38
rs372334778 Chr.9: 137436798-137436798 on Build GRCh38
rs898744147 Chr.9: 137436929-137436929 on Build GRCh38
rs758162819 Chr.9: 137436996-137436996 on Build GRCh38
rs764847969 Chr.9: 137437051-137437051 on Build GRCh38
rs143455567 Chr.9: 137436961-137436961 on Build GRCh38
rs766777881 Chr.9: 137437121-137437127 on Build GRCh38
rs777588687 Chr.9: 137437005-137437005 on Build GRCh38
rs903039347 Chr.9: 137436777-137436777 on Build GRCh38
rs746841951 Chr.9: 137437012-137437012 on Build GRCh38
rs761018753 Chr.9: 137436970-137436970 on Build GRCh38
rs370632675 Chr.9: 137437028-137437028 on Build GRCh38
rs758690977 Chr.9: 137436779-137436779 on Build GRCh38
rs777473391 Chr.9: 137436780-137436780 on Build GRCh38
rs960510730 Chr.9: 137437092-137437092 on Build GRCh38
rs759269087 Chr.9: 137437207-137437207 on Build GRCh38
rs891263947 Chr.9: 137436958-137436958 on Build GRCh38
rs1000513533 Chr.9: 137436789-137436789 on Build GRCh38
rs745405388 Chr.9: 137436793-137436793 on Build GRCh38
rs1020153555 Chr.9: 137436810-137436822 on Build GRCh38
rs748261207 Chr.9: 137437179-137437179 on Build GRCh38
rs747370376 Chr.9: 137437194-137437194 on Build GRCh38
rs766076261 Chr.9: 137436982-137436982 on Build GRCh38
rs28606499 Chr.9: 137437096-137437096 on Build GRCh38
rs778708451 Chr.9: 137436993-137436993 on Build GRCh38
rs533175240 Chr.9: 137436878-137436878 on Build GRCh38
rs753417933 Chr.9: 137436931-137436931 on Build GRCh38
rs780136184 Chr.9: 137436877-137436877 on Build GRCh38
rs61730142 Chr.9: 137437198-137437198 on Build GRCh38
rs771917149 Chr.9: 137436946-137436946 on Build GRCh38
rs376653113 Chr.9: 137436803-137436803 on Build GRCh38
rs199654541 Chr.9: 137437119-137437119 on Build GRCh38
rs773116259 Chr.9: 137437204-137437204 on Build GRCh38
rs779751159 Chr.9: 137436925-137436925 on Build GRCh38
rs375616394 Chr.9: 137436790-137436790 on Build GRCh38
rs780137460 Chr.9: 137437121-137437121 on Build GRCh38
rs1033821970 Chr.9: 137437016-137437016 on Build GRCh38
rs745317623 Chr.9: 137436865-137436865 on Build GRCh38
rs200288034 Chr.9: 137436962-137436962 on Build GRCh38
rs987727728 Chr.9: 137436880-137436880 on Build GRCh38
rs201213794 Chr.9: 137436832-137436832 on Build GRCh38
rs547613223 Chr.9: 137436800-137436800 on Build GRCh38
rs760362592 Chr.9: 137436981-137436981 on Build GRCh38
rs920591634 Chr.9: 137437126-137437126 on Build GRCh38
rs141698920 Chr.9: 137436890-137436890 on Build GRCh38
rs759621709 Chr.9: 137436845-137436845 on Build GRCh38
rs768988574 Chr.9: 137436966-137436966 on Build GRCh38
rs772647050 Chr.9: 137437180-137437180 on Build GRCh38
rs992273217 Chr.9: 137437095-137437095 on Build GRCh38
rs892219752 Chr.9: 137437201-137437201 on Build GRCh38
rs778919634 Chr.9: 137436943-137436943 on Build GRCh38
rs754135698 Chr.9: 137437184-137437184 on Build GRCh38
rs1038374917 Chr.9: 137437110-137437110 on Build GRCh38
rs889010121 Chr.9: 137436900-137436900 on Build GRCh38
rs61730138 Chr.9: 137437200-137437200 on Build GRCh38
rs761133443 Chr.9: 137437036-137437036 on Build GRCh38
rs770403483 Chr.9: 137436799-137436799 on Build GRCh38
rs765661709 Chr.9: 137436846-137436846 on Build GRCh38
rs765753413 Chr.9: 137436770-137436770 on Build GRCh38
rs541802399 Chr.9: 137437105-137437105 on Build GRCh38
rs755436342 Chr.9: 137436766-137436766 on Build GRCh38
rs773858173 Chr.9: 137437187-137437187 on Build GRCh38
rs928945008 Chr.9: 137437141-137437141 on Build GRCh38
rs746701597 Chr.9: 137436787-137436787 on Build GRCh38
rs762264022 Chr.9: 137436974-137436974 on Build GRCh38
rs956599665 Chr.9: 137436969-137436969 on Build GRCh38
rs752999322 Chr.9: 137436773-137436773 on Build GRCh38
rs746141018 Chr.9: 137436952-137436952 on Build GRCh38
rs371455616 Chr.9: 137436792-137436792 on Build GRCh38
rs954528307 Chr.9: 137436867-137436867 on Build GRCh38
rs373018363 Chr.9: 137436989-137436989 on Build GRCh38
rs752733354 Chr.9: 137436995-137436995 on Build GRCh38
rs367656206 Chr.9: 137436894-137436894 on Build GRCh38
rs773661492 Chr.9: 137437033-137437033 on Build GRCh38
rs200175060 Chr.9: 137437075-137437075 on Build GRCh38
rs758780532 Chr.9: 137436848-137436848 on Build GRCh38
rs750068355 Chr.9: 137436980-137436980 on Build GRCh38
rs145935333 Chr.9: 137436922-137436922 on Build GRCh38
rs765877671 Chr.9: 137437040-137437040 on Build GRCh38
rs1024928651 Chr.9: 137437029-137437029 on Build GRCh38
rs376780742 Chr.9: 137437072-137437072 on Build GRCh38

View Full Product Details