Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137615202-137615700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC1
Entrez Gene ID
92714
Gene Name
arrestin domain containing 1
Gene Aliases
-
UniGene
Hs.12999
Gene Chromosome Location
Chr.9: 137605640-137615360 on Build GRCh38
Transcripts
NM_001317968.1 NM_152285.3 XM_005266119.1 XM_006717320.3 XM_006717321.3 XM_006717322.3 XM_017015288.1 NR_122035.1 NR_122036.1

Gene Symbol
ARRDC1-AS1
Entrez Gene ID
85026
Gene Name
ARRDC1 antisense RNA 1
Gene Aliases
C9orf37
UniGene
Hs.522520
Gene Chromosome Location
Chr.9: 137615332-137618895 on Build GRCh38
Transcripts
NM_001317968.1 NM_152285.3 XM_005266119.1 XM_006717320.3 XM_006717321.3 XM_006717322.3 XM_017015288.1 NR_122035.1 NR_122036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285641 c.525T>C p.G175G Chr.9: 137615675-137615675 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000134992 Chr.9: 137615336-137615336 on Build GRCh38
rs1035020601 Chr.9: 137615425-137615425 on Build GRCh38
rs951510097 Chr.9: 137615465-137615465 on Build GRCh38
rs185438817 Chr.9: 137615373-137615373 on Build GRCh38
rs890243555 Chr.9: 137615344-137615344 on Build GRCh38
rs888812043 Chr.9: 137615380-137615380 on Build GRCh38
rs931681063 Chr.9: 137615289-137615289 on Build GRCh38
rs1018745262 Chr.9: 137615397-137615397 on Build GRCh38
rs901466736 Chr.9: 137615575-137615575 on Build GRCh38
rs761332133 Chr.9: 137615642-137615642 on Build GRCh38
rs1004684735 Chr.9: 137615600-137615600 on Build GRCh38
rs558221589 Chr.9: 137615522-137615548 on Build GRCh38
rs762108093 Chr.9: 137615570-137615570 on Build GRCh38
rs3177227 Chr.9: 137615347-137615347 on Build GRCh38
rs963537965 Chr.9: 137615627-137615627 on Build GRCh38
rs766970979 Chr.9: 137615676-137615676 on Build GRCh38
rs3174164 Chr.9: 137615263-137615263 on Build GRCh38
rs973278873 Chr.9: 137615491-137615491 on Build GRCh38
rs1475494 Chr.9: 137615329-137615329 on Build GRCh38
rs919536531 Chr.9: 137615224-137615224 on Build GRCh38
rs1033934758 Chr.9: 137615617-137615617 on Build GRCh38
rs111508914 Chr.9: 137615508-137615508 on Build GRCh38
rs982898906 Chr.9: 137615471-137615471 on Build GRCh38
rs114986132 Chr.9: 137615271-137615271 on Build GRCh38
rs774882876 Chr.9: 137615407-137615407 on Build GRCh38
rs560291200 Chr.9: 137615514-137615514 on Build GRCh38
rs1028787927 Chr.9: 137615653-137615653 on Build GRCh38
rs959610383 Chr.9: 137615628-137615628 on Build GRCh38
rs763692127 Chr.9: 137615490-137615490 on Build GRCh38
rs905185880 Chr.9: 137615415-137615415 on Build GRCh38
rs1034988153 Chr.9: 137615565-137615565 on Build GRCh38
rs952338408 Chr.9: 137615236-137615236 on Build GRCh38
rs769591479 Chr.9: 137615322-137615322 on Build GRCh38
rs112136439 Chr.9: 137615223-137615223 on Build GRCh38
rs935100336 Chr.9: 137615353-137615353 on Build GRCh38
rs532021726 Chr.9: 137615576-137615576 on Build GRCh38
rs893948538 Chr.9: 137615522-137615522 on Build GRCh38
rs577645167 Chr.9: 137615547-137615547 on Build GRCh38
rs953264556 Chr.9: 137615657-137615657 on Build GRCh38
rs911525187 Chr.9: 137615679-137615679 on Build GRCh38
rs1026486131 Chr.9: 137615458-137615458 on Build GRCh38
rs151003285 Chr.9: 137615666-137615666 on Build GRCh38
rs552612651 Chr.9: 137615548-137615548 on Build GRCh38
rs553044554 Chr.9: 137615253-137615253 on Build GRCh38
rs770635803 Chr.9: 137615637-137615637 on Build GRCh38
rs1040047924 Chr.9: 137615567-137615567 on Build GRCh38
rs14357 Chr.9: 137615249-137615249 on Build GRCh38
rs1052998902 Chr.9: 137615356-137615356 on Build GRCh38
rs773887815 Chr.9: 137615513-137615513 on Build GRCh38
rs776269668 Chr.9: 137615639-137615639 on Build GRCh38
rs537046769 Chr.9: 137615622-137615622 on Build GRCh38
rs377335997 Chr.9: 137615530-137615530 on Build GRCh38
rs1003584543 Chr.9: 137615422-137615422 on Build GRCh38
rs181357513 Chr.9: 137615272-137615272 on Build GRCh38
rs945481306 Chr.9: 137615519-137615519 on Build GRCh38
rs758987670 Chr.9: 137615663-137615663 on Build GRCh38
rs772447555 Chr.9: 137615619-137615619 on Build GRCh38
rs548848133 Chr.9: 137615588-137615588 on Build GRCh38
rs924480964 Chr.9: 137615535-137615535 on Build GRCh38
rs992328301 Chr.9: 137615629-137615629 on Build GRCh38
rs993619835 Chr.9: 137615427-137615427 on Build GRCh38
rs532510724 Chr.9: 137615540-137615540 on Build GRCh38
rs949965454 Chr.9: 137615273-137615273 on Build GRCh38
rs188291368 Chr.9: 137615633-137615633 on Build GRCh38
rs754396232 Chr.9: 137615549-137615549 on Build GRCh38
rs540292107 Chr.9: 137615507-137615507 on Build GRCh38
rs1006328618 Chr.9: 137615614-137615614 on Build GRCh38
rs116415813 Chr.9: 137615391-137615391 on Build GRCh38
rs3750386 Chr.9: 137615607-137615607 on Build GRCh38
rs762591060 Chr.9: 137615392-137615392 on Build GRCh38
rs761251348 Chr.9: 137615521-137615521 on Build GRCh38
rs751471089 Chr.9: 137615681-137615681 on Build GRCh38
rs563232735 Chr.9: 137615557-137615557 on Build GRCh38
rs560724541 Chr.9: 137615494-137615494 on Build GRCh38
rs760416491 Chr.9: 137615412-137615413 on Build GRCh38
rs574635966 Chr.9: 137615499-137615499 on Build GRCh38
rs1015145964 Chr.9: 137615483-137615483 on Build GRCh38

View Full Product Details