Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137752203-137752699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
UniGene
Hs.495511
Gene Chromosome Location
Chr.9: 137618988-137836127 on Build GRCh38
Transcripts
NM_001145527.1 NM_024757.4 XM_005266105.4 XM_005266110.1 XM_006717288.2 XM_011519021.2 XM_011519022.2 XM_011519023.2 XM_011519024.2 XM_011519025.1 XM_011519026.2 XM_011519027.2 XM_011519028.2 XM_011519033.2 XM_017015134.1 XM_017015135.1 XM_017015136.1 XM_017015137.1 XM_017015138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5875978 c.1139G>A p.G380D Chr.9: 137752392-137752392 on Build GRCh38
5875977 c.1232G>A p.G411D Chr.9: 137752392-137752392 on Build GRCh38
6203433 c.1122_1124delGGA p.E378delE Chr.9: 137752375-137752377 on Build GRCh38
6203432 c.1215_1217delGGA p.E409delE Chr.9: 137752375-137752377 on Build GRCh38
3906167 c.1176C>T p.D392D Chr.9: 137752336-137752336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749977131 Chr.9: 137752460-137752460 on Build GRCh38
rs763737811 Chr.9: 137752221-137752221 on Build GRCh38
rs1032319342 Chr.9: 137752509-137752509 on Build GRCh38
rs1022654481 Chr.9: 137752493-137752493 on Build GRCh38
rs532903216 Chr.9: 137752516-137752516 on Build GRCh38
rs533474318 Chr.9: 137752233-137752233 on Build GRCh38
rs1031695920 Chr.9: 137752426-137752426 on Build GRCh38
rs982834494 Chr.9: 137752607-137752607 on Build GRCh38
rs867506224 Chr.9: 137752510-137752510 on Build GRCh38
rs745856672 Chr.9: 137752448-137752448 on Build GRCh38
rs777305555 Chr.9: 137752424-137752424 on Build GRCh38
rs980124173 Chr.9: 137752557-137752557 on Build GRCh38
rs777413326 Chr.9: 137752301-137752301 on Build GRCh38
rs780631623 Chr.9: 137752450-137752450 on Build GRCh38
rs11137202 Chr.9: 137752626-137752626 on Build GRCh38
rs755473562 Chr.9: 137752498-137752498 on Build GRCh38
rs551967315 Chr.9: 137752543-137752543 on Build GRCh38
rs539526279 Chr.9: 137752592-137752592 on Build GRCh38
rs535429498 Chr.9: 137752643-137752643 on Build GRCh38
rs556292023 Chr.9: 137752598-137752598 on Build GRCh38
rs894613555 Chr.9: 137752358-137752358 on Build GRCh38
rs549814854 Chr.9: 137752317-137752317 on Build GRCh38
rs745619662 Chr.9: 137752316-137752316 on Build GRCh38
rs754380060 Chr.9: 137752391-137752391 on Build GRCh38
rs990569265 Chr.9: 137752527-137752527 on Build GRCh38
rs1056407860 Chr.9: 137752591-137752591 on Build GRCh38
rs922350033 Chr.9: 137752237-137752237 on Build GRCh38
rs756631550 Chr.9: 137752316-137752317 on Build GRCh38
rs529239920 Chr.9: 137752429-137752429 on Build GRCh38
rs549300129 Chr.9: 137752433-137752433 on Build GRCh38
rs374067222 Chr.9: 137752396-137752396 on Build GRCh38
rs781303008 Chr.9: 137752313-137752313 on Build GRCh38
rs768444103 Chr.9: 137752421-137752421 on Build GRCh38
rs1044893501 Chr.9: 137752267-137752267 on Build GRCh38
rs369984436 Chr.9: 137752364-137752364 on Build GRCh38
rs117256904 Chr.9: 137752222-137752222 on Build GRCh38
rs766991704 Chr.9: 137752432-137752432 on Build GRCh38
rs375739205 Chr.9: 137752345-137752345 on Build GRCh38
rs753035963 Chr.9: 137752658-137752658 on Build GRCh38
rs377468927 Chr.9: 137752602-137752602 on Build GRCh38
rs963886995 Chr.9: 137752232-137752232 on Build GRCh38
rs373204327 Chr.9: 137752579-137752579 on Build GRCh38
rs773281152 Chr.9: 137752341-137752341 on Build GRCh38
rs373083497 Chr.9: 137752336-137752336 on Build GRCh38
rs987108055 Chr.9: 137752488-137752488 on Build GRCh38
rs772187480 Chr.9: 137752337-137752337 on Build GRCh38
rs555371598 Chr.9: 137752672-137752672 on Build GRCh38
rs1045753654 Chr.9: 137752390-137752390 on Build GRCh38
rs377629839 Chr.9: 137752420-137752420 on Build GRCh38
rs754348302 Chr.9: 137752446-137752446 on Build GRCh38
rs962468518 Chr.9: 137752407-137752407 on Build GRCh38
rs955515244 Chr.9: 137752487-137752487 on Build GRCh38
rs926493309 Chr.9: 137752563-137752563 on Build GRCh38
rs769706934 Chr.9: 137752457-137752457 on Build GRCh38
rs566340735 Chr.9: 137752439-137752439 on Build GRCh38
rs770935375 Chr.9: 137752308-137752308 on Build GRCh38
rs192219035 Chr.9: 137752578-137752578 on Build GRCh38
rs963833908 Chr.9: 137752500-137752500 on Build GRCh38
rs187988466 Chr.9: 137752529-137752529 on Build GRCh38
rs746509340 Chr.9: 137752307-137752307 on Build GRCh38
rs766828352 Chr.9: 137752373-137752373 on Build GRCh38
rs948808310 Chr.9: 137752545-137752545 on Build GRCh38
rs774904451 Chr.9: 137752227-137752227 on Build GRCh38

View Full Product Details