Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2910202-2910706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
UniGene
Hs.528631
Gene Chromosome Location
Chr.X: 2903970-2929375 on Build GRCh38
Transcripts
NM_001669.3 XM_005274514.2 XM_005274515.2

SNP Detail

SNP ID Chromosome Location
rs780452551 Chr.X: 2910459-2910459 on Build GRCh38
rs7357997 Chr.X: 2910530-2910530 on Build GRCh38
rs372941673 Chr.X: 2910625-2910625 on Build GRCh38
rs769256693 Chr.X: 2910624-2910624 on Build GRCh38
rs201397958 Chr.X: 2910316-2910317 on Build GRCh38
rs1027525415 Chr.X: 2910280-2910281 on Build GRCh38
rs868618320 Chr.X: 2910316-2910316 on Build GRCh38
rs922046854 Chr.X: 2910364-2910364 on Build GRCh38
rs151160381 Chr.X: 2910672-2910672 on Build GRCh38
rs747427502 Chr.X: 2910594-2910594 on Build GRCh38
rs966274951 Chr.X: 2910342-2910342 on Build GRCh38
rs750033876 Chr.X: 2910462-2910462 on Build GRCh38
rs1053132713 Chr.X: 2910536-2910536 on Build GRCh38
rs183333373 Chr.X: 2910446-2910446 on Build GRCh38
rs750529722 Chr.X: 2910283-2910284 on Build GRCh38
rs1015709545 Chr.X: 2910351-2910361 on Build GRCh38
rs866105632 Chr.X: 2910319-2910319 on Build GRCh38
rs760561589 Chr.X: 2910300-2910300 on Build GRCh38
rs868504970 Chr.X: 2910324-2910324 on Build GRCh38
rs780608947 Chr.X: 2910679-2910679 on Build GRCh38
rs144786076 Chr.X: 2910634-2910634 on Build GRCh38
rs61634042 Chr.X: 2910318-2910319 on Build GRCh38
rs201426112 Chr.X: 2910284-2910286 on Build GRCh38
rs781272269 Chr.X: 2910235-2910235 on Build GRCh38
rs750645297 Chr.X: 2910383-2910383 on Build GRCh38
rs1001189832 Chr.X: 2910245-2910245 on Build GRCh38
rs907677771 Chr.X: 2910314-2910314 on Build GRCh38
rs925510450 Chr.X: 2910522-2910522 on Build GRCh38
rs199573361 Chr.X: 2910345-2910352 on Build GRCh38
rs1048988213 Chr.X: 2910307-2910307 on Build GRCh38
rs765609350 Chr.X: 2910623-2910623 on Build GRCh38
rs190679072 Chr.X: 2910367-2910367 on Build GRCh38
rs879073274 Chr.X: 2910533-2910533 on Build GRCh38
rs1045476125 Chr.X: 2910630-2910630 on Build GRCh38
rs747575791 Chr.X: 2910671-2910671 on Build GRCh38
rs757990044 Chr.X: 2910489-2910489 on Build GRCh38
rs951993159 Chr.X: 2910311-2910311 on Build GRCh38
rs756323480 Chr.X: 2910447-2910447 on Build GRCh38
rs866820231 Chr.X: 2910323-2910323 on Build GRCh38
rs983786730 Chr.X: 2910314-2910316 on Build GRCh38
rs937215232 Chr.X: 2910279-2910279 on Build GRCh38
rs931969519 Chr.X: 2910387-2910387 on Build GRCh38

View Full Product Details