Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3024121-3024646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSH
Entrez Gene ID
347527
Gene Name
arylsulfatase family member H
Gene Aliases
sulfatase
UniGene
Hs.351533
Gene Chromosome Location
Chr.X: 3006613-3033385 on Build GRCh38
Transcripts
NM_001011719.1

SNP Detail

SNP ID Chromosome Location
rs370929934 Chr.X: 3024142-3024142 on Build GRCh38
rs963957629 Chr.X: 3024392-3024392 on Build GRCh38
rs940920020 Chr.X: 3024478-3024478 on Build GRCh38
rs369705244 Chr.X: 3024301-3024301 on Build GRCh38
rs888281349 Chr.X: 3024344-3024344 on Build GRCh38
rs761919702 Chr.X: 3024289-3024289 on Build GRCh38
rs181215928 Chr.X: 3024418-3024418 on Build GRCh38
rs927660384 Chr.X: 3024575-3024575 on Build GRCh38
rs1046480857 Chr.X: 3024574-3024574 on Build GRCh38
rs776422541 Chr.X: 3024521-3024521 on Build GRCh38
rs769439892 Chr.X: 3024396-3024396 on Build GRCh38
rs764625295 Chr.X: 3024611-3024611 on Build GRCh38
rs777607675 Chr.X: 3024168-3024168 on Build GRCh38
rs772832290 Chr.X: 3024357-3024357 on Build GRCh38
rs374868562 Chr.X: 3024201-3024201 on Build GRCh38
rs989678334 Chr.X: 3024529-3024529 on Build GRCh38
rs948368935 Chr.X: 3024563-3024563 on Build GRCh38
rs958207859 Chr.X: 3024528-3024528 on Build GRCh38
rs984791976 Chr.X: 3024471-3024471 on Build GRCh38
rs879225506 Chr.X: 3024289-3024290 on Build GRCh38
rs761456640 Chr.X: 3024174-3024179 on Build GRCh38
rs972213248 Chr.X: 3024505-3024505 on Build GRCh38
rs1010092426 Chr.X: 3024296-3024296 on Build GRCh38
rs34062105 Chr.X: 3024476-3024477 on Build GRCh38
rs996560068 Chr.X: 3024417-3024417 on Build GRCh38
rs761438928 Chr.X: 3024562-3024562 on Build GRCh38
rs372731165 Chr.X: 3024502-3024502 on Build GRCh38
rs768703783 Chr.X: 3024509-3024509 on Build GRCh38
rs1032956287 Chr.X: 3024199-3024199 on Build GRCh38
rs923402850 Chr.X: 3024511-3024511 on Build GRCh38
rs769762002 Chr.X: 3024156-3024156 on Build GRCh38
rs772010741 Chr.X: 3024421-3024421 on Build GRCh38
rs775741142 Chr.X: 3024185-3024185 on Build GRCh38
rs760690985 Chr.X: 3024499-3024499 on Build GRCh38
rs775363068 Chr.X: 3024448-3024448 on Build GRCh38
rs60737314 Chr.X: 3024301-3024302 on Build GRCh38
rs745920172 Chr.X: 3024174-3024174 on Build GRCh38
rs878948325 Chr.X: 3024270-3024270 on Build GRCh38
rs1054098694 Chr.X: 3024596-3024596 on Build GRCh38
rs188338185 Chr.X: 3024369-3024369 on Build GRCh38
rs920667958 Chr.X: 3024503-3024503 on Build GRCh38
rs144609105 Chr.X: 3024210-3024210 on Build GRCh38
rs775183846 Chr.X: 3024599-3024599 on Build GRCh38
rs778095761 Chr.X: 3024407-3024407 on Build GRCh38
rs1025864749 Chr.X: 3024319-3024319 on Build GRCh38
rs954948333 Chr.X: 3024470-3024470 on Build GRCh38
rs957255209 Chr.X: 3024275-3024275 on Build GRCh38
rs909488599 Chr.X: 3024475-3024475 on Build GRCh38
rs747948261 Chr.X: 3024143-3024143 on Build GRCh38
rs916816264 Chr.X: 3024545-3024545 on Build GRCh38
rs772229342 Chr.X: 3024178-3024178 on Build GRCh38
rs760912036 Chr.X: 3024187-3024187 on Build GRCh38
rs796622372 Chr.X: 3024379-3024379 on Build GRCh38

View Full Product Details