Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 8798267-8798764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9A
Entrez Gene ID
171482
Gene Name
family with sequence similarity 9 member A
Gene Aliases
TEX39A
UniGene
Hs.382062
Gene Chromosome Location
Chr.X: 8790795-8801383 on Build GRCh38
Transcripts
NM_001171186.1 NM_174951.3

SNP Detail

SNP ID Chromosome Location
rs377189496 Chr.X: 8798440-8798440 on Build GRCh38
rs1030097134 Chr.X: 8798465-8798465 on Build GRCh38
rs140609226 Chr.X: 8798462-8798462 on Build GRCh38
rs779932539 Chr.X: 8798617-8798617 on Build GRCh38
rs749521495 Chr.X: 8798464-8798464 on Build GRCh38
rs200743802 Chr.X: 8798504-8798504 on Build GRCh38
rs938159500 Chr.X: 8798708-8798708 on Build GRCh38
rs1048602374 Chr.X: 8798315-8798315 on Build GRCh38
rs369620752 Chr.X: 8798459-8798459 on Build GRCh38
rs745854208 Chr.X: 8798416-8798416 on Build GRCh38
rs780913810 Chr.X: 8798535-8798535 on Build GRCh38
rs760110676 Chr.X: 8798490-8798490 on Build GRCh38
rs753554786 Chr.X: 8798510-8798510 on Build GRCh38
rs755797410 Chr.X: 8798371-8798371 on Build GRCh38
rs772513481 Chr.X: 8798393-8798393 on Build GRCh38
rs372813665 Chr.X: 8798340-8798340 on Build GRCh38
rs926703907 Chr.X: 8798643-8798643 on Build GRCh38
rs982138286 Chr.X: 8798640-8798640 on Build GRCh38
rs771466775 Chr.X: 8798466-8798466 on Build GRCh38
rs141739673 Chr.X: 8798359-8798359 on Build GRCh38
rs760118289 Chr.X: 8798505-8798505 on Build GRCh38
rs768709413 Chr.X: 8798694-8798694 on Build GRCh38
rs755931139 Chr.X: 8798611-8798611 on Build GRCh38
rs745458725 Chr.X: 8798574-8798574 on Build GRCh38
rs970718436 Chr.X: 8798633-8798633 on Build GRCh38
rs772420118 Chr.X: 8798495-8798495 on Build GRCh38
rs763356301 Chr.X: 8798329-8798329 on Build GRCh38
rs778674943 Chr.X: 8798412-8798412 on Build GRCh38
rs376701169 Chr.X: 8798374-8798374 on Build GRCh38
rs376479459 Chr.X: 8798522-8798522 on Build GRCh38
rs941695959 Chr.X: 8798349-8798349 on Build GRCh38
rs781352634 Chr.X: 8798429-8798429 on Build GRCh38
rs112920459 Chr.X: 8798551-8798551 on Build GRCh38
rs752101252 Chr.X: 8798361-8798361 on Build GRCh38
rs888928926 Chr.X: 8798481-8798481 on Build GRCh38
rs956803542 Chr.X: 8798544-8798544 on Build GRCh38
rs774558446 Chr.X: 8798471-8798471 on Build GRCh38
rs774993127 Chr.X: 8798341-8798341 on Build GRCh38
rs773783416 Chr.X: 8798427-8798428 on Build GRCh38
rs1020942477 Chr.X: 8798581-8798581 on Build GRCh38
rs1007805864 Chr.X: 8798493-8798493 on Build GRCh38
rs773720734 Chr.X: 8798316-8798316 on Build GRCh38
rs769447147 Chr.X: 8798641-8798641 on Build GRCh38
rs866060373 Chr.X: 8798358-8798358 on Build GRCh38
rs759929838 Chr.X: 8798347-8798347 on Build GRCh38
rs761022532 Chr.X: 8798503-8798504 on Build GRCh38
rs753642195 Chr.X: 8798407-8798407 on Build GRCh38
rs112014143 Chr.X: 8798565-8798565 on Build GRCh38
rs373017947 Chr.X: 8798521-8798521 on Build GRCh38
rs758241499 Chr.X: 8798427-8798427 on Build GRCh38
rs144824648 Chr.X: 8798569-8798569 on Build GRCh38
rs749268941 Chr.X: 8798682-8798682 on Build GRCh38
rs757101733 Chr.X: 8798408-8798408 on Build GRCh38

View Full Product Details