Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 10158172-10158674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN4
Entrez Gene ID
1183
Gene Name
chloride voltage-gated channel 4
Gene Aliases
CLC4, ClC-4, ClC-4A
UniGene
Hs.495674
Gene Chromosome Location
Chr.X: 10156945-10237660 on Build GRCh38
Transcripts
NM_001256944.1 NM_001830.3

SNP Detail

SNP ID Chromosome Location
rs971288832 Chr.X: 10158528-10158528 on Build GRCh38
rs914733423 Chr.X: 10158642-10158642 on Build GRCh38
rs1039555532 Chr.X: 10158611-10158611 on Build GRCh38
rs952000384 Chr.X: 10158501-10158501 on Build GRCh38
rs1041364398 Chr.X: 10158648-10158648 on Build GRCh38
rs1040053244 Chr.X: 10158337-10158337 on Build GRCh38
rs927312646 Chr.X: 10158554-10158554 on Build GRCh38
rs7877167 Chr.X: 10158392-10158392 on Build GRCh38
rs754456637 Chr.X: 10158474-10158474 on Build GRCh38
rs1042391024 Chr.X: 10158216-10158216 on Build GRCh38
rs912382811 Chr.X: 10158575-10158575 on Build GRCh38
rs892782583 Chr.X: 10158347-10158347 on Build GRCh38
rs907820691 Chr.X: 10158527-10158527 on Build GRCh38
rs959162918 Chr.X: 10158457-10158457 on Build GRCh38
rs963618369 Chr.X: 10158504-10158504 on Build GRCh38
rs1052313218 Chr.X: 10158572-10158572 on Build GRCh38
rs919269966 Chr.X: 10158522-10158522 on Build GRCh38
rs112523997 Chr.X: 10158394-10158394 on Build GRCh38
rs981375501 Chr.X: 10158537-10158537 on Build GRCh38
rs779572152 Chr.X: 10158393-10158393 on Build GRCh38
rs1006798176 Chr.X: 10158313-10158313 on Build GRCh38
rs1008607969 Chr.X: 10158383-10158383 on Build GRCh38
rs893029374 Chr.X: 10158612-10158612 on Build GRCh38
rs1054495472 Chr.X: 10158625-10158625 on Build GRCh38
rs991743332 Chr.X: 10158467-10158467 on Build GRCh38
rs1002626680 Chr.X: 10158407-10158407 on Build GRCh38
rs942549320 Chr.X: 10158603-10158603 on Build GRCh38
rs866128637 Chr.X: 10158425-10158425 on Build GRCh38
rs937405486 Chr.X: 10158621-10158621 on Build GRCh38
rs972011914 Chr.X: 10158521-10158521 on Build GRCh38
rs550467927 Chr.X: 10158217-10158217 on Build GRCh38
rs1011223311 Chr.X: 10158361-10158361 on Build GRCh38
rs983377939 Chr.X: 10158524-10158524 on Build GRCh38
rs1061287 Chr.X: 10158391-10158391 on Build GRCh38

View Full Product Details