Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13319776-13320284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN3L
Entrez Gene ID
92552
Gene Name
ataxin 3 like
Gene Aliases
MJDL
UniGene
Hs.382641
Gene Chromosome Location
Chr.X: 13318647-13320399 on Build GRCh38
Transcripts
NM_001135995.1 NR_046087.1

Gene Symbol
GS1-600G8.3
Entrez Gene ID
100093698
Gene Name
unknown transcript
Gene Aliases
-
UniGene
Hs.643612
Gene Chromosome Location
Chr.X: 13310652-13319933 on Build GRCh38
Transcripts
NM_001135995.1 NR_046087.1

SNP Detail

SNP ID Chromosome Location
rs767189434 Chr.X: 13320068-13320068 on Build GRCh38
rs917866768 Chr.X: 13319857-13319857 on Build GRCh38
rs968140111 Chr.X: 13319803-13319803 on Build GRCh38
rs748416747 Chr.X: 13319919-13319919 on Build GRCh38
rs908043296 Chr.X: 13319994-13319994 on Build GRCh38
rs1040542258 Chr.X: 13320057-13320057 on Build GRCh38
rs998273864 Chr.X: 13319902-13319902 on Build GRCh38
rs373180769 Chr.X: 13319948-13319948 on Build GRCh38
rs763881218 Chr.X: 13319818-13319818 on Build GRCh38
rs1044552887 Chr.X: 13319822-13319822 on Build GRCh38
rs774300596 Chr.X: 13319967-13319967 on Build GRCh38
rs192281200 Chr.X: 13320229-13320229 on Build GRCh38
rs767076385 Chr.X: 13319974-13319974 on Build GRCh38
rs761000735 Chr.X: 13320088-13320088 on Build GRCh38
rs748261338 Chr.X: 13320124-13320124 on Build GRCh38
rs755713922 Chr.X: 13319808-13319808 on Build GRCh38
rs904693853 Chr.X: 13319829-13319829 on Build GRCh38
rs780101194 Chr.X: 13320072-13320072 on Build GRCh38
rs879168064 Chr.X: 13320034-13320034 on Build GRCh38
rs781376546 Chr.X: 13319884-13319884 on Build GRCh38
rs747101778 Chr.X: 13319868-13319868 on Build GRCh38
rs926149328 Chr.X: 13320251-13320251 on Build GRCh38
rs991997735 Chr.X: 13319848-13319848 on Build GRCh38
rs778907160 Chr.X: 13319862-13319862 on Build GRCh38
rs1000960651 Chr.X: 13320199-13320199 on Build GRCh38
rs765291844 Chr.X: 13319950-13319950 on Build GRCh38
rs773722796 Chr.X: 13319947-13319947 on Build GRCh38
rs774889081 Chr.X: 13319968-13319968 on Build GRCh38
rs753022254 Chr.X: 13320075-13320075 on Build GRCh38
rs973512742 Chr.X: 13320049-13320049 on Build GRCh38
rs771406493 Chr.X: 13319965-13319965 on Build GRCh38
rs940887037 Chr.X: 13320009-13320009 on Build GRCh38
rs773573952 Chr.X: 13320247-13320247 on Build GRCh38
rs956713973 Chr.X: 13319828-13319828 on Build GRCh38
rs755066004 Chr.X: 13319881-13319881 on Build GRCh38
rs891490053 Chr.X: 13319892-13319892 on Build GRCh38
rs769747653 Chr.X: 13320039-13320039 on Build GRCh38
rs903817856 Chr.X: 13320173-13320173 on Build GRCh38
rs1000929284 Chr.X: 13319809-13319809 on Build GRCh38
rs980434795 Chr.X: 13319863-13319863 on Build GRCh38
rs201381068 Chr.X: 13319859-13319859 on Build GRCh38
rs1050437250 Chr.X: 13320138-13320138 on Build GRCh38
rs369799117 Chr.X: 13319935-13319935 on Build GRCh38
rs917221473 Chr.X: 13320222-13320222 on Build GRCh38
rs371393419 Chr.X: 13319853-13319853 on Build GRCh38
rs1016816651 Chr.X: 13320001-13320001 on Build GRCh38
rs1031539940 Chr.X: 13320200-13320200 on Build GRCh38
rs1006803980 Chr.X: 13319991-13319991 on Build GRCh38
rs140774548 Chr.X: 13319969-13319969 on Build GRCh38
rs764372724 Chr.X: 13320231-13320231 on Build GRCh38

View Full Product Details