Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130450186-130450688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASS1
Entrez Gene ID
445
Gene Name
argininosuccinate synthase 1
Gene Aliases
ASS, CTLN1
UniGene
Hs.160786
Gene Chromosome Location
Chr.9: 130444707-130501274 on Build GRCh38
Transcripts
NM_000050.4 NM_054012.3 XM_005272200.3 XM_011518705.2 XM_017014729.1

SNP Detail

SNP ID Chromosome Location
rs891087400 Chr.9: 130450449-130450449 on Build GRCh38
rs757321147 Chr.9: 130450501-130450501 on Build GRCh38
rs575191736 Chr.9: 130450306-130450306 on Build GRCh38
rs916753693 Chr.9: 130450550-130450550 on Build GRCh38
rs995658510 Chr.9: 130450484-130450484 on Build GRCh38
rs781330201 Chr.9: 130450587-130450587 on Build GRCh38
rs140853298 Chr.9: 130450257-130450257 on Build GRCh38
rs891060988 Chr.9: 130450418-130450418 on Build GRCh38
rs933155655 Chr.9: 130450659-130450659 on Build GRCh38
rs988917564 Chr.9: 130450607-130450607 on Build GRCh38
rs903528557 Chr.9: 130450368-130450368 on Build GRCh38
rs898784451 Chr.9: 130450482-130450482 on Build GRCh38
rs947214966 Chr.9: 130450388-130450388 on Build GRCh38
rs952101534 Chr.9: 130450276-130450276 on Build GRCh38
rs549216391 Chr.9: 130450309-130450309 on Build GRCh38
rs995780459 Chr.9: 130450661-130450661 on Build GRCh38
rs1039555820 Chr.9: 130450654-130450655 on Build GRCh38
rs561651898 Chr.9: 130450660-130450660 on Build GRCh38
rs545051848 Chr.9: 130450644-130450644 on Build GRCh38
rs1018077926 Chr.9: 130450463-130450463 on Build GRCh38
rs1022721250 Chr.9: 130450275-130450275 on Build GRCh38
rs866436174 Chr.9: 130450425-130450425 on Build GRCh38
rs992388017 Chr.9: 130450533-130450533 on Build GRCh38
rs924630790 Chr.9: 130450619-130450619 on Build GRCh38
rs1014906529 Chr.9: 130450531-130450531 on Build GRCh38
rs1023169563 Chr.9: 130450493-130450493 on Build GRCh38
rs1056668739 Chr.9: 130450319-130450319 on Build GRCh38
rs1052560681 Chr.9: 130450614-130450614 on Build GRCh38
rs1043829391 Chr.9: 130450363-130450363 on Build GRCh38
rs952153088 Chr.9: 130450511-130450511 on Build GRCh38
rs537034604 Chr.9: 130450406-130450406 on Build GRCh38
rs877326 Chr.9: 130450601-130450601 on Build GRCh38
rs866262416 Chr.9: 130450248-130450248 on Build GRCh38
rs974270619 Chr.9: 130450242-130450242 on Build GRCh38
rs1004627104 Chr.9: 130450455-130450455 on Build GRCh38
rs920623587 Chr.9: 130450232-130450232 on Build GRCh38
rs1015835982 Chr.9: 130450481-130450481 on Build GRCh38
rs948170125 Chr.9: 130450558-130450558 on Build GRCh38
rs533855091 Chr.9: 130450515-130450515 on Build GRCh38
rs1051142587 Chr.9: 130450439-130450439 on Build GRCh38
rs190094151 Chr.9: 130450605-130450605 on Build GRCh38
rs184403245 Chr.9: 130450320-130450320 on Build GRCh38
rs75725212 Chr.9: 130450638-130450638 on Build GRCh38
rs537889984 Chr.9: 130450327-130450327 on Build GRCh38
rs145212223 Chr.9: 130450307-130450307 on Build GRCh38
rs932707973 Chr.9: 130450429-130450429 on Build GRCh38
rs575976662 Chr.9: 130450639-130450639 on Build GRCh38
rs75335697 Chr.9: 130450371-130450371 on Build GRCh38
rs947334541 Chr.9: 130450627-130450627 on Build GRCh38
rs377349262 Chr.9: 130450625-130450625 on Build GRCh38
rs12004384 Chr.9: 130450341-130450341 on Build GRCh38
rs536298037 Chr.9: 130450626-130450626 on Build GRCh38
rs925063578 Chr.9: 130450604-130450604 on Build GRCh38

View Full Product Details