Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130471389-130471894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASS1
Entrez Gene ID
445
Gene Name
argininosuccinate synthase 1
Gene Aliases
ASS, CTLN1
UniGene
Hs.160786
Gene Chromosome Location
Chr.9: 130444707-130501274 on Build GRCh38
Transcripts
NM_000050.4 NM_054012.3 XM_005272200.3 XM_011518705.2 XM_017014729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7252448 c.567-5C>T p.? Chr.9: 130471480-130471480 on Build GRCh38
1193644 c.571G>A p.E191K Chr.9: 130471489-130471489 on Build GRCh38
4407022 c.597+8C>T p.? Chr.9: 130471523-130471523 on Build GRCh38
6606094 c.593C>A p.P198H Chr.9: 130471511-130471511 on Build GRCh38
5472680 c.570C>T p.Y190Y Chr.9: 130471488-130471488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980488068 Chr.9: 130471807-130471807 on Build GRCh38
rs573861905 Chr.9: 130471750-130471750 on Build GRCh38
rs147147298 Chr.9: 130471463-130471463 on Build GRCh38
rs376535206 Chr.9: 130471730-130471730 on Build GRCh38
rs534299937 Chr.9: 130471687-130471694 on Build GRCh38
rs577747166 Chr.9: 130471795-130471795 on Build GRCh38
rs373954259 Chr.9: 130471688-130471695 on Build GRCh38
rs641510 Chr.9: 130471596-130471596 on Build GRCh38
rs369407001 Chr.9: 130471482-130471482 on Build GRCh38
rs920751996 Chr.9: 130471642-130471642 on Build GRCh38
rs1057517259 Chr.9: 130471484-130471484 on Build GRCh38
rs761420352 Chr.9: 130471470-130471470 on Build GRCh38
rs796051936 Chr.9: 130471502-130471502 on Build GRCh38
rs927672961 Chr.9: 130471670-130471670 on Build GRCh38
rs1009007360 Chr.9: 130471778-130471778 on Build GRCh38
rs899717122 Chr.9: 130471574-130471574 on Build GRCh38
rs764078809 Chr.9: 130471514-130471514 on Build GRCh38
rs944868686 Chr.9: 130471866-130471866 on Build GRCh38
rs754758285 Chr.9: 130471481-130471481 on Build GRCh38
rs144320037 Chr.9: 130471801-130471801 on Build GRCh38
rs572357790 Chr.9: 130471681-130471681 on Build GRCh38
rs529055563 Chr.9: 130471451-130471451 on Build GRCh38
rs778270266 Chr.9: 130471437-130471437 on Build GRCh38
rs973689163 Chr.9: 130471637-130471637 on Build GRCh38
rs913593358 Chr.9: 130471700-130471700 on Build GRCh38
rs969645083 Chr.9: 130471841-130471841 on Build GRCh38
rs486889 Chr.9: 130471693-130471693 on Build GRCh38
rs758648129 Chr.9: 130471521-130471521 on Build GRCh38
rs973967493 Chr.9: 130471441-130471441 on Build GRCh38
rs373477755 Chr.9: 130471562-130471562 on Build GRCh38
rs184410370 Chr.9: 130471721-130471721 on Build GRCh38
rs369403556 Chr.9: 130471544-130471544 on Build GRCh38
rs908082110 Chr.9: 130471432-130471432 on Build GRCh38
rs567075493 Chr.9: 130471578-130471578 on Build GRCh38
rs753543907 Chr.9: 130471480-130471480 on Build GRCh38
rs760732303 Chr.9: 130471560-130471560 on Build GRCh38
rs887666633 Chr.9: 130471727-130471727 on Build GRCh38
rs917277258 Chr.9: 130471664-130471665 on Build GRCh38
rs555694491 Chr.9: 130471617-130471617 on Build GRCh38
rs189046174 Chr.9: 130471816-130471816 on Build GRCh38
rs1024385943 Chr.9: 130471793-130471793 on Build GRCh38
rs652313 Chr.9: 130471533-130471533 on Build GRCh38
rs749644572 Chr.9: 130471672-130471672 on Build GRCh38
rs560065283 Chr.9: 130471424-130471424 on Build GRCh38
rs375812499 Chr.9: 130471472-130471472 on Build GRCh38
rs768162825 Chr.9: 130471461-130471461 on Build GRCh38
rs773347177 Chr.9: 130471473-130471473 on Build GRCh38
rs756402539 Chr.9: 130471699-130471699 on Build GRCh38
rs926162906 Chr.9: 130471686-130471686 on Build GRCh38
rs992899479 Chr.9: 130471659-130471659 on Build GRCh38
rs1043660180 Chr.9: 130471723-130471723 on Build GRCh38
rs531425163 Chr.9: 130471809-130471809 on Build GRCh38
rs28633982 Chr.9: 130471682-130471682 on Build GRCh38
rs760675113 Chr.9: 130471474-130471474 on Build GRCh38
rs575650478 Chr.9: 130471628-130471628 on Build GRCh38
rs374516554 Chr.9: 130471420-130471420 on Build GRCh38
rs758420576 Chr.9: 130471435-130471435 on Build GRCh38
rs1015199909 Chr.9: 130471731-130471731 on Build GRCh38
rs747575810 Chr.9: 130471494-130471494 on Build GRCh38
rs1009289949 Chr.9: 130471597-130471597 on Build GRCh38
rs140368861 Chr.9: 130471575-130471575 on Build GRCh38
rs890507972 Chr.9: 130471766-130471766 on Build GRCh38
rs752862441 Chr.9: 130471486-130471486 on Build GRCh38
rs551393780 Chr.9: 130471532-130471532 on Build GRCh38
rs77343702 Chr.9: 130471523-130471523 on Build GRCh38
rs544350156 Chr.9: 130471643-130471643 on Build GRCh38
rs781652117 Chr.9: 130471511-130471511 on Build GRCh38
rs980937023 Chr.9: 130471655-130471655 on Build GRCh38
rs181412427 Chr.9: 130471576-130471576 on Build GRCh38
rs776847891 Chr.9: 130471447-130471447 on Build GRCh38
rs368882572 Chr.9: 130471438-130471438 on Build GRCh38
rs1027868121 Chr.9: 130471839-130471839 on Build GRCh38
rs772824744 Chr.9: 130471552-130471552 on Build GRCh38
rs758481857 Chr.9: 130471488-130471488 on Build GRCh38
rs984256519 Chr.9: 130471411-130471411 on Build GRCh38
rs568893606 Chr.9: 130471492-130471492 on Build GRCh38
rs771355143 Chr.9: 130471442-130471442 on Build GRCh38
rs560359118 Chr.9: 130471724-130471724 on Build GRCh38
rs747738960 Chr.9: 130471757-130471757 on Build GRCh38
rs951921567 Chr.9: 130471585-130471585 on Build GRCh38
rs980905815 Chr.9: 130471869-130471869 on Build GRCh38
rs376351776 Chr.9: 130471433-130471433 on Build GRCh38
rs777828000 Chr.9: 130471489-130471489 on Build GRCh38
rs777266339 Chr.9: 130471823-130471823 on Build GRCh38
rs141620505 Chr.9: 130471410-130471410 on Build GRCh38

View Full Product Details