Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130695070-130695611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5225486 c.123-1delG p.? Chr.9: 130695491-130695491 on Build GRCh38
7099076 c.185G>C p.R62T Chr.9: 130695554-130695554 on Build GRCh38
292948 c.123G>T p.R41R Chr.9: 130695492-130695492 on Build GRCh38
6657293 c.156C>T p.L52L Chr.9: 130695525-130695525 on Build GRCh38
4468892 c.156C>G p.L52L Chr.9: 130695525-130695525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027565324 Chr.9: 130695572-130695572 on Build GRCh38
rs951742226 Chr.9: 130695303-130695306 on Build GRCh38
rs1044211628 Chr.9: 130695348-130695348 on Build GRCh38
rs928215325 Chr.9: 130695252-130695252 on Build GRCh38
rs532255445 Chr.9: 130695194-130695194 on Build GRCh38
rs907616912 Chr.9: 130695330-130695330 on Build GRCh38
rs917479363 Chr.9: 130695096-130695096 on Build GRCh38
rs777097865 Chr.9: 130695522-130695522 on Build GRCh38
rs373286412 Chr.9: 130695555-130695555 on Build GRCh38
rs745895735 Chr.9: 130695201-130695201 on Build GRCh38
rs1008717545 Chr.9: 130695192-130695192 on Build GRCh38
rs749542829 Chr.9: 130695279-130695279 on Build GRCh38
rs376037709 Chr.9: 130695450-130695450 on Build GRCh38
rs948973048 Chr.9: 130695122-130695122 on Build GRCh38
rs960465326 Chr.9: 130695344-130695344 on Build GRCh38
rs112461394 Chr.9: 130695444-130695444 on Build GRCh38
rs80050395 Chr.9: 130695326-130695326 on Build GRCh38
rs778394466 Chr.9: 130695505-130695505 on Build GRCh38
rs117466604 Chr.9: 130695347-130695347 on Build GRCh38
rs543445788 Chr.9: 130695126-130695126 on Build GRCh38
rs373490705 Chr.9: 130695447-130695447 on Build GRCh38
rs754110717 Chr.9: 130695479-130695479 on Build GRCh38
rs780076705 Chr.9: 130695207-130695207 on Build GRCh38
rs768823088 Chr.9: 130695317-130695317 on Build GRCh38
rs199841784 Chr.9: 130695564-130695564 on Build GRCh38
rs372828936 Chr.9: 130695177-130695177 on Build GRCh38
rs773672235 Chr.9: 130695448-130695448 on Build GRCh38
rs948906606 Chr.9: 130695368-130695368 on Build GRCh38
rs982437550 Chr.9: 130695142-130695142 on Build GRCh38
rs752164925 Chr.9: 130695490-130695490 on Build GRCh38
rs761827459 Chr.9: 130695458-130695458 on Build GRCh38
rs996074998 Chr.9: 130695241-130695241 on Build GRCh38
rs771446451 Chr.9: 130695521-130695521 on Build GRCh38
rs370282291 Chr.9: 130695560-130695560 on Build GRCh38
rs770027199 Chr.9: 130695203-130695203 on Build GRCh38
rs748117919 Chr.9: 130695503-130695503 on Build GRCh38
rs369974111 Chr.9: 130695497-130695497 on Build GRCh38
rs548222813 Chr.9: 130695382-130695382 on Build GRCh38
rs988370294 Chr.9: 130695552-130695552 on Build GRCh38
rs996451470 Chr.9: 130695533-130695533 on Build GRCh38
rs973816391 Chr.9: 130695476-130695476 on Build GRCh38
rs570396756 Chr.9: 130695472-130695472 on Build GRCh38
rs980948275 Chr.9: 130695229-130695229 on Build GRCh38
rs186124210 Chr.9: 130695480-130695480 on Build GRCh38
rs112939010 Chr.9: 130695419-130695419 on Build GRCh38
rs760500253 Chr.9: 130695379-130695379 on Build GRCh38
rs563649230 Chr.9: 130695176-130695176 on Build GRCh38
rs1052371595 Chr.9: 130695286-130695286 on Build GRCh38
rs1045054677 Chr.9: 130695141-130695141 on Build GRCh38
rs373224770 Chr.9: 130695469-130695469 on Build GRCh38
rs148157757 Chr.9: 130695531-130695531 on Build GRCh38
rs369221914 Chr.9: 130695459-130695459 on Build GRCh38
rs550383127 Chr.9: 130695504-130695504 on Build GRCh38
rs181660957 Chr.9: 130695108-130695108 on Build GRCh38
rs1029589190 Chr.9: 130695118-130695118 on Build GRCh38
rs891916914 Chr.9: 130695178-130695178 on Build GRCh38
rs776570598 Chr.9: 130695508-130695508 on Build GRCh38
rs887720575 Chr.9: 130695581-130695581 on Build GRCh38
rs377234336 Chr.9: 130695571-130695571 on Build GRCh38
rs1000645967 Chr.9: 130695164-130695164 on Build GRCh38
rs996745856 Chr.9: 130695109-130695109 on Build GRCh38
rs748977410 Chr.9: 130695554-130695554 on Build GRCh38
rs7856911 Chr.9: 130695435-130695435 on Build GRCh38
rs939587418 Chr.9: 130695541-130695541 on Build GRCh38
rs376021506 Chr.9: 130695253-130695253 on Build GRCh38

View Full Product Details