Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130703610-130704108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991365 c.802-1G>T p.? Chr.9: 130703693-130703693 on Build GRCh38
6657294 c.802-8delT p.? Chr.9: 130703686-130703686 on Build GRCh38
1105991 c.808G>A p.D270N Chr.9: 130703700-130703700 on Build GRCh38
6031442 c.873G>A p.Q291Q Chr.9: 130703765-130703765 on Build GRCh38
4999047 c.854G>T p.R285L Chr.9: 130703746-130703746 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530567915 Chr.9: 130703890-130703890 on Build GRCh38
rs112701247 Chr.9: 130704042-130704042 on Build GRCh38
rs532912873 Chr.9: 130704034-130704034 on Build GRCh38
rs770189512 Chr.9: 130703784-130703784 on Build GRCh38
rs770845097 Chr.9: 130703752-130703752 on Build GRCh38
rs548741912 Chr.9: 130703940-130703940 on Build GRCh38
rs371501959 Chr.9: 130703736-130703736 on Build GRCh38
rs201500823 Chr.9: 130703793-130703793 on Build GRCh38
rs759692462 Chr.9: 130703661-130703661 on Build GRCh38
rs1056004045 Chr.9: 130703841-130703841 on Build GRCh38
rs762281819 Chr.9: 130703823-130703823 on Build GRCh38
rs376194925 Chr.9: 130703794-130703794 on Build GRCh38
rs748140544 Chr.9: 130703682-130703682 on Build GRCh38
rs1054500783 Chr.9: 130704055-130704055 on Build GRCh38
rs929970554 Chr.9: 130703791-130703791 on Build GRCh38
rs951655091 Chr.9: 130703975-130703975 on Build GRCh38
rs148480990 Chr.9: 130703900-130703900 on Build GRCh38
rs534604208 Chr.9: 130703906-130703906 on Build GRCh38
rs559987015 Chr.9: 130703916-130703916 on Build GRCh38
rs568249955 Chr.9: 130703966-130703966 on Build GRCh38
rs778509334 Chr.9: 130703742-130703742 on Build GRCh38
rs757712197 Chr.9: 130703731-130703731 on Build GRCh38
rs546138489 Chr.9: 130703914-130703914 on Build GRCh38
rs942054228 Chr.9: 130703996-130703997 on Build GRCh38
rs1014547293 Chr.9: 130703880-130703880 on Build GRCh38
rs770662804 Chr.9: 130703651-130703651 on Build GRCh38
rs991590247 Chr.9: 130704009-130704009 on Build GRCh38
rs980689619 Chr.9: 130703874-130703874 on Build GRCh38
rs1027550891 Chr.9: 130703638-130703638 on Build GRCh38
rs765822902 Chr.9: 130703751-130703754 on Build GRCh38
rs1048410650 Chr.9: 130703795-130703795 on Build GRCh38
rs746926392 Chr.9: 130703749-130703749 on Build GRCh38
rs773512685 Chr.9: 130703745-130703745 on Build GRCh38
rs532857774 Chr.9: 130703901-130703901 on Build GRCh38
rs965986468 Chr.9: 130703716-130703716 on Build GRCh38
rs746916936 Chr.9: 130703650-130703650 on Build GRCh38
rs749438442 Chr.9: 130703667-130703667 on Build GRCh38
rs144576118 Chr.9: 130703746-130703746 on Build GRCh38
rs766776428 Chr.9: 130703802-130703802 on Build GRCh38
rs976921840 Chr.9: 130703960-130703960 on Build GRCh38
rs566735734 Chr.9: 130704037-130704037 on Build GRCh38
rs530881474 Chr.9: 130703985-130703985 on Build GRCh38
rs777298280 Chr.9: 130703747-130703747 on Build GRCh38
rs946219090 Chr.9: 130703916-130703916 on Build GRCh38
rs993607249 Chr.9: 130704022-130704022 on Build GRCh38
rs775307152 Chr.9: 130703646-130703646 on Build GRCh38
rs763071172 Chr.9: 130703655-130703656 on Build GRCh38
rs986015752 Chr.9: 130703713-130703713 on Build GRCh38
rs371877055 Chr.9: 130703885-130703885 on Build GRCh38
rs370647022 Chr.9: 130703993-130703993 on Build GRCh38
rs115596953 Chr.9: 130704047-130704047 on Build GRCh38
rs555563445 Chr.9: 130704048-130704048 on Build GRCh38
rs774429658 Chr.9: 130703820-130703820 on Build GRCh38
rs775751006 Chr.9: 130703712-130703712 on Build GRCh38
rs768068113 Chr.9: 130703826-130703826 on Build GRCh38
rs957680514 Chr.9: 130704077-130704077 on Build GRCh38
rs889106527 Chr.9: 130703994-130703994 on Build GRCh38
rs879751862 Chr.9: 130703948-130703948 on Build GRCh38
rs528727312 Chr.9: 130703922-130703922 on Build GRCh38
rs754695417 Chr.9: 130703739-130703739 on Build GRCh38
rs146807696 Chr.9: 130703715-130703715 on Build GRCh38
rs563080155 Chr.9: 130703840-130703840 on Build GRCh38
rs764219579 Chr.9: 130703726-130703726 on Build GRCh38
rs767784154 Chr.9: 130703734-130703734 on Build GRCh38
rs937458651 Chr.9: 130703830-130703830 on Build GRCh38
rs895656777 Chr.9: 130703872-130703872 on Build GRCh38
rs776421866 Chr.9: 130703658-130703658 on Build GRCh38
rs977969547 Chr.9: 130703965-130703965 on Build GRCh38
rs773114946 Chr.9: 130703643-130703643 on Build GRCh38
rs182960894 Chr.9: 130703992-130703992 on Build GRCh38
rs34680820 Chr.9: 130703765-130703765 on Build GRCh38
rs745643416 Chr.9: 130703770-130703770 on Build GRCh38
rs188916148 Chr.9: 130704016-130704016 on Build GRCh38
rs909894688 Chr.9: 130703995-130703995 on Build GRCh38
rs993759812 Chr.9: 130703828-130703828 on Build GRCh38
rs752033760 Chr.9: 130703730-130703730 on Build GRCh38
rs546622361 Chr.9: 130704028-130704028 on Build GRCh38
rs142735531 Chr.9: 130704001-130704001 on Build GRCh38
rs913671442 Chr.9: 130703915-130703915 on Build GRCh38

View Full Product Details