Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132630183-132630691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX31
Entrez Gene ID
64794
Gene Name
DEAD-box helicase 31
Gene Aliases
PPP1R25
UniGene
Hs.660767
Gene Chromosome Location
Chr.9: 132592997-132670403 on Build GRCh38
Transcripts
NM_001322340.1 NM_001322341.1 NM_001322342.1 NM_001322343.1 NM_022779.8 NR_136309.1 XM_005272207.4 XM_006717236.3 XM_011518921.2 XM_011518922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4962821 c.1844G>A p.R615Q Chr.9: 132630366-132630366 on Build GRCh38
7277960 c.1919T>C p.V640A Chr.9: 132630291-132630291 on Build GRCh38
455525 c.1861C>T p.R621W Chr.9: 132630349-132630349 on Build GRCh38
3905126 c.1858G>A p.A620T Chr.9: 132630352-132630352 on Build GRCh38
291986 c.1894T>C p.L632L Chr.9: 132630316-132630316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548078213 Chr.9: 132630366-132630366 on Build GRCh38
rs747739580 Chr.9: 132630364-132630364 on Build GRCh38
rs1038794691 Chr.9: 132630593-132630593 on Build GRCh38
rs761033105 Chr.9: 132630302-132630302 on Build GRCh38
rs924268597 Chr.9: 132630519-132630519 on Build GRCh38
rs1037654063 Chr.9: 132630212-132630212 on Build GRCh38
rs781173884 Chr.9: 132630233-132630233 on Build GRCh38
rs774184361 Chr.9: 132630287-132630287 on Build GRCh38
rs919077552 Chr.9: 132630222-132630222 on Build GRCh38
rs959681366 Chr.9: 132630403-132630403 on Build GRCh38
rs199790657 Chr.9: 132630284-132630284 on Build GRCh38
rs929372463 Chr.9: 132630624-132630624 on Build GRCh38
rs533793755 Chr.9: 132630398-132630398 on Build GRCh38
rs774145543 Chr.9: 132630358-132630358 on Build GRCh38
rs755974339 Chr.9: 132630410-132630410 on Build GRCh38
rs753876186 Chr.9: 132630305-132630305 on Build GRCh38
rs767839862 Chr.9: 132630290-132630290 on Build GRCh38
rs771344843 Chr.9: 132630446-132630446 on Build GRCh38
rs781299305 Chr.9: 132630667-132630667 on Build GRCh38
rs752972843 Chr.9: 132630310-132630310 on Build GRCh38
rs779137730 Chr.9: 132630215-132630215 on Build GRCh38
rs757095425 Chr.9: 132630227-132630227 on Build GRCh38
rs910785962 Chr.9: 132630577-132630577 on Build GRCh38
rs143275845 Chr.9: 132630377-132630377 on Build GRCh38
rs765425773 Chr.9: 132630309-132630309 on Build GRCh38
rs759601011 Chr.9: 132630383-132630383 on Build GRCh38
rs574447114 Chr.9: 132630534-132630534 on Build GRCh38
rs575013252 Chr.9: 132630613-132630613 on Build GRCh38
rs771819641 Chr.9: 132630403-132630406 on Build GRCh38
rs899265007 Chr.9: 132630617-132630617 on Build GRCh38
rs777172373 Chr.9: 132630381-132630381 on Build GRCh38
rs745857827 Chr.9: 132630236-132630236 on Build GRCh38
rs139345088 Chr.9: 132630367-132630367 on Build GRCh38
rs1038638187 Chr.9: 132630306-132630306 on Build GRCh38
rs1042055329 Chr.9: 132630626-132630626 on Build GRCh38
rs368427298 Chr.9: 132630444-132630444 on Build GRCh38
rs1007681170 Chr.9: 132630378-132630378 on Build GRCh38
rs375494425 Chr.9: 132630438-132630438 on Build GRCh38
rs546529762 Chr.9: 132630208-132630208 on Build GRCh38
rs946409773 Chr.9: 132630625-132630625 on Build GRCh38
rs780113834 Chr.9: 132630412-132630412 on Build GRCh38
rs755091161 Chr.9: 132630308-132630308 on Build GRCh38
rs919149580 Chr.9: 132630507-132630507 on Build GRCh38
rs762243465 Chr.9: 132630288-132630288 on Build GRCh38
rs73659425 Chr.9: 132630575-132630575 on Build GRCh38
rs746776535 Chr.9: 132630225-132630225 on Build GRCh38
rs201397560 Chr.9: 132630447-132630447 on Build GRCh38
rs189999986 Chr.9: 132630207-132630207 on Build GRCh38
rs929249826 Chr.9: 132630510-132630510 on Build GRCh38
rs921996022 Chr.9: 132630252-132630252 on Build GRCh38
rs761132557 Chr.9: 132630368-132630368 on Build GRCh38
rs962731975 Chr.9: 132630460-132630460 on Build GRCh38
rs752825220 Chr.9: 132630469-132630469 on Build GRCh38
rs556157017 Chr.9: 132630474-132630474 on Build GRCh38
rs750140971 Chr.9: 132630405-132630405 on Build GRCh38
rs372385164 Chr.9: 132630285-132630285 on Build GRCh38
rs766931553 Chr.9: 132630303-132630303 on Build GRCh38
rs778512898 Chr.9: 132630435-132630435 on Build GRCh38
rs537457236 Chr.9: 132630463-132630463 on Build GRCh38
rs911734892 Chr.9: 132630552-132630552 on Build GRCh38
rs368961280 Chr.9: 132630251-132630251 on Build GRCh38
rs753882010 Chr.9: 132630423-132630423 on Build GRCh38
rs780782677 Chr.9: 132630337-132630337 on Build GRCh38
rs150301518 Chr.9: 132630397-132630397 on Build GRCh38
rs893514103 Chr.9: 132630339-132630339 on Build GRCh38
rs865902634 Chr.9: 132630341-132630341 on Build GRCh38
rs973297149 Chr.9: 132630499-132630499 on Build GRCh38
rs932982044 Chr.9: 132630539-132630541 on Build GRCh38
rs570085819 Chr.9: 132630458-132630458 on Build GRCh38
rs746001561 Chr.9: 132630348-132630348 on Build GRCh38
rs768329211 Chr.9: 132630353-132630353 on Build GRCh38
rs181254770 Chr.9: 132630237-132630237 on Build GRCh38
rs200225130 Chr.9: 132630280-132630280 on Build GRCh38
rs902735849 Chr.9: 132630653-132630653 on Build GRCh38
rs890170845 Chr.9: 132630265-132630265 on Build GRCh38
rs765366527 Chr.9: 132630384-132630384 on Build GRCh38
rs1035602117 Chr.9: 132630456-132630456 on Build GRCh38
rs755069984 Chr.9: 132630352-132630352 on Build GRCh38
rs371246050 Chr.9: 132630428-132630428 on Build GRCh38
rs780168692 Chr.9: 132630349-132630349 on Build GRCh38
rs993918510 Chr.9: 132630427-132630427 on Build GRCh38
rs991306676 Chr.9: 132630461-132630461 on Build GRCh38
rs914318652 Chr.9: 132630468-132630468 on Build GRCh38
rs773002070 Chr.9: 132630417-132630417 on Build GRCh38
rs773634176 Chr.9: 132630296-132630296 on Build GRCh38
rs771584499 Chr.9: 132630441-132630441 on Build GRCh38
rs148854898 Chr.9: 132630336-132630336 on Build GRCh38
rs187073347 Chr.9: 132630276-132630276 on Build GRCh38
rs777522339 Chr.9: 132630442-132630442 on Build GRCh38
rs764399681 Chr.9: 132630402-132630402 on Build GRCh38
rs761486264 Chr.9: 132630213-132630213 on Build GRCh38

View Full Product Details