Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132638180-132638688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX31
Entrez Gene ID
64794
Gene Name
DEAD-box helicase 31
Gene Aliases
PPP1R25
UniGene
Hs.660767
Gene Chromosome Location
Chr.9: 132592997-132670403 on Build GRCh38
Transcripts
NM_001322340.1 NM_001322341.1 NM_001322342.1 NM_001322343.1 NM_022779.8 NM_138620.1 NR_136309.1 XM_005272207.4 XM_006717236.3 XM_011518921.2 XM_011518922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1489756 c.1757G>A p.*586* Chr.9: 132638362-132638362 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935122445 Chr.9: 132638219-132638219 on Build GRCh38
rs780435598 Chr.9: 132638259-132638259 on Build GRCh38
rs917791014 Chr.9: 132638490-132638490 on Build GRCh38
rs36103714 Chr.9: 132638350-132638350 on Build GRCh38
rs534681858 Chr.9: 132638467-132638467 on Build GRCh38
rs997441448 Chr.9: 132638646-132638646 on Build GRCh38
rs976860577 Chr.9: 132638651-132638651 on Build GRCh38
rs1019778322 Chr.9: 132638301-132638301 on Build GRCh38
rs762040037 Chr.9: 132638314-132638314 on Build GRCh38
rs374181405 Chr.9: 132638480-132638480 on Build GRCh38
rs1013262359 Chr.9: 132638539-132638539 on Build GRCh38
rs756850441 Chr.9: 132638358-132638358 on Build GRCh38
rs190731201 Chr.9: 132638275-132638275 on Build GRCh38
rs768368092 Chr.9: 132638414-132638414 on Build GRCh38
rs762809852 Chr.9: 132638497-132638497 on Build GRCh38
rs1028723387 Chr.9: 132638366-132638366 on Build GRCh38
rs139140678 Chr.9: 132638548-132638548 on Build GRCh38
rs925157045 Chr.9: 132638449-132638449 on Build GRCh38
rs549287434 Chr.9: 132638399-132638399 on Build GRCh38
rs979182896 Chr.9: 132638426-132638426 on Build GRCh38
rs796786833 Chr.9: 132638658-132638658 on Build GRCh38
rs761564237 Chr.9: 132638606-132638606 on Build GRCh38
rs537298676 Chr.9: 132638308-132638308 on Build GRCh38
rs551938853 Chr.9: 132638285-132638285 on Build GRCh38
rs1041180460 Chr.9: 132638597-132638597 on Build GRCh38
rs532555098 Chr.9: 132638220-132638220 on Build GRCh38
rs1050316140 Chr.9: 132638484-132638484 on Build GRCh38
rs746200091 Chr.9: 132638384-132638384 on Build GRCh38
rs936826927 Chr.9: 132638295-132638295 on Build GRCh38
rs879268796 Chr.9: 132638371-132638371 on Build GRCh38
rs980985541 Chr.9: 132638280-132638280 on Build GRCh38
rs757844760 Chr.9: 132638349-132638349 on Build GRCh38
rs751041426 Chr.9: 132638357-132638357 on Build GRCh38
rs1006031251 Chr.9: 132638494-132638494 on Build GRCh38
rs376706108 Chr.9: 132638650-132638651 on Build GRCh38
rs749870785 Chr.9: 132638402-132638402 on Build GRCh38
rs567086260 Chr.9: 132638210-132638210 on Build GRCh38
rs569214979 Chr.9: 132638571-132638571 on Build GRCh38
rs944055419 Chr.9: 132638422-132638422 on Build GRCh38
rs574469871 Chr.9: 132638302-132638302 on Build GRCh38
rs112143616 Chr.9: 132638233-132638233 on Build GRCh38
rs771116904 Chr.9: 132638291-132638291 on Build GRCh38
rs767170333 Chr.9: 132638608-132638608 on Build GRCh38
rs144435946 Chr.9: 132638253-132638253 on Build GRCh38
rs1052483989 Chr.9: 132638332-132638332 on Build GRCh38
rs1044846846 Chr.9: 132638538-132638538 on Build GRCh38
rs1045118269 Chr.9: 132638250-132638250 on Build GRCh38
rs368009931 Chr.9: 132638650-132638650 on Build GRCh38
rs78676585 Chr.9: 132638273-132638273 on Build GRCh38
rs775352299 Chr.9: 132638574-132638574 on Build GRCh38
rs374746580 Chr.9: 132638340-132638340 on Build GRCh38
rs969006138 Chr.9: 132638567-132638567 on Build GRCh38
rs73562912 Chr.9: 132638595-132638595 on Build GRCh38
rs765245370 Chr.9: 132638343-132638343 on Build GRCh38
rs1004297468 Chr.9: 132638283-132638283 on Build GRCh38
rs956612599 Chr.9: 132638457-132638457 on Build GRCh38
rs567287556 Chr.9: 132638441-132638441 on Build GRCh38
rs904888337 Chr.9: 132638266-132638266 on Build GRCh38
rs578042765 Chr.9: 132638559-132638559 on Build GRCh38
rs768611724 Chr.9: 132638580-132638580 on Build GRCh38
rs780850797 Chr.9: 132638367-132638367 on Build GRCh38
rs769058304 Chr.9: 132638405-132638405 on Build GRCh38
rs186199937 Chr.9: 132638211-132638211 on Build GRCh38
rs921290112 Chr.9: 132638654-132638654 on Build GRCh38
rs375210059 Chr.9: 132638281-132638281 on Build GRCh38
rs763590483 Chr.9: 132638353-132638353 on Build GRCh38
rs767877776 Chr.9: 132638330-132638330 on Build GRCh38
rs981016781 Chr.9: 132638570-132638570 on Build GRCh38
rs747078988 Chr.9: 132638224-132638224 on Build GRCh38

View Full Product Details