Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133155576-133156113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBGT1
Entrez Gene ID
26301
Gene Name
globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
A3GALNT, FS, UNQ2513
UniGene
Hs.495419
Gene Chromosome Location
Chr.9: 133152948-133163945 on Build GRCh38
Transcripts
NM_001282629.1 NM_001282632.1 NM_001288572.1 NM_001288573.1 NM_021996.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905250 c.256G>A p.A86T Chr.9: 133155906-133155906 on Build GRCh38
6473005 c.189-7G>T p.? Chr.9: 133155943-133155943 on Build GRCh38
5495413 c.176_179delAGCC p.Q59fs*21 Chr.9: 133156024-133156027 on Build GRCh38
3413415 c.148C>A p.L50M Chr.9: 133156055-133156055 on Build GRCh38
5495414 c.213_216delAGCC p.A72fs*70 Chr.9: 133156024-133156027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530378627 Chr.9: 133155792-133155792 on Build GRCh38
rs376647799 Chr.9: 133156037-133156037 on Build GRCh38
rs951395534 Chr.9: 133155731-133155731 on Build GRCh38
rs1035615052 Chr.9: 133156035-133156036 on Build GRCh38
rs774409259 Chr.9: 133156089-133156089 on Build GRCh38
rs981327390 Chr.9: 133155997-133155997 on Build GRCh38
rs368926549 Chr.9: 133155950-133155950 on Build GRCh38
rs1013186161 Chr.9: 133155690-133155690 on Build GRCh38
rs777253584 Chr.9: 133155871-133155871 on Build GRCh38
rs75456737 Chr.9: 133156038-133156038 on Build GRCh38
rs962638926 Chr.9: 133155681-133155681 on Build GRCh38
rs775740988 Chr.9: 133155981-133155981 on Build GRCh38
rs199608295 Chr.9: 133155984-133155984 on Build GRCh38
rs35906929 Chr.9: 133155983-133155983 on Build GRCh38
rs747444889 Chr.9: 133155868-133155868 on Build GRCh38
rs373328427 Chr.9: 133155936-133155936 on Build GRCh38
rs746629735 Chr.9: 133155964-133155964 on Build GRCh38
rs548646335 Chr.9: 133155805-133155805 on Build GRCh38
rs139796375 Chr.9: 133156025-133156025 on Build GRCh38
rs796310350 Chr.9: 133155958-133155959 on Build GRCh38
rs776227646 Chr.9: 133156005-133156005 on Build GRCh38
rs575167551 Chr.9: 133155624-133155624 on Build GRCh38
rs898988141 Chr.9: 133155704-133155704 on Build GRCh38
rs370033002 Chr.9: 133155979-133155979 on Build GRCh38
rs1006588964 Chr.9: 133155640-133155640 on Build GRCh38
rs77339890 Chr.9: 133155700-133155700 on Build GRCh38
rs777534578 Chr.9: 133155963-133155963 on Build GRCh38
rs973875638 Chr.9: 133155694-133155694 on Build GRCh38
rs753878697 Chr.9: 133155929-133155929 on Build GRCh38
rs1055065585 Chr.9: 133155789-133155789 on Build GRCh38
rs532613368 Chr.9: 133155759-133155759 on Build GRCh38
rs780755250 Chr.9: 133156040-133156040 on Build GRCh38
rs763509041 Chr.9: 133156023-133156023 on Build GRCh38
rs749796330 Chr.9: 133156062-133156062 on Build GRCh38
rs145977218 Chr.9: 133155768-133155768 on Build GRCh38
rs563181112 Chr.9: 133155781-133155781 on Build GRCh38
rs772554707 Chr.9: 133155980-133155980 on Build GRCh38
rs759504776 Chr.9: 133156016-133156016 on Build GRCh38
rs565508993 Chr.9: 133155750-133155750 on Build GRCh38
rs539929626 Chr.9: 133155742-133155742 on Build GRCh38
rs202127534 Chr.9: 133155770-133155770 on Build GRCh38
rs775652901 Chr.9: 133155882-133155882 on Build GRCh38
rs542218310 Chr.9: 133155651-133155651 on Build GRCh38
rs746391395 Chr.9: 133155974-133155974 on Build GRCh38
rs749419341 Chr.9: 133155982-133155982 on Build GRCh38
rs778031991 Chr.9: 133155867-133155867 on Build GRCh38
rs73660458 Chr.9: 133155958-133155958 on Build GRCh38
rs755730083 Chr.9: 133156034-133156034 on Build GRCh38
rs201026152 Chr.9: 133156031-133156031 on Build GRCh38
rs187297599 Chr.9: 133155697-133155697 on Build GRCh38
rs763288546 Chr.9: 133155905-133155905 on Build GRCh38
rs771665971 Chr.9: 133155862-133155862 on Build GRCh38
rs773627027 Chr.9: 133155730-133155730 on Build GRCh38
rs747800086 Chr.9: 133155860-133155860 on Build GRCh38
rs764370932 Chr.9: 133155907-133155907 on Build GRCh38
rs955014823 Chr.9: 133155639-133155639 on Build GRCh38
rs766197789 Chr.9: 133156001-133156001 on Build GRCh38
rs975997466 Chr.9: 133155828-133155828 on Build GRCh38
rs761932271 Chr.9: 133155990-133155990 on Build GRCh38
rs768475752 Chr.9: 133155862-133155862 on Build GRCh38
rs371901748 Chr.9: 133155870-133155870 on Build GRCh38
rs374147391 Chr.9: 133156022-133156022 on Build GRCh38
rs2073927 Chr.9: 133155959-133155959 on Build GRCh38
rs921461786 Chr.9: 133155832-133155832 on Build GRCh38
rs994781044 Chr.9: 133155724-133155724 on Build GRCh38
rs560599170 Chr.9: 133155625-133155625 on Build GRCh38
rs781345868 Chr.9: 133155973-133155973 on Build GRCh38
rs150605526 Chr.9: 133156019-133156019 on Build GRCh38
rs746374893 Chr.9: 133155874-133155874 on Build GRCh38
rs117595304 Chr.9: 133155928-133155928 on Build GRCh38
rs193135740 Chr.9: 133155877-133155877 on Build GRCh38
rs1051273164 Chr.9: 133155900-133155900 on Build GRCh38
rs778789902 Chr.9: 133156088-133156088 on Build GRCh38
rs370102294 Chr.9: 133155927-133155927 on Build GRCh38
rs757706490 Chr.9: 133155967-133155967 on Build GRCh38
rs750121195 Chr.9: 133156024-133156024 on Build GRCh38
rs990746579 Chr.9: 133156048-133156048 on Build GRCh38
rs770373664 Chr.9: 133155980-133155980 on Build GRCh38
rs377538663 Chr.9: 133155966-133155966 on Build GRCh38
rs774404360 Chr.9: 133155908-133155908 on Build GRCh38
rs778387138 Chr.9: 133155854-133155854 on Build GRCh38
rs1004180639 Chr.9: 133155740-133155740 on Build GRCh38
rs1057293998 Chr.9: 133155648-133155648 on Build GRCh38
rs572933304 Chr.9: 133155723-133155723 on Build GRCh38
rs867808470 Chr.9: 133155652-133155652 on Build GRCh38
rs766608683 Chr.9: 133155906-133155906 on Build GRCh38
rs547031073 Chr.9: 133155627-133155627 on Build GRCh38
rs768843054 Chr.9: 133156078-133156078 on Build GRCh38
rs968838891 Chr.9: 133155834-133155834 on Build GRCh38

View Full Product Details