Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154445692-154446233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50A
Entrez Gene ID
9130
Gene Name
family with sequence similarity 50 member A
Gene Aliases
9F, DXS9928E, HXC-26, HXC26, XAP5
UniGene
Hs.54277
Gene Chromosome Location
Chr.X: 154444126-154450654 on Build GRCh38
Transcripts
NM_004699.3

SNP Detail

SNP ID Chromosome Location
rs782492408 Chr.X: 154445770-154445770 on Build GRCh38
rs1034673524 Chr.X: 154445974-154445974 on Build GRCh38
rs782053855 Chr.X: 154445864-154445864 on Build GRCh38
rs782813545 Chr.X: 154445828-154445828 on Build GRCh38
rs782175770 Chr.X: 154445927-154445927 on Build GRCh38
rs781954950 Chr.X: 154446197-154446197 on Build GRCh38
rs782253943 Chr.X: 154445956-154445956 on Build GRCh38
rs200788225 Chr.X: 154445746-154445746 on Build GRCh38
rs782527911 Chr.X: 154445924-154445924 on Build GRCh38
rs781785055 Chr.X: 154445776-154445776 on Build GRCh38
rs782449565 Chr.X: 154445799-154445799 on Build GRCh38
rs782610296 Chr.X: 154446065-154446065 on Build GRCh38
rs183908507 Chr.X: 154445998-154445998 on Build GRCh38
rs782224227 Chr.X: 154445792-154445792 on Build GRCh38
rs370092930 Chr.X: 154445789-154445789 on Build GRCh38
rs781860807 Chr.X: 154445756-154445756 on Build GRCh38
rs782124643 Chr.X: 154446147-154446147 on Build GRCh38
rs367875244 Chr.X: 154445923-154445923 on Build GRCh38
rs782245650 Chr.X: 154446077-154446077 on Build GRCh38
rs781792574 Chr.X: 154445769-154445769 on Build GRCh38
rs781902781 Chr.X: 154445964-154445964 on Build GRCh38
rs781942257 Chr.X: 154445846-154445846 on Build GRCh38
rs910790917 Chr.X: 154446076-154446076 on Build GRCh38
rs781962345 Chr.X: 154445743-154445743 on Build GRCh38
rs1036307452 Chr.X: 154445747-154445747 on Build GRCh38
rs782158349 Chr.X: 154445731-154445731 on Build GRCh38
rs200076498 Chr.X: 154445765-154445765 on Build GRCh38
rs782676382 Chr.X: 154446061-154446061 on Build GRCh38
rs782425221 Chr.X: 154445963-154445963 on Build GRCh38
rs1024422203 Chr.X: 154446012-154446012 on Build GRCh38
rs782164633 Chr.X: 154445817-154445817 on Build GRCh38
rs782030673 Chr.X: 154446152-154446152 on Build GRCh38
rs73638283 Chr.X: 154446068-154446068 on Build GRCh38
rs147042000 Chr.X: 154445713-154445713 on Build GRCh38
rs1040917526 Chr.X: 154446128-154446128 on Build GRCh38
rs782337282 Chr.X: 154445796-154445796 on Build GRCh38
rs782204207 Chr.X: 154446040-154446040 on Build GRCh38
rs782390406 Chr.X: 154446151-154446151 on Build GRCh38
rs377764840 Chr.X: 154445730-154445730 on Build GRCh38
rs1000080652 Chr.X: 154445971-154445971 on Build GRCh38
rs116166090 Chr.X: 154446064-154446064 on Build GRCh38
rs917319596 Chr.X: 154445745-154445745 on Build GRCh38
rs781826792 Chr.X: 154445894-154445894 on Build GRCh38
rs944923506 Chr.X: 154446117-154446117 on Build GRCh38
rs782751834 Chr.X: 154445915-154445915 on Build GRCh38
rs782759848 Chr.X: 154445753-154445753 on Build GRCh38
rs782523141 Chr.X: 154445911-154445911 on Build GRCh38
rs782482972 Chr.X: 154445943-154445943 on Build GRCh38
rs900942800 Chr.X: 154446135-154446135 on Build GRCh38
rs374036563 Chr.X: 154445873-154445873 on Build GRCh38
rs782588013 Chr.X: 154445926-154445926 on Build GRCh38
rs782597125 Chr.X: 154445955-154445955 on Build GRCh38
rs146728107 Chr.X: 154445766-154445766 on Build GRCh38
rs781913922 Chr.X: 154445790-154445790 on Build GRCh38
rs367635365 Chr.X: 154445783-154445783 on Build GRCh38
rs782604470 Chr.X: 154445773-154445773 on Build GRCh38
rs782181197 Chr.X: 154446169-154446169 on Build GRCh38
rs782042773 Chr.X: 154446072-154446072 on Build GRCh38
rs782255264 Chr.X: 154445775-154445775 on Build GRCh38
rs782639797 Chr.X: 154445991-154445991 on Build GRCh38
rs889254552 Chr.X: 154445798-154445798 on Build GRCh38
rs376540647 Chr.X: 154446056-154446056 on Build GRCh38
rs782277066 Chr.X: 154446060-154446060 on Build GRCh38
rs782395187 Chr.X: 154446167-154446167 on Build GRCh38
rs782659661 Chr.X: 154445940-154445940 on Build GRCh38
rs958605504 Chr.X: 154445986-154445986 on Build GRCh38
rs369954132 Chr.X: 154446211-154446211 on Build GRCh38

View Full Product Details