Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154511756-154512109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3A
Entrez Gene ID
60343
Gene Name
family with sequence similarity 3 member A
Gene Aliases
2.19, DLD, DXS560S, XAP-7
UniGene
Hs.289108
Gene Chromosome Location
Chr.X: 154506159-154516249 on Build GRCh38
Transcripts
NM_001171132.2 NM_001171133.2 NM_001171134.2 NM_001282311.1 NM_001282312.1 NM_021806.3 XM_005274714.3 XM_005274716.3 XM_005277879.4 XM_006724831.2 XM_006724832.3 XM_006724833.2 XM_006724834.2 XM_011531185.2 XM_017029699.1 XM_017029700.1 XM_017029701.1 XM_017029702.1

SNP Detail

SNP ID Chromosome Location
rs782696977 Chr.X: 154511988-154511988 on Build GRCh38
rs147214885 Chr.X: 154511785-154511785 on Build GRCh38
rs185585368 Chr.X: 154512002-154512002 on Build GRCh38
rs782709383 Chr.X: 154511922-154511922 on Build GRCh38
rs782199811 Chr.X: 154511858-154511858 on Build GRCh38
rs782245221 Chr.X: 154511948-154511948 on Build GRCh38
rs782019548 Chr.X: 154511968-154511968 on Build GRCh38
rs781969873 Chr.X: 154511884-154511884 on Build GRCh38
rs1052431673 Chr.X: 154511911-154511911 on Build GRCh38
rs1052055625 Chr.X: 154511804-154511804 on Build GRCh38
rs782273686 Chr.X: 154511834-154511834 on Build GRCh38
rs782530487 Chr.X: 154511784-154511784 on Build GRCh38
rs782119310 Chr.X: 154511905-154511905 on Build GRCh38
rs935454242 Chr.X: 154511859-154511859 on Build GRCh38
rs371947699 Chr.X: 154511816-154511816 on Build GRCh38
rs1005040791 Chr.X: 154511985-154511985 on Build GRCh38
rs782431757 Chr.X: 154511844-154511844 on Build GRCh38
rs200583876 Chr.X: 154511921-154511921 on Build GRCh38
rs1003787848 Chr.X: 154511965-154511965 on Build GRCh38
rs782075598 Chr.X: 154512054-154512054 on Build GRCh38
rs782202107 Chr.X: 154511794-154511794 on Build GRCh38
rs145680913 Chr.X: 154511868-154511868 on Build GRCh38
rs372889876 Chr.X: 154511920-154511920 on Build GRCh38
rs921816959 Chr.X: 154511791-154511791 on Build GRCh38
rs782385334 Chr.X: 154511949-154511949 on Build GRCh38
rs371419000 Chr.X: 154511912-154511912 on Build GRCh38
rs782164443 Chr.X: 154511984-154511984 on Build GRCh38
rs782243962 Chr.X: 154511837-154511837 on Build GRCh38
rs781816981 Chr.X: 154511923-154511923 on Build GRCh38
rs782657572 Chr.X: 154511825-154511825 on Build GRCh38
rs973040227 Chr.X: 154512073-154512073 on Build GRCh38
rs374780401 Chr.X: 154511845-154511845 on Build GRCh38

View Full Product Details