Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154512738-154513219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3A
Entrez Gene ID
60343
Gene Name
family with sequence similarity 3 member A
Gene Aliases
2.19, DLD, DXS560S, XAP-7
UniGene
Hs.289108
Gene Chromosome Location
Chr.X: 154506159-154516249 on Build GRCh38
Transcripts
NM_001171132.2 NM_001171133.2 NM_001171134.2 NM_001282311.1 NM_001282312.1 NM_021806.3 XM_005274714.3 XM_005274716.3 XM_005277879.4 XM_006724831.2 XM_006724832.3 XM_006724833.2 XM_006724834.2 XM_011531185.2 XM_017029699.1 XM_017029700.1 XM_017029701.1 XM_017029702.1

SNP Detail

SNP ID Chromosome Location
rs781787283 Chr.X: 154513100-154513100 on Build GRCh38
rs181188384 Chr.X: 154512910-154512910 on Build GRCh38
rs782325572 Chr.X: 154512767-154512767 on Build GRCh38
rs782017805 Chr.X: 154512928-154512928 on Build GRCh38
rs782134864 Chr.X: 154512973-154512973 on Build GRCh38
rs781850082 Chr.X: 154513113-154513113 on Build GRCh38
rs782046134 Chr.X: 154512858-154512858 on Build GRCh38
rs1022126051 Chr.X: 154513196-154513196 on Build GRCh38
rs782020400 Chr.X: 154512955-154512955 on Build GRCh38
rs782771596 Chr.X: 154513018-154513018 on Build GRCh38
rs782765703 Chr.X: 154512799-154512799 on Build GRCh38
rs148149959 Chr.X: 154512885-154512885 on Build GRCh38
rs1008541107 Chr.X: 154513106-154513106 on Build GRCh38
rs1019010120 Chr.X: 154513117-154513117 on Build GRCh38
rs782469898 Chr.X: 154513131-154513131 on Build GRCh38
rs782332239 Chr.X: 154512905-154512905 on Build GRCh38
rs781926880 Chr.X: 154512797-154512797 on Build GRCh38
rs202125428 Chr.X: 154512896-154512896 on Build GRCh38
rs201768658 Chr.X: 154512807-154512807 on Build GRCh38
rs782164224 Chr.X: 154512935-154512935 on Build GRCh38
rs376012862 Chr.X: 154512981-154512981 on Build GRCh38
rs782441606 Chr.X: 154512788-154512788 on Build GRCh38
rs781984027 Chr.X: 154512830-154512830 on Build GRCh38
rs371195332 Chr.X: 154512980-154512980 on Build GRCh38
rs377098720 Chr.X: 154512786-154512786 on Build GRCh38
rs782692210 Chr.X: 154512893-154512893 on Build GRCh38
rs377424756 Chr.X: 154512982-154512982 on Build GRCh38
rs377755058 Chr.X: 154512947-154512947 on Build GRCh38
rs782074773 Chr.X: 154512806-154512806 on Build GRCh38
rs782306026 Chr.X: 154513002-154513002 on Build GRCh38
rs782638937 Chr.X: 154512886-154512886 on Build GRCh38
rs782791470 Chr.X: 154512856-154512856 on Build GRCh38
rs782453375 Chr.X: 154513103-154513103 on Build GRCh38
rs200730944 Chr.X: 154512843-154512843 on Build GRCh38
rs988013144 Chr.X: 154513149-154513149 on Build GRCh38
rs35985994 Chr.X: 154512887-154512887 on Build GRCh38
rs939331269 Chr.X: 154512778-154512778 on Build GRCh38
rs781899769 Chr.X: 154512857-154512857 on Build GRCh38
rs782386917 Chr.X: 154512927-154512927 on Build GRCh38
rs782180409 Chr.X: 154512804-154512804 on Build GRCh38
rs782236332 Chr.X: 154512919-154512919 on Build GRCh38
rs940235574 Chr.X: 154512824-154512824 on Build GRCh38
rs375008708 Chr.X: 154513174-154513174 on Build GRCh38
rs782487249 Chr.X: 154512878-154512878 on Build GRCh38
rs782312403 Chr.X: 154512940-154512940 on Build GRCh38
rs781822318 Chr.X: 154512869-154512869 on Build GRCh38
rs782082955 Chr.X: 154512974-154512974 on Build GRCh38
rs782726344 Chr.X: 154512859-154512859 on Build GRCh38
rs141040279 Chr.X: 154512888-154512888 on Build GRCh38

View Full Product Details