Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154531616-154532115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PD
Entrez Gene ID
2539
Gene Name
glucose-6-phosphate dehydrogenase
Gene Aliases
G6PD1
UniGene
Hs.461047
Gene Chromosome Location
Chr.X: 154531390-154547586 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2

SNP Detail

SNP ID Chromosome Location
rs782648605 Chr.X: 154531862-154531862 on Build GRCh38
rs1034742794 Chr.X: 154531635-154531635 on Build GRCh38
rs782082882 Chr.X: 154531946-154531946 on Build GRCh38
rs782351135 Chr.X: 154532079-154532079 on Build GRCh38
rs781857931 Chr.X: 154531912-154531912 on Build GRCh38
rs782513228 Chr.X: 154531997-154531997 on Build GRCh38
rs782140549 Chr.X: 154531962-154531962 on Build GRCh38
rs781788462 Chr.X: 154531980-154531980 on Build GRCh38
rs975719422 Chr.X: 154531719-154531719 on Build GRCh38
rs955205814 Chr.X: 154531798-154531798 on Build GRCh38
rs782371841 Chr.X: 154531840-154531840 on Build GRCh38
rs398123543 Chr.X: 154531953-154531953 on Build GRCh38
rs782600380 Chr.X: 154531990-154531990 on Build GRCh38
rs1003923823 Chr.X: 154531677-154531677 on Build GRCh38
rs782799512 Chr.X: 154531963-154531963 on Build GRCh38
rs1057515821 Chr.X: 154531635-154531635 on Build GRCh38
rs782817663 Chr.X: 154531979-154531979 on Build GRCh38
rs782681737 Chr.X: 154531658-154531658 on Build GRCh38
rs781913124 Chr.X: 154531964-154531964 on Build GRCh38
rs934738398 Chr.X: 154531830-154531830 on Build GRCh38
rs782020659 Chr.X: 154531955-154531955 on Build GRCh38
rs111776132 Chr.X: 154531938-154531938 on Build GRCh38
rs914543891 Chr.X: 154531846-154531846 on Build GRCh38
rs781866772 Chr.X: 154531975-154531975 on Build GRCh38
rs907435124 Chr.X: 154531924-154531924 on Build GRCh38
rs782094249 Chr.X: 154531969-154531969 on Build GRCh38
rs112950723 Chr.X: 154531728-154531728 on Build GRCh38
rs782643722 Chr.X: 154532009-154532009 on Build GRCh38
rs782455613 Chr.X: 154531986-154531986 on Build GRCh38
rs782605257 Chr.X: 154531718-154531718 on Build GRCh38
rs1017005312 Chr.X: 154531691-154531691 on Build GRCh38
rs781923155 Chr.X: 154532063-154532063 on Build GRCh38
rs201294737 Chr.X: 154531950-154531950 on Build GRCh38
rs191319738 Chr.X: 154531711-154531711 on Build GRCh38
rs111485003 Chr.X: 154531696-154531696 on Build GRCh38
rs1050757 Chr.X: 154531643-154531643 on Build GRCh38
rs986597786 Chr.X: 154531814-154531814 on Build GRCh38
rs147551350 Chr.X: 154531869-154531869 on Build GRCh38
rs782143571 Chr.X: 154531667-154531667 on Build GRCh38
rs782206043 Chr.X: 154532078-154532078 on Build GRCh38
rs921159359 Chr.X: 154531766-154531766 on Build GRCh38
rs782745892 Chr.X: 154531974-154531974 on Build GRCh38
rs782233788 Chr.X: 154531991-154531991 on Build GRCh38
rs782522167 Chr.X: 154531978-154531978 on Build GRCh38
rs1047844230 Chr.X: 154531930-154531930 on Build GRCh38
rs782397732 Chr.X: 154531954-154531954 on Build GRCh38
rs938857639 Chr.X: 154531925-154531925 on Build GRCh38
rs781935516 Chr.X: 154531935-154531935 on Build GRCh38
rs368255747 Chr.X: 154531951-154531951 on Build GRCh38
rs375285369 Chr.X: 154532031-154532031 on Build GRCh38
rs782422580 Chr.X: 154532055-154532055 on Build GRCh38
rs781784222 Chr.X: 154531682-154531682 on Build GRCh38
rs782224094 Chr.X: 154531824-154531824 on Build GRCh38

View Full Product Details