Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154652038-154652526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAG2
Entrez Gene ID
30848
Gene Name
cancer/testis antigen 2
Gene Aliases
CAMEL, CT2, CT6.2, CT6.2a, CT6.2b, ESO2, LAGE-1, LAGE2B
UniGene
Hs.713312
Gene Chromosome Location
Chr.X: 154651972-154653579 on Build GRCh38
Transcripts
NM_020994.4 NM_172377.4

SNP Detail

SNP ID Chromosome Location
rs112780678 Chr.X: 154652295-154652295 on Build GRCh38
rs782434846 Chr.X: 154652176-154652176 on Build GRCh38
rs199533324 Chr.X: 154652429-154652429 on Build GRCh38
rs201803251 Chr.X: 154652152-154652152 on Build GRCh38
rs782759822 Chr.X: 154652113-154652113 on Build GRCh38
rs782075668 Chr.X: 154652116-154652116 on Build GRCh38
rs199885620 Chr.X: 154652338-154652338 on Build GRCh38
rs781822611 Chr.X: 154652183-154652183 on Build GRCh38
rs113459988 Chr.X: 154652360-154652360 on Build GRCh38
rs781989911 Chr.X: 154652213-154652213 on Build GRCh38
rs782641911 Chr.X: 154652419-154652419 on Build GRCh38
rs781923494 Chr.X: 154652347-154652347 on Build GRCh38
rs782394278 Chr.X: 154652492-154652492 on Build GRCh38
rs782273548 Chr.X: 154652337-154652337 on Build GRCh38
rs782134176 Chr.X: 154652450-154652450 on Build GRCh38
rs147124424 Chr.X: 154652306-154652306 on Build GRCh38
rs782495026 Chr.X: 154652481-154652481 on Build GRCh38
rs146270536 Chr.X: 154652221-154652221 on Build GRCh38
rs782232934 Chr.X: 154652506-154652506 on Build GRCh38
rs782535427 Chr.X: 154652326-154652326 on Build GRCh38
rs374234710 Chr.X: 154652488-154652488 on Build GRCh38
rs781785685 Chr.X: 154652469-154652469 on Build GRCh38
rs151065825 Chr.X: 154652244-154652244 on Build GRCh38
rs782427536 Chr.X: 154652086-154652086 on Build GRCh38
rs782632339 Chr.X: 154652499-154652499 on Build GRCh38
rs782342647 Chr.X: 154652507-154652507 on Build GRCh38
rs781895845 Chr.X: 154652416-154652416 on Build GRCh38
rs782563025 Chr.X: 154652406-154652406 on Build GRCh38
rs781869752 Chr.X: 154652166-154652166 on Build GRCh38
rs782130741 Chr.X: 154652257-154652257 on Build GRCh38
rs782747935 Chr.X: 154652444-154652444 on Build GRCh38
rs782753339 Chr.X: 154652383-154652383 on Build GRCh38
rs782359239 Chr.X: 154652422-154652422 on Build GRCh38
rs143422691 Chr.X: 154652405-154652405 on Build GRCh38
rs782664059 Chr.X: 154652161-154652161 on Build GRCh38
rs782196572 Chr.X: 154652173-154652173 on Build GRCh38
rs782502444 Chr.X: 154652167-154652167 on Build GRCh38
rs782621780 Chr.X: 154652482-154652482 on Build GRCh38
rs782786808 Chr.X: 154652369-154652369 on Build GRCh38
rs200456175 Chr.X: 154652417-154652417 on Build GRCh38
rs147744363 Chr.X: 154652336-154652336 on Build GRCh38
rs782160090 Chr.X: 154652476-154652476 on Build GRCh38
rs782597963 Chr.X: 154652168-154652168 on Build GRCh38
rs267606399 Chr.X: 154652186-154652186 on Build GRCh38
rs782096932 Chr.X: 154652344-154652344 on Build GRCh38
rs144179580 Chr.X: 154652240-154652240 on Build GRCh38
rs781870770 Chr.X: 154652480-154652480 on Build GRCh38
rs782790185 Chr.X: 154652477-154652477 on Build GRCh38
rs201129178 Chr.X: 154652309-154652309 on Build GRCh38
rs781826758 Chr.X: 154652266-154652266 on Build GRCh38
rs782087117 Chr.X: 154652468-154652468 on Build GRCh38
rs782448909 Chr.X: 154652397-154652397 on Build GRCh38
rs782024343 Chr.X: 154652090-154652090 on Build GRCh38
rs781784571 Chr.X: 154652133-154652133 on Build GRCh38
rs370153454 Chr.X: 154652123-154652123 on Build GRCh38
rs372538197 Chr.X: 154652390-154652390 on Build GRCh38
rs374957448 Chr.X: 154652292-154652292 on Build GRCh38
rs781819203 Chr.X: 154652484-154652484 on Build GRCh38
rs782123123 Chr.X: 154652422-154652422 on Build GRCh38
rs139568434 Chr.X: 154652237-154652237 on Build GRCh38
rs782649630 Chr.X: 154652330-154652330 on Build GRCh38
rs782347991 Chr.X: 154652343-154652343 on Build GRCh38
rs781834317 Chr.X: 154652115-154652115 on Build GRCh38
rs782139896 Chr.X: 154652103-154652103 on Build GRCh38
rs782251849 Chr.X: 154652302-154652302 on Build GRCh38
rs782019457 Chr.X: 154652252-154652252 on Build GRCh38
rs782186886 Chr.X: 154652380-154652380 on Build GRCh38
rs782240656 Chr.X: 154652421-154652421 on Build GRCh38
rs781972947 Chr.X: 154652464-154652464 on Build GRCh38
rs145829050 Chr.X: 154652284-154652284 on Build GRCh38
rs148011170 Chr.X: 154652439-154652439 on Build GRCh38
rs782686361 Chr.X: 154652489-154652489 on Build GRCh38
rs143520716 Chr.X: 154652203-154652203 on Build GRCh38
rs368057025 Chr.X: 154652379-154652379 on Build GRCh38
rs782717473 Chr.X: 154652346-154652346 on Build GRCh38
rs782424285 Chr.X: 154652433-154652433 on Build GRCh38
rs782288697 Chr.X: 154652200-154652200 on Build GRCh38
rs782235402 Chr.X: 154652227-154652227 on Build GRCh38
rs782192001 Chr.X: 154652324-154652324 on Build GRCh38
rs782386555 Chr.X: 154652461-154652461 on Build GRCh38
rs149831151 Chr.X: 154652259-154652259 on Build GRCh38

View Full Product Details