Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154830637-154831136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM9
Entrez Gene ID
100132963
Gene Name
small integral membrane protein 9
Gene Aliases
CXorf68
UniGene
Hs.220762
Gene Chromosome Location
Chr.X: 154823348-154834662 on Build GRCh38
Transcripts
NM_001162936.1

SNP Detail

SNP ID Chromosome Location
rs782490191 Chr.X: 154831090-154831090 on Build GRCh38
rs376427110 Chr.X: 154830864-154830864 on Build GRCh38
rs1009775668 Chr.X: 154830913-154830913 on Build GRCh38
rs980655689 Chr.X: 154831008-154831008 on Build GRCh38
rs1056125197 Chr.X: 154830669-154830669 on Build GRCh38
rs192303481 Chr.X: 154830732-154830732 on Build GRCh38
rs188397606 Chr.X: 154830906-154830906 on Build GRCh38
rs782020888 Chr.X: 154831048-154831048 on Build GRCh38
rs782443981 Chr.X: 154830818-154830818 on Build GRCh38
rs782048065 Chr.X: 154830761-154830761 on Build GRCh38
rs184141613 Chr.X: 154830770-154830770 on Build GRCh38
rs199975971 Chr.X: 154830724-154830724 on Build GRCh38
rs940159688 Chr.X: 154830717-154830717 on Build GRCh38
rs782518347 Chr.X: 154830688-154830688 on Build GRCh38
rs891112446 Chr.X: 154830853-154830853 on Build GRCh38
rs1022136573 Chr.X: 154830947-154830947 on Build GRCh38
rs782758076 Chr.X: 154831058-154831058 on Build GRCh38
rs782602739 Chr.X: 154831104-154831104 on Build GRCh38
rs73247664 Chr.X: 154831068-154831068 on Build GRCh38
rs781850793 Chr.X: 154830899-154830899 on Build GRCh38
rs12009271 Chr.X: 154830834-154830834 on Build GRCh38
rs782295828 Chr.X: 154830985-154830985 on Build GRCh38
rs782656009 Chr.X: 154830903-154830903 on Build GRCh38
rs886873382 Chr.X: 154830712-154830712 on Build GRCh38
rs782165440 Chr.X: 154831049-154831049 on Build GRCh38
rs991802685 Chr.X: 154831063-154831063 on Build GRCh38
rs782266601 Chr.X: 154830861-154830861 on Build GRCh38
rs939791974 Chr.X: 154831073-154831073 on Build GRCh38

View Full Product Details