Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154860191-154860707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F8
Entrez Gene ID
2157
Gene Name
coagulation factor VIII
Gene Aliases
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
UniGene
Hs.632836
Gene Chromosome Location
Chr.X: 154835788-155022723 on Build GRCh38
Transcripts
NM_000132.3 NM_019863.2

SNP Detail

SNP ID Chromosome Location
rs988099238 Chr.X: 154860500-154860500 on Build GRCh38
rs782479558 Chr.X: 154860580-154860580 on Build GRCh38
rs895445033 Chr.X: 154860272-154860272 on Build GRCh38
rs782539098 Chr.X: 154860291-154860291 on Build GRCh38
rs145813556 Chr.X: 154860347-154860347 on Build GRCh38
rs782654096 Chr.X: 154860608-154860608 on Build GRCh38
rs781846388 Chr.X: 154860444-154860444 on Build GRCh38
rs782012093 Chr.X: 154860616-154860616 on Build GRCh38
rs782496606 Chr.X: 154860394-154860395 on Build GRCh38
rs782792257 Chr.X: 154860414-154860414 on Build GRCh38
rs781799682 Chr.X: 154860604-154860604 on Build GRCh38
rs137852470 Chr.X: 154860538-154860538 on Build GRCh38
rs387906465 Chr.X: 154860537-154860538 on Build GRCh38
rs372502385 Chr.X: 154860398-154860398 on Build GRCh38
rs144069929 Chr.X: 154860269-154860269 on Build GRCh38
rs782291195 Chr.X: 154860628-154860628 on Build GRCh38
rs782541492 Chr.X: 154860389-154860390 on Build GRCh38
rs137852471 Chr.X: 154860467-154860467 on Build GRCh38
rs186375692 Chr.X: 154860656-154860656 on Build GRCh38
rs782557042 Chr.X: 154860518-154860518 on Build GRCh38
rs371791984 Chr.X: 154860647-154860647 on Build GRCh38
rs973162043 Chr.X: 154860363-154860363 on Build GRCh38
rs1014569441 Chr.X: 154860322-154860322 on Build GRCh38
rs782207803 Chr.X: 154860402-154860402 on Build GRCh38
rs782212661 Chr.X: 154860327-154860327 on Build GRCh38
rs782063102 Chr.X: 154860410-154860410 on Build GRCh38
rs782766041 Chr.X: 154860390-154860390 on Build GRCh38
rs781920027 Chr.X: 154860406-154860406 on Build GRCh38
rs781799587 Chr.X: 154860364-154860364 on Build GRCh38
rs139348729 Chr.X: 154860461-154860461 on Build GRCh38
rs782354758 Chr.X: 154860405-154860405 on Build GRCh38
rs782717799 Chr.X: 154860497-154860497 on Build GRCh38
rs781890169 Chr.X: 154860514-154860514 on Build GRCh38
rs28370229 Chr.X: 154860249-154860249 on Build GRCh38
rs782608496 Chr.X: 154860584-154860584 on Build GRCh38
rs905701703 Chr.X: 154860238-154860238 on Build GRCh38
rs377550329 Chr.X: 154860606-154860606 on Build GRCh38
rs782290071 Chr.X: 154860388-154860388 on Build GRCh38
rs112699109 Chr.X: 154860566-154860566 on Build GRCh38
rs782370974 Chr.X: 154860390-154860390 on Build GRCh38
rs782107575 Chr.X: 154860418-154860418 on Build GRCh38
rs137852469 Chr.X: 154860588-154860588 on Build GRCh38
rs782027402 Chr.X: 154860416-154860416 on Build GRCh38
rs782729286 Chr.X: 154860433-154860433 on Build GRCh38
rs371143627 Chr.X: 154860643-154860643 on Build GRCh38
rs1800297 Chr.X: 154860563-154860563 on Build GRCh38

View Full Product Details