Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155054828-155055343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUNDC2
Entrez Gene ID
65991
Gene Name
FUN14 domain containing 2
Gene Aliases
DC44, HCBP6, HCC3, PD03104
UniGene
Hs.356050
Gene Chromosome Location
Chr.X: 155026789-155056916 on Build GRCh38
Transcripts
NM_023934.3

SNP Detail

SNP ID Chromosome Location
rs902563813 Chr.X: 155054969-155054969 on Build GRCh38
rs1022925747 Chr.X: 155055092-155055092 on Build GRCh38
rs1007780602 Chr.X: 155054907-155054907 on Build GRCh38
rs782137184 Chr.X: 155054896-155054896 on Build GRCh38
rs185529445 Chr.X: 155054983-155054983 on Build GRCh38
rs189192040 Chr.X: 155055116-155055116 on Build GRCh38
rs781816032 Chr.X: 155055131-155055131 on Build GRCh38
rs781900427 Chr.X: 155055191-155055191 on Build GRCh38
rs909369292 Chr.X: 155055237-155055237 on Build GRCh38
rs1039290129 Chr.X: 155054918-155054918 on Build GRCh38
rs782090496 Chr.X: 155055019-155055019 on Build GRCh38
rs1049188247 Chr.X: 155054895-155054895 on Build GRCh38
rs781801337 Chr.X: 155055245-155055245 on Build GRCh38
rs981069423 Chr.X: 155055127-155055127 on Build GRCh38
rs991411197 Chr.X: 155055055-155055055 on Build GRCh38
rs782024044 Chr.X: 155054928-155054928 on Build GRCh38
rs1020988316 Chr.X: 155054965-155054965 on Build GRCh38
rs782798751 Chr.X: 155054952-155054952 on Build GRCh38
rs943556594 Chr.X: 155055252-155055252 on Build GRCh38
rs143802254 Chr.X: 155055257-155055257 on Build GRCh38
rs1001361875 Chr.X: 155054970-155054970 on Build GRCh38
rs997512909 Chr.X: 155054957-155054957 on Build GRCh38
rs782071520 Chr.X: 155054966-155054966 on Build GRCh38
rs929046735 Chr.X: 155055176-155055176 on Build GRCh38
rs901833431 Chr.X: 155054935-155054935 on Build GRCh38
rs1048795 Chr.X: 155054906-155054906 on Build GRCh38
rs782720062 Chr.X: 155055110-155055110 on Build GRCh38
rs933583357 Chr.X: 155055309-155055309 on Build GRCh38
rs782756518 Chr.X: 155055167-155055167 on Build GRCh38
rs782523293 Chr.X: 155055149-155055149 on Build GRCh38

View Full Product Details