Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155276769-155277319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC2
Entrez Gene ID
1193
Gene Name
chloride intracellular channel 2
Gene Aliases
CLIC2b, MRXS32, XAP121
UniGene
Hs.655445
Gene Chromosome Location
Chr.X: 155276207-155334681 on Build GRCh38
Transcripts
NM_001289.5

SNP Detail

SNP ID Chromosome Location
rs782489111 Chr.X: 155276833-155276833 on Build GRCh38
rs145583465 Chr.X: 155276976-155276976 on Build GRCh38
rs1005429346 Chr.X: 155277217-155277217 on Build GRCh38
rs782448069 Chr.X: 155276956-155276956 on Build GRCh38
rs991630929 Chr.X: 155277077-155277077 on Build GRCh38
rs950321424 Chr.X: 155277138-155277138 on Build GRCh38
rs782282895 Chr.X: 155276966-155276966 on Build GRCh38
rs899778815 Chr.X: 155277238-155277238 on Build GRCh38
rs925674575 Chr.X: 155276988-155276988 on Build GRCh38
rs1018612073 Chr.X: 155276846-155276846 on Build GRCh38
rs897806949 Chr.X: 155277167-155277167 on Build GRCh38
rs782507833 Chr.X: 155277124-155277124 on Build GRCh38
rs782731075 Chr.X: 155276882-155276882 on Build GRCh38
rs781955607 Chr.X: 155277146-155277146 on Build GRCh38
rs916188158 Chr.X: 155277136-155277136 on Build GRCh38
rs887824147 Chr.X: 155277216-155277216 on Build GRCh38
rs56064215 Chr.X: 155277071-155277071 on Build GRCh38
rs781806541 Chr.X: 155276951-155276951 on Build GRCh38
rs782298468 Chr.X: 155277248-155277248 on Build GRCh38
rs967126353 Chr.X: 155276943-155276943 on Build GRCh38
rs936178331 Chr.X: 155277045-155277045 on Build GRCh38
rs782683622 Chr.X: 155276822-155276822 on Build GRCh38
rs1027717374 Chr.X: 155276809-155276809 on Build GRCh38
rs782225748 Chr.X: 155277208-155277209 on Build GRCh38
rs929263345 Chr.X: 155277181-155277181 on Build GRCh38
rs1029320072 Chr.X: 155276908-155276908 on Build GRCh38
rs781929438 Chr.X: 155277237-155277237 on Build GRCh38
rs1049181997 Chr.X: 155277195-155277195 on Build GRCh38
rs998096340 Chr.X: 155277242-155277242 on Build GRCh38
rs782338105 Chr.X: 155277225-155277225 on Build GRCh38
rs976859390 Chr.X: 155276895-155276895 on Build GRCh38

View Full Product Details