Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155298928-155299339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC2
Entrez Gene ID
1193
Gene Name
chloride intracellular channel 2
Gene Aliases
CLIC2b, MRXS32, XAP121
UniGene
Hs.655445
Gene Chromosome Location
Chr.X: 155276207-155334681 on Build GRCh38
Transcripts
NM_001289.5

SNP Detail

SNP ID Chromosome Location
rs782807344 Chr.X: 155298990-155298990 on Build GRCh38
rs1012912805 Chr.X: 155299206-155299206 on Build GRCh38
rs981912087 Chr.X: 155299240-155299240 on Build GRCh38
rs782589527 Chr.X: 155299171-155299171 on Build GRCh38
rs782013964 Chr.X: 155299007-155299007 on Build GRCh38
rs782072010 Chr.X: 155298998-155298998 on Build GRCh38
rs782218666 Chr.X: 155299243-155299243 on Build GRCh38
rs782272592 Chr.X: 155299196-155299196 on Build GRCh38
rs782495285 Chr.X: 155299167-155299167 on Build GRCh38
rs782783664 Chr.X: 155299151-155299151 on Build GRCh38
rs782684108 Chr.X: 155299021-155299021 on Build GRCh38
rs1034821626 Chr.X: 155299284-155299284 on Build GRCh38
rs1023502014 Chr.X: 155299213-155299213 on Build GRCh38
rs934176781 Chr.X: 155299099-155299099 on Build GRCh38
rs782029089 Chr.X: 155299194-155299194 on Build GRCh38
rs782588490 Chr.X: 155299159-155299159 on Build GRCh38
rs782061068 Chr.X: 155299020-155299020 on Build GRCh38
rs5983864 Chr.X: 155299238-155299238 on Build GRCh38
rs781828011 Chr.X: 155299059-155299059 on Build GRCh38
rs782367152 Chr.X: 155299190-155299190 on Build GRCh38
rs368081571 Chr.X: 155299016-155299016 on Build GRCh38
rs41312792 Chr.X: 155299182-155299182 on Build GRCh38
rs377459528 Chr.X: 155299187-155299187 on Build GRCh38
rs782520088 Chr.X: 155299070-155299070 on Build GRCh38
rs782380904 Chr.X: 155299221-155299221 on Build GRCh38
rs782670176 Chr.X: 155298966-155298966 on Build GRCh38
rs782435839 Chr.X: 155298958-155298958 on Build GRCh38
rs782187128 Chr.X: 155299164-155299164 on Build GRCh38

View Full Product Details