Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 51698022-51698528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF2T
Entrez Gene ID
23283
Gene Name
cleavage stimulation factor subunit 2, tau variant
Gene Aliases
CstF-64T
UniGene
Hs.591358
Gene Chromosome Location
Chr.10: 51695486-51699595 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Gene Symbol
PRKG1
Entrez Gene ID
5592
Gene Name
protein kinase, cGMP-dependent, type I
Gene Aliases
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha
UniGene
Hs.407535
Gene Chromosome Location
Chr.10: 50991151-52298350 on Build GRCh38
Transcripts
NM_015235.2 NM_001098512.2 NM_006258.3 XM_011539952.2 XM_017016412.1 XM_017016413.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4443787 c.1307G>A p.R436K Chr.10: 51698243-51698243 on Build GRCh38
5518974 c.1075delC p.H359fs*22 Chr.10: 51698475-51698475 on Build GRCh38
5451081 c.1453G>A p.A485T Chr.10: 51698097-51698097 on Build GRCh38
3438643 c.1117C>T p.R373C Chr.10: 51698433-51698433 on Build GRCh38
918841 c.1324G>A p.A442T Chr.10: 51698226-51698226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771847513 Chr.10: 51698288-51698288 on Build GRCh38
rs750851950 Chr.10: 51698267-51698267 on Build GRCh38
rs146369924 Chr.10: 51698282-51698282 on Build GRCh38
rs139988598 Chr.10: 51698093-51698093 on Build GRCh38
rs750928413 Chr.10: 51698193-51698193 on Build GRCh38
rs779350917 Chr.10: 51698072-51698072 on Build GRCh38
rs369282662 Chr.10: 51698481-51698481 on Build GRCh38
rs3740228 Chr.10: 51698287-51698287 on Build GRCh38
rs1036729694 Chr.10: 51698323-51698323 on Build GRCh38
rs770746990 Chr.10: 51698129-51698129 on Build GRCh38
rs748345504 Chr.10: 51698210-51698210 on Build GRCh38
rs1003268596 Chr.10: 51698358-51698358 on Build GRCh38
rs759400764 Chr.10: 51698154-51698154 on Build GRCh38
rs963653879 Chr.10: 51698351-51698351 on Build GRCh38
rs201315901 Chr.10: 51698195-51698195 on Build GRCh38
rs765545916 Chr.10: 51698263-51698263 on Build GRCh38
rs945562538 Chr.10: 51698245-51698245 on Build GRCh38
rs758199123 Chr.10: 51698395-51698395 on Build GRCh38
rs200607148 Chr.10: 51698224-51698224 on Build GRCh38
rs772996942 Chr.10: 51698324-51698324 on Build GRCh38
rs767044550 Chr.10: 51698156-51698156 on Build GRCh38
rs878977306 Chr.10: 51698044-51698044 on Build GRCh38
rs764098088 Chr.10: 51698484-51698484 on Build GRCh38
rs779274176 Chr.10: 51698192-51698192 on Build GRCh38
rs369546537 Chr.10: 51698062-51698062 on Build GRCh38
rs774806602 Chr.10: 51698464-51698464 on Build GRCh38
rs768689903 Chr.10: 51698419-51698419 on Build GRCh38
rs772039924 Chr.10: 51698502-51698504 on Build GRCh38
rs144891671 Chr.10: 51698289-51698289 on Build GRCh38
rs75877881 Chr.10: 51698437-51698437 on Build GRCh38
rs775981203 Chr.10: 51698294-51698294 on Build GRCh38
rs1035286543 Chr.10: 51698087-51698087 on Build GRCh38
rs915168532 Chr.10: 51698109-51698109 on Build GRCh38
rs369368999 Chr.10: 51698283-51698283 on Build GRCh38
rs772286622 Chr.10: 51698074-51698074 on Build GRCh38
rs866839962 Chr.10: 51698466-51698466 on Build GRCh38
rs761210544 Chr.10: 51698319-51698319 on Build GRCh38
rs754061074 Chr.10: 51698492-51698492 on Build GRCh38
rs763471233 Chr.10: 51698331-51698331 on Build GRCh38
rs371185885 Chr.10: 51698362-51698362 on Build GRCh38
rs752100435 Chr.10: 51698277-51698277 on Build GRCh38
rs760341333 Chr.10: 51698322-51698322 on Build GRCh38
rs139186594 Chr.10: 51698286-51698286 on Build GRCh38
rs776404351 Chr.10: 51698426-51698426 on Build GRCh38
rs761490173 Chr.10: 51698261-51698261 on Build GRCh38
rs752868245 Chr.10: 51698366-51698366 on Build GRCh38
rs764346235 Chr.10: 51698432-51698432 on Build GRCh38
rs3824686 Chr.10: 51698407-51698407 on Build GRCh38
rs937252250 Chr.10: 51698285-51698285 on Build GRCh38
rs762563728 Chr.10: 51698310-51698318 on Build GRCh38
rs747127200 Chr.10: 51698203-51698203 on Build GRCh38
rs758806447 Chr.10: 51698272-51698272 on Build GRCh38
rs768087095 Chr.10: 51698293-51698293 on Build GRCh38
rs774169015 Chr.10: 51698144-51698144 on Build GRCh38
rs199709239 Chr.10: 51698112-51698112 on Build GRCh38
rs757426073 Chr.10: 51698496-51698496 on Build GRCh38
rs978770104 Chr.10: 51698186-51698186 on Build GRCh38
rs773013114 Chr.10: 51698434-51698434 on Build GRCh38
rs749495404 Chr.10: 51698386-51698386 on Build GRCh38
rs752151308 Chr.10: 51698162-51698162 on Build GRCh38
rs374193081 Chr.10: 51698469-51698469 on Build GRCh38
rs780687784 Chr.10: 51698197-51698197 on Build GRCh38
rs757615922 Chr.10: 51698175-51698175 on Build GRCh38
rs780244509 Chr.10: 51698085-51698085 on Build GRCh38
rs755122614 Chr.10: 51698278-51698278 on Build GRCh38
rs757607894 Chr.10: 51698067-51698067 on Build GRCh38
rs551079956 Chr.10: 51698080-51698080 on Build GRCh38
rs766869247 Chr.10: 51698081-51698081 on Build GRCh38
rs778120310 Chr.10: 51698375-51698375 on Build GRCh38
rs753137311 Chr.10: 51698477-51698477 on Build GRCh38
rs924691336 Chr.10: 51698202-51698202 on Build GRCh38
rs1008374857 Chr.10: 51698303-51698303 on Build GRCh38
rs766792935 Chr.10: 51698338-51698338 on Build GRCh38
rs768108943 Chr.10: 51698474-51698474 on Build GRCh38
rs376951469 Chr.10: 51698348-51698348 on Build GRCh38
rs373436302 Chr.10: 51698433-51698433 on Build GRCh38
rs781265932 Chr.10: 51698279-51698279 on Build GRCh38
rs759490518 Chr.10: 51698228-51698228 on Build GRCh38
rs775371688 Chr.10: 51698254-51698254 on Build GRCh38
rs111331961 Chr.10: 51698273-51698273 on Build GRCh38
rs774154367 Chr.10: 51698225-51698225 on Build GRCh38
rs112642722 Chr.10: 51698061-51698061 on Build GRCh38
rs991114121 Chr.10: 51698102-51698102 on Build GRCh38
rs768854110 Chr.10: 51698207-51698207 on Build GRCh38
rs571252591 Chr.10: 51698222-51698222 on Build GRCh38
rs755471672 Chr.10: 51698353-51698353 on Build GRCh38
rs781327433 Chr.10: 51698209-51698209 on Build GRCh38
rs530966056 Chr.10: 51698259-51698259 on Build GRCh38
rs571544210 Chr.10: 51698257-51698257 on Build GRCh38
rs148622678 Chr.10: 51698300-51698300 on Build GRCh38
rs753520233 Chr.10: 51698165-51698165 on Build GRCh38
rs572345819 Chr.10: 51698050-51698050 on Build GRCh38
rs142115985 Chr.10: 51698230-51698230 on Build GRCh38
rs769050592 Chr.10: 51698108-51698108 on Build GRCh38
rs766879975 Chr.10: 51698454-51698454 on Build GRCh38
rs535610984 Chr.10: 51698400-51698400 on Build GRCh38
rs748117556 Chr.10: 51698476-51698490 on Build GRCh38
rs760153053 Chr.10: 51698164-51698164 on Build GRCh38
rs959432517 Chr.10: 51698380-51698380 on Build GRCh38

View Full Product Details