Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58269015-58269512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD1
Entrez Gene ID
55847
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
C10orf70, MDS029, ZCD1, mitoNEET
UniGene
Hs.370102
Gene Chromosome Location
Chr.10: 58269102-58290553 on Build GRCh38
Transcripts
NM_018464.4 XM_017016013.1

Gene Symbol
IPMK
Entrez Gene ID
253430
Gene Name
inositol polyphosphate multikinase
Gene Aliases
-
UniGene
Hs.30280
Gene Chromosome Location
Chr.10: 58191517-58269837 on Build GRCh38
Transcripts
NM_018464.4 XM_017016013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966565 c.20C>A p.S7Y Chr.10: 58269293-58269293 on Build GRCh38
5370874 c.12T>C p.T4T Chr.10: 58269285-58269285 on Build GRCh38
6248894 c.23G>T p.S8I Chr.10: 58269296-58269296 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1044003664 Chr.10: 58269318-58269318 on Build GRCh38
rs925511321 Chr.10: 58269336-58269336 on Build GRCh38
rs776749254 Chr.10: 58269255-58269255 on Build GRCh38
rs748910021 Chr.10: 58269324-58269324 on Build GRCh38
rs370461677 Chr.10: 58269254-58269254 on Build GRCh38
rs961865056 Chr.10: 58269359-58269359 on Build GRCh38
rs778438933 Chr.10: 58269130-58269130 on Build GRCh38
rs575657868 Chr.10: 58269159-58269159 on Build GRCh38
rs1038808983 Chr.10: 58269191-58269191 on Build GRCh38
rs1034104411 Chr.10: 58269470-58269470 on Build GRCh38
rs754961383 Chr.10: 58269290-58269290 on Build GRCh38
rs535002429 Chr.10: 58269120-58269120 on Build GRCh38
rs992241611 Chr.10: 58269483-58269483 on Build GRCh38
rs760089570 Chr.10: 58269347-58269347 on Build GRCh38
rs997242118 Chr.10: 58269199-58269199 on Build GRCh38
rs909921499 Chr.10: 58269103-58269103 on Build GRCh38
rs371715015 Chr.10: 58269244-58269244 on Build GRCh38
rs752760288 Chr.10: 58269245-58269245 on Build GRCh38
rs747876636 Chr.10: 58269227-58269227 on Build GRCh38
rs372053931 Chr.10: 58269270-58269270 on Build GRCh38
rs777324011 Chr.10: 58269319-58269319 on Build GRCh38
rs1041319607 Chr.10: 58269452-58269452 on Build GRCh38
rs557468646 Chr.10: 58269143-58269143 on Build GRCh38
rs941357767 Chr.10: 58269105-58269105 on Build GRCh38
rs398013634 Chr.10: 58269080-58269081 on Build GRCh38
rs560892775 Chr.10: 58269361-58269361 on Build GRCh38
rs1011888138 Chr.10: 58269289-58269289 on Build GRCh38
rs965926839 Chr.10: 58269050-58269050 on Build GRCh38
rs902770043 Chr.10: 58269457-58269457 on Build GRCh38
rs886298977 Chr.10: 58269390-58269390 on Build GRCh38
rs999270388 Chr.10: 58269042-58269042 on Build GRCh38
rs35093457 Chr.10: 58269088-58269088 on Build GRCh38
rs2251039 Chr.10: 58269134-58269134 on Build GRCh38
rs780995447 Chr.10: 58269112-58269112 on Build GRCh38
rs757354920 Chr.10: 58269325-58269326 on Build GRCh38
rs150889076 Chr.10: 58269215-58269215 on Build GRCh38
rs142533752 Chr.10: 58269275-58269275 on Build GRCh38
rs2790171 Chr.10: 58269175-58269175 on Build GRCh38
rs776217903 Chr.10: 58269224-58269224 on Build GRCh38
rs747037831 Chr.10: 58269298-58269299 on Build GRCh38
rs907698672 Chr.10: 58269124-58269124 on Build GRCh38
rs375164960 Chr.10: 58269310-58269310 on Build GRCh38
rs950616774 Chr.10: 58269085-58269085 on Build GRCh38
rs926687671 Chr.10: 58269115-58269115 on Build GRCh38
rs541472900 Chr.10: 58269238-58269238 on Build GRCh38
rs970399456 Chr.10: 58269429-58269429 on Build GRCh38
rs1008467267 Chr.10: 58269188-58269188 on Build GRCh38
rs541226587 Chr.10: 58269242-58269242 on Build GRCh38
rs975899073 Chr.10: 58269071-58269071 on Build GRCh38
rs541369297 Chr.10: 58269210-58269210 on Build GRCh38
rs780910983 Chr.10: 58269301-58269301 on Build GRCh38
rs1007616736 Chr.10: 58269186-58269186 on Build GRCh38
rs369617882 Chr.10: 58269342-58269342 on Build GRCh38
rs901601056 Chr.10: 58269193-58269193 on Build GRCh38
rs772545630 Chr.10: 58269355-58269355 on Build GRCh38
rs552933342 Chr.10: 58269129-58269129 on Build GRCh38
rs201057069 Chr.10: 58269269-58269269 on Build GRCh38
rs779937526 Chr.10: 58269278-58269278 on Build GRCh38
rs201855607 Chr.10: 58269237-58269237 on Build GRCh38
rs958036102 Chr.10: 58269253-58269253 on Build GRCh38
rs1006184857 Chr.10: 58269217-58269217 on Build GRCh38
rs944054352 Chr.10: 58269422-58269422 on Build GRCh38
rs570784890 Chr.10: 58269111-58269111 on Build GRCh38
rs775586107 Chr.10: 58269356-58269356 on Build GRCh38
rs771285785 Chr.10: 58269327-58269327 on Build GRCh38
rs1026162497 Chr.10: 58269203-58269203 on Build GRCh38
rs751548592 Chr.10: 58269287-58269287 on Build GRCh38
rs543557066 Chr.10: 58269165-58269165 on Build GRCh38
rs67338868 Chr.10: 58269078-58269079 on Build GRCh38
rs960686168 Chr.10: 58269480-58269480 on Build GRCh38
rs1017767099 Chr.10: 58269044-58269044 on Build GRCh38
rs74675316 Chr.10: 58269404-58269404 on Build GRCh38
rs530545136 Chr.10: 58269241-58269241 on Build GRCh38
rs367769431 Chr.10: 58269240-58269240 on Build GRCh38
rs1047511771 Chr.10: 58269388-58269388 on Build GRCh38
rs1001420863 Chr.10: 58269455-58269455 on Build GRCh38
rs577171846 Chr.10: 58269180-58269180 on Build GRCh38
rs907836482 Chr.10: 58269358-58269358 on Build GRCh38
rs747592583 Chr.10: 58269308-58269308 on Build GRCh38
rs769319492 Chr.10: 58269234-58269234 on Build GRCh38
rs142456520 Chr.10: 58269133-58269133 on Build GRCh38
rs940551845 Chr.10: 58269125-58269125 on Build GRCh38
rs973651493 Chr.10: 58269364-58269364 on Build GRCh38
rs919993489 Chr.10: 58269385-58269385 on Build GRCh38
rs901475891 Chr.10: 58269357-58269357 on Build GRCh38
rs945638048 Chr.10: 58269145-58269145 on Build GRCh38
rs926622008 Chr.10: 58269243-58269243 on Build GRCh38
rs1026121727 Chr.10: 58269075-58269075 on Build GRCh38
rs878869720 Chr.10: 58269101-58269101 on Build GRCh38
rs1416763 Chr.10: 58269403-58269403 on Build GRCh38

View Full Product Details