Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58287799-58288309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CISD1
Entrez Gene ID
55847
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
C10orf70, MDS029, ZCD1, mitoNEET
UniGene
Hs.370102
Gene Chromosome Location
Chr.10: 58269102-58290553 on Build GRCh38
Transcripts
NM_018464.4 XM_017016409.1

SNP Detail

SNP ID Chromosome Location
rs905074293 Chr.10: 58288174-58288174 on Build GRCh38
rs1046917326 Chr.10: 58288115-58288115 on Build GRCh38
rs777422032 Chr.10: 58288108-58288108 on Build GRCh38
rs895922017 Chr.10: 58288193-58288193 on Build GRCh38
rs913371723 Chr.10: 58287985-58287985 on Build GRCh38
rs60626434 Chr.10: 58287833-58287833 on Build GRCh38
rs1001515697 Chr.10: 58287869-58287869 on Build GRCh38
rs781087329 Chr.10: 58287968-58287968 on Build GRCh38
rs986114637 Chr.10: 58288188-58288188 on Build GRCh38
rs199920654 Chr.10: 58287863-58287863 on Build GRCh38
rs201018053 Chr.10: 58287864-58287864 on Build GRCh38
rs574372590 Chr.10: 58288073-58288073 on Build GRCh38
rs929714549 Chr.10: 58288114-58288114 on Build GRCh38
rs777249828 Chr.10: 58287931-58287931 on Build GRCh38
rs929464736 Chr.10: 58288168-58288168 on Build GRCh38
rs796985790 Chr.10: 58288272-58288272 on Build GRCh38
rs114589568 Chr.10: 58288178-58288178 on Build GRCh38
rs970062088 Chr.10: 58287849-58287849 on Build GRCh38
rs920588742 Chr.10: 58287879-58287879 on Build GRCh38
rs201677186 Chr.10: 58287861-58287861 on Build GRCh38
rs1012559106 Chr.10: 58288096-58288096 on Build GRCh38
rs1041849424 Chr.10: 58287966-58287966 on Build GRCh38
rs922409967 Chr.10: 58288112-58288112 on Build GRCh38
rs372198623 Chr.10: 58287832-58287832 on Build GRCh38
rs769533510 Chr.10: 58288038-58288038 on Build GRCh38
rs79076458 Chr.10: 58287863-58287863 on Build GRCh38
rs1040652817 Chr.10: 58288110-58288110 on Build GRCh38
rs1051098776 Chr.10: 58287908-58287908 on Build GRCh38
rs796474833 Chr.10: 58287849-58287849 on Build GRCh38
rs1035783506 Chr.10: 58287888-58287888 on Build GRCh38
rs140699213 Chr.10: 58288177-58288177 on Build GRCh38
rs1010722236 Chr.10: 58288276-58288276 on Build GRCh38
rs1004540634 Chr.10: 58288137-58288137 on Build GRCh38
rs56776648 Chr.10: 58287864-58287864 on Build GRCh38
rs185315951 Chr.10: 58288223-58288223 on Build GRCh38
rs978110011 Chr.10: 58288273-58288273 on Build GRCh38
rs200440932 Chr.10: 58288166-58288166 on Build GRCh38
rs924758218 Chr.10: 58288275-58288275 on Build GRCh38
rs1047641 Chr.10: 58287862-58287862 on Build GRCh38
rs193118266 Chr.10: 58288181-58288181 on Build GRCh38
rs138033937 Chr.10: 58288209-58288210 on Build GRCh38
rs77230219 Chr.10: 58287836-58287836 on Build GRCh38
rs369446826 Chr.10: 58288116-58288116 on Build GRCh38
rs953605961 Chr.10: 58288184-58288184 on Build GRCh38
rs959463446 Chr.10: 58287851-58287851 on Build GRCh38
rs79650878 Chr.10: 58287833-58287833 on Build GRCh38
rs538797459 Chr.10: 58288222-58288222 on Build GRCh38
rs999937387 Chr.10: 58288011-58288011 on Build GRCh38
rs941146381 Chr.10: 58288130-58288130 on Build GRCh38
rs371048809 Chr.10: 58287825-58287825 on Build GRCh38
rs759205736 Chr.10: 58287863-58287864 on Build GRCh38
rs74146625 Chr.10: 58287832-58287832 on Build GRCh38
rs912118606 Chr.10: 58287919-58287919 on Build GRCh38
rs2590369 Chr.10: 58288034-58288034 on Build GRCh38

View Full Product Details