Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59262344-59262849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
UniGene
Hs.607594
Gene Chromosome Location
Chr.10: 59245933-59363186 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
684817 c.1076C>G p.P359R Chr.10: 59262594-59262594 on Build GRCh38
6883229 c.1114G>T p.E372* Chr.10: 59262556-59262556 on Build GRCh38
4461763 c.1219C>T p.P407S Chr.10: 59262451-59262451 on Build GRCh38
5544942 c.1174G>C p.D392H Chr.10: 59262496-59262496 on Build GRCh38
6883228 c.1114G>T p.E372* Chr.10: 59262556-59262556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915553219 Chr.10: 59262759-59262759 on Build GRCh38
rs890534941 Chr.10: 59262694-59262694 on Build GRCh38
rs773481958 Chr.10: 59262605-59262605 on Build GRCh38
rs34696661 Chr.10: 59262673-59262674 on Build GRCh38
rs752603616 Chr.10: 59262549-59262549 on Build GRCh38
rs967710269 Chr.10: 59262776-59262776 on Build GRCh38
rs573596509 Chr.10: 59262567-59262567 on Build GRCh38
rs377472100 Chr.10: 59262417-59262417 on Build GRCh38
rs370922025 Chr.10: 59262527-59262527 on Build GRCh38
rs930378783 Chr.10: 59262726-59262726 on Build GRCh38
rs753741976 Chr.10: 59262570-59262570 on Build GRCh38
rs1038288194 Chr.10: 59262654-59262654 on Build GRCh38
rs774534265 Chr.10: 59262691-59262691 on Build GRCh38
rs999679880 Chr.10: 59262386-59262386 on Build GRCh38
rs199675574 Chr.10: 59262414-59262414 on Build GRCh38
rs757180554 Chr.10: 59262571-59262571 on Build GRCh38
rs189991779 Chr.10: 59262671-59262671 on Build GRCh38
rs146006694 Chr.10: 59262781-59262781 on Build GRCh38
rs201233580 Chr.10: 59262542-59262542 on Build GRCh38
rs779734354 Chr.10: 59262668-59262668 on Build GRCh38
rs35187307 Chr.10: 59262745-59262745 on Build GRCh38
rs114156972 Chr.10: 59262463-59262463 on Build GRCh38
rs991662005 Chr.10: 59262727-59262727 on Build GRCh38
rs781421231 Chr.10: 59262672-59262672 on Build GRCh38
rs771772201 Chr.10: 59262579-59262579 on Build GRCh38
rs759756641 Chr.10: 59262628-59262628 on Build GRCh38
rs771156309 Chr.10: 59262686-59262686 on Build GRCh38
rs954715542 Chr.10: 59262406-59262406 on Build GRCh38
rs1020652257 Chr.10: 59262486-59262486 on Build GRCh38
rs770488077 Chr.10: 59262681-59262681 on Build GRCh38
rs140430636 Chr.10: 59262543-59262543 on Build GRCh38
rs113016699 Chr.10: 59262533-59262533 on Build GRCh38
rs758649167 Chr.10: 59262403-59262403 on Build GRCh38
rs758676876 Chr.10: 59262660-59262660 on Build GRCh38
rs778401167 Chr.10: 59262574-59262574 on Build GRCh38
rs770274840 Chr.10: 59262603-59262603 on Build GRCh38
rs763827338 Chr.10: 59262629-59262629 on Build GRCh38
rs746668566 Chr.10: 59262670-59262670 on Build GRCh38
rs987513422 Chr.10: 59262560-59262560 on Build GRCh38
rs766747856 Chr.10: 59262612-59262612 on Build GRCh38
rs777564914 Chr.10: 59262468-59262468 on Build GRCh38
rs200083976 Chr.10: 59262453-59262453 on Build GRCh38
rs200879419 Chr.10: 59262534-59262534 on Build GRCh38
rs544779823 Chr.10: 59262780-59262780 on Build GRCh38
rs767450934 Chr.10: 59262784-59262784 on Build GRCh38
rs775849830 Chr.10: 59262518-59262518 on Build GRCh38
rs767722407 Chr.10: 59262693-59262693 on Build GRCh38
rs1000090949 Chr.10: 59262779-59262779 on Build GRCh38
rs752482559 Chr.10: 59262415-59262415 on Build GRCh38
rs750004093 Chr.10: 59262661-59262661 on Build GRCh38
rs375098743 Chr.10: 59262581-59262581 on Build GRCh38
rs1020248311 Chr.10: 59262744-59262744 on Build GRCh38
rs765149770 Chr.10: 59262659-59262659 on Build GRCh38
rs745550781 Chr.10: 59262497-59262497 on Build GRCh38
rs776976417 Chr.10: 59262530-59262530 on Build GRCh38
rs151012926 Chr.10: 59262493-59262493 on Build GRCh38
rs367803531 Chr.10: 59262652-59262652 on Build GRCh38
rs763119533 Chr.10: 59262611-59262611 on Build GRCh38
rs760604744 Chr.10: 59262676-59262676 on Build GRCh38
rs747444856 Chr.10: 59262586-59262586 on Build GRCh38
rs771647200 Chr.10: 59262503-59262503 on Build GRCh38
rs748604510 Chr.10: 59262597-59262597 on Build GRCh38
rs768921251 Chr.10: 59262595-59262595 on Build GRCh38
rs533230915 Chr.10: 59262823-59262823 on Build GRCh38
rs769274515 Chr.10: 59262528-59262528 on Build GRCh38
rs773821398 Chr.10: 59262684-59262684 on Build GRCh38
rs377477932 Chr.10: 59262822-59262822 on Build GRCh38
rs774691934 Chr.10: 59262613-59262613 on Build GRCh38
rs769164016 Chr.10: 59262805-59262805 on Build GRCh38
rs866320960 Chr.10: 59262548-59262548 on Build GRCh38
rs368763247 Chr.10: 59262692-59262692 on Build GRCh38
rs752809470 Chr.10: 59262796-59262796 on Build GRCh38
rs748868878 Chr.10: 59262470-59262470 on Build GRCh38
rs546787378 Chr.10: 59262412-59262412 on Build GRCh38
rs745328080 Chr.10: 59262578-59262578 on Build GRCh38
rs755833524 Chr.10: 59262550-59262550 on Build GRCh38
rs749427683 Chr.10: 59262685-59262685 on Build GRCh38
rs371741777 Chr.10: 59262664-59262664 on Build GRCh38
rs979007685 Chr.10: 59262507-59262507 on Build GRCh38
rs1031208398 Chr.10: 59262405-59262405 on Build GRCh38
rs35458924 Chr.10: 59262677-59262678 on Build GRCh38
rs958556535 Chr.10: 59262791-59262791 on Build GRCh38
rs140859038 Chr.10: 59262704-59262704 on Build GRCh38
rs770327303 Chr.10: 59262487-59262487 on Build GRCh38

View Full Product Details