Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59269810-59270323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
UniGene
Hs.607594
Gene Chromosome Location
Chr.10: 59245933-59363186 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682599 c.641C>T p.P214L Chr.10: 59270061-59270061 on Build GRCh38
7212444 c.603A>C p.P201P Chr.10: 59270099-59270099 on Build GRCh38
684815 c.719G>A p.R240Q Chr.10: 59269983-59269983 on Build GRCh38
5673053 c.607C>A p.H203N Chr.10: 59270095-59270095 on Build GRCh38
3686800 c.619C>T p.Q207* Chr.10: 59270083-59270083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773841608 Chr.10: 59269864-59269864 on Build GRCh38
rs771314305 Chr.10: 59270030-59270030 on Build GRCh38
rs367555975 Chr.10: 59270009-59270009 on Build GRCh38
rs752888748 Chr.10: 59270055-59270055 on Build GRCh38
rs973581445 Chr.10: 59270052-59270052 on Build GRCh38
rs755028671 Chr.10: 59270099-59270099 on Build GRCh38
rs768651945 Chr.10: 59269983-59269983 on Build GRCh38
rs190127197 Chr.10: 59270165-59270165 on Build GRCh38
rs758181480 Chr.10: 59270086-59270087 on Build GRCh38
rs187251609 Chr.10: 59270039-59270039 on Build GRCh38
rs143474599 Chr.10: 59270067-59270067 on Build GRCh38
rs777296280 Chr.10: 59270127-59270127 on Build GRCh38
rs1012742613 Chr.10: 59270295-59270295 on Build GRCh38
rs756416635 Chr.10: 59269901-59269901 on Build GRCh38
rs777854836 Chr.10: 59269907-59269907 on Build GRCh38
rs746977874 Chr.10: 59269970-59269970 on Build GRCh38
rs535596917 Chr.10: 59269948-59269948 on Build GRCh38
rs181798562 Chr.10: 59270017-59270017 on Build GRCh38
rs761197844 Chr.10: 59270153-59270153 on Build GRCh38
rs976593191 Chr.10: 59270247-59270247 on Build GRCh38
rs780002914 Chr.10: 59270083-59270083 on Build GRCh38
rs948527964 Chr.10: 59269904-59269904 on Build GRCh38
rs770512808 Chr.10: 59270131-59270131 on Build GRCh38
rs529098149 Chr.10: 59270022-59270022 on Build GRCh38
rs759891121 Chr.10: 59270133-59270133 on Build GRCh38
rs149406709 Chr.10: 59269988-59269988 on Build GRCh38
rs374333238 Chr.10: 59269877-59269877 on Build GRCh38
rs1007640414 Chr.10: 59270208-59270208 on Build GRCh38
rs752081543 Chr.10: 59269866-59269866 on Build GRCh38
rs779949458 Chr.10: 59269964-59269964 on Build GRCh38
rs371136281 Chr.10: 59270150-59270150 on Build GRCh38
rs759995374 Chr.10: 59269867-59269867 on Build GRCh38
rs1015185358 Chr.10: 59270018-59270018 on Build GRCh38
rs977101559 Chr.10: 59270209-59270209 on Build GRCh38
rs772608949 Chr.10: 59270152-59270152 on Build GRCh38
rs202010856 Chr.10: 59269914-59269914 on Build GRCh38
rs968309017 Chr.10: 59270195-59270195 on Build GRCh38
rs891436556 Chr.10: 59270040-59270040 on Build GRCh38
rs764779870 Chr.10: 59269862-59269862 on Build GRCh38
rs764428234 Chr.10: 59270069-59270069 on Build GRCh38
rs928311040 Chr.10: 59270217-59270217 on Build GRCh38
rs769207376 Chr.10: 59270124-59270124 on Build GRCh38
rs747602970 Chr.10: 59270001-59270001 on Build GRCh38
rs752371603 Chr.10: 59269886-59269887 on Build GRCh38
rs751346288 Chr.10: 59270090-59270090 on Build GRCh38
rs748896508 Chr.10: 59270130-59270130 on Build GRCh38
rs200777981 Chr.10: 59269897-59269897 on Build GRCh38
rs781240537 Chr.10: 59270106-59270106 on Build GRCh38
rs368410005 Chr.10: 59270071-59270071 on Build GRCh38
rs747740196 Chr.10: 59270120-59270120 on Build GRCh38
rs150637641 Chr.10: 59270072-59270072 on Build GRCh38
rs148013871 Chr.10: 59270068-59270068 on Build GRCh38
rs1024044275 Chr.10: 59269982-59269982 on Build GRCh38
rs774755628 Chr.10: 59270132-59270132 on Build GRCh38
rs773956207 Chr.10: 59270297-59270297 on Build GRCh38
rs762615807 Chr.10: 59270029-59270029 on Build GRCh38
rs181830368 Chr.10: 59270229-59270229 on Build GRCh38
rs938608267 Chr.10: 59270224-59270224 on Build GRCh38
rs539092134 Chr.10: 59270108-59270108 on Build GRCh38
rs1044293423 Chr.10: 59269931-59269931 on Build GRCh38
rs534462817 Chr.10: 59270278-59270278 on Build GRCh38
rs773689302 Chr.10: 59269863-59269863 on Build GRCh38
rs894240443 Chr.10: 59269949-59269949 on Build GRCh38
rs767999626 Chr.10: 59270050-59270050 on Build GRCh38
rs2290852 Chr.10: 59269881-59269881 on Build GRCh38
rs920736357 Chr.10: 59270066-59270066 on Build GRCh38
rs919375511 Chr.10: 59270296-59270296 on Build GRCh38
rs372255393 Chr.10: 59270033-59270033 on Build GRCh38
rs766918029 Chr.10: 59269865-59269865 on Build GRCh38
rs758108362 Chr.10: 59269940-59269940 on Build GRCh38

View Full Product Details