Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59790246-59790746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC6
Entrez Gene ID
8030
Gene Name
coiled-coil domain containing 6
Gene Aliases
D10S170, H4, PTC, TPC, TST1
UniGene
Hs.591360
Gene Chromosome Location
Chr.10: 59788747-59906656 on Build GRCh38
Transcripts
NM_005436.4

SNP Detail

SNP ID Chromosome Location
rs973794903 Chr.10: 59790491-59790491 on Build GRCh38
rs989704092 Chr.10: 59790451-59790451 on Build GRCh38
rs371528873 Chr.10: 59790564-59790564 on Build GRCh38
rs779703533 Chr.10: 59790526-59790526 on Build GRCh38
rs533132006 Chr.10: 59790634-59790634 on Build GRCh38
rs906591587 Chr.10: 59790638-59790638 on Build GRCh38
rs559988531 Chr.10: 59790374-59790374 on Build GRCh38
rs1006158813 Chr.10: 59790316-59790316 on Build GRCh38
rs758234337 Chr.10: 59790441-59790441 on Build GRCh38
rs140814509 Chr.10: 59790574-59790574 on Build GRCh38
rs530212450 Chr.10: 59790547-59790547 on Build GRCh38
rs796481194 Chr.10: 59790442-59790442 on Build GRCh38
rs925252613 Chr.10: 59790637-59790637 on Build GRCh38
rs993728774 Chr.10: 59790598-59790601 on Build GRCh38
rs914039074 Chr.10: 59790455-59790455 on Build GRCh38
rs1016553568 Chr.10: 59790474-59790474 on Build GRCh38
rs779807803 Chr.10: 59790478-59790478 on Build GRCh38
rs572147966 Chr.10: 59790405-59790405 on Build GRCh38
rs1050062338 Chr.10: 59790506-59790506 on Build GRCh38
rs933115885 Chr.10: 59790504-59790504 on Build GRCh38
rs150979800 Chr.10: 59790437-59790437 on Build GRCh38
rs545017664 Chr.10: 59790355-59790355 on Build GRCh38
rs977912421 Chr.10: 59790595-59790595 on Build GRCh38
rs564867920 Chr.10: 59790632-59790632 on Build GRCh38
rs756884651 Chr.10: 59790286-59790286 on Build GRCh38
rs1033612394 Chr.10: 59790684-59790684 on Build GRCh38
rs755314795 Chr.10: 59790394-59790394 on Build GRCh38
rs993554812 Chr.10: 59790404-59790404 on Build GRCh38
rs932979831 Chr.10: 59790293-59790293 on Build GRCh38
rs1024837553 Chr.10: 59790630-59790630 on Build GRCh38
rs1039055800 Chr.10: 59790687-59790687 on Build GRCh38
rs781548262 Chr.10: 59790426-59790426 on Build GRCh38
rs936569552 Chr.10: 59790639-59790639 on Build GRCh38
rs897660015 Chr.10: 59790399-59790399 on Build GRCh38
rs531472951 Chr.10: 59790573-59790573 on Build GRCh38
rs747013141 Chr.10: 59790612-59790612 on Build GRCh38
rs1033744840 Chr.10: 59790494-59790494 on Build GRCh38
rs981166686 Chr.10: 59790428-59790428 on Build GRCh38
rs897780737 Chr.10: 59790588-59790589 on Build GRCh38
rs560752653 Chr.10: 59790556-59790556 on Build GRCh38
rs923007942 Chr.10: 59790490-59790490 on Build GRCh38
rs3802695 Chr.10: 59790299-59790299 on Build GRCh38
rs372307347 Chr.10: 59790500-59790500 on Build GRCh38
rs945540015 Chr.10: 59790460-59790460 on Build GRCh38
rs561293349 Chr.10: 59790310-59790310 on Build GRCh38
rs901472743 Chr.10: 59790270-59790270 on Build GRCh38

View Full Product Details