Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68431754-68432256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNA2
Entrez Gene ID
1763
Gene Name
DNA replication helicase/nuclease 2
Gene Aliases
DNA2L, hDNA2
UniGene
Hs.532446
Gene Chromosome Location
Chr.10: 68414064-68472521 on Build GRCh38
Transcripts
NM_001080449.2 NR_102264.1 XM_006717680.3 XM_011539417.1 XM_017015799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6505012 c.1918T>C p.S640P Chr.10: 68431927-68431927 on Build GRCh38
3807530 c.1826C>G p.S609C Chr.10: 68432253-68432253 on Build GRCh38
4015272 c.2186A>G p.Y729C Chr.10: 68431917-68431917 on Build GRCh38
6505011 c.2176T>C p.S726P Chr.10: 68431927-68431927 on Build GRCh38
280449 c.1824T>G p.S608R Chr.10: 68432255-68432255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779899034 Chr.10: 68432167-68432167 on Build GRCh38
rs763355093 Chr.10: 68431948-68431948 on Build GRCh38
rs780293065 Chr.10: 68431867-68431867 on Build GRCh38
rs745691227 Chr.10: 68431907-68431907 on Build GRCh38
rs376585293 Chr.10: 68431965-68431965 on Build GRCh38
rs775102780 Chr.10: 68431823-68431823 on Build GRCh38
rs992336938 Chr.10: 68431847-68431847 on Build GRCh38
rs190231374 Chr.10: 68432139-68432139 on Build GRCh38
rs776214390 Chr.10: 68431944-68431944 on Build GRCh38
rs776145149 Chr.10: 68432175-68432178 on Build GRCh38
rs768376767 Chr.10: 68432184-68432184 on Build GRCh38
rs747501129 Chr.10: 68431926-68431926 on Build GRCh38
rs530713841 Chr.10: 68432118-68432118 on Build GRCh38
rs1007417870 Chr.10: 68432129-68432129 on Build GRCh38
rs766576587 Chr.10: 68431952-68431952 on Build GRCh38
rs773159668 Chr.10: 68431808-68431810 on Build GRCh38
rs1031386686 Chr.10: 68432205-68432205 on Build GRCh38
rs61855090 Chr.10: 68431874-68431874 on Build GRCh38
rs745639863 Chr.10: 68432010-68432010 on Build GRCh38
rs954877139 Chr.10: 68432051-68432051 on Build GRCh38
rs900000038 Chr.10: 68432112-68432129 on Build GRCh38
rs775963035 Chr.10: 68432187-68432187 on Build GRCh38
rs755073004 Chr.10: 68431875-68431875 on Build GRCh38
rs570858843 Chr.10: 68432105-68432105 on Build GRCh38
rs771009196 Chr.10: 68431897-68431897 on Build GRCh38
rs532891518 Chr.10: 68431812-68431812 on Build GRCh38
rs771877791 Chr.10: 68431818-68431818 on Build GRCh38
rs765526721 Chr.10: 68431856-68431856 on Build GRCh38
rs755233490 Chr.10: 68431972-68431972 on Build GRCh38
rs779038287 Chr.10: 68432006-68432006 on Build GRCh38
rs1017867198 Chr.10: 68432219-68432219 on Build GRCh38
rs977179809 Chr.10: 68431788-68431788 on Build GRCh38
rs935153751 Chr.10: 68432118-68432119 on Build GRCh38
rs775059467 Chr.10: 68431914-68431914 on Build GRCh38
rs753314834 Chr.10: 68432144-68432144 on Build GRCh38
rs767980817 Chr.10: 68431937-68431937 on Build GRCh38
rs757222938 Chr.10: 68431999-68431999 on Build GRCh38
rs765880716 Chr.10: 68431860-68431870 on Build GRCh38
rs1017963757 Chr.10: 68431957-68431957 on Build GRCh38
rs559735066 Chr.10: 68431862-68431862 on Build GRCh38
rs933127553 Chr.10: 68432100-68432100 on Build GRCh38
rs766587630 Chr.10: 68432227-68432227 on Build GRCh38
rs369587693 Chr.10: 68431893-68431893 on Build GRCh38
rs773532624 Chr.10: 68431947-68431947 on Build GRCh38
rs948304605 Chr.10: 68431921-68431921 on Build GRCh38
rs767274530 Chr.10: 68431945-68431945 on Build GRCh38
rs747695993 Chr.10: 68432190-68432190 on Build GRCh38
rs186375359 Chr.10: 68431837-68431837 on Build GRCh38
rs754365021 Chr.10: 68431845-68431845 on Build GRCh38
rs760122439 Chr.10: 68431848-68431850 on Build GRCh38
rs778805630 Chr.10: 68431900-68431900 on Build GRCh38
rs750808724 Chr.10: 68431858-68431858 on Build GRCh38
rs771822514 Chr.10: 68431913-68431913 on Build GRCh38
rs758330868 Chr.10: 68432013-68432013 on Build GRCh38
rs116704917 Chr.10: 68431946-68431946 on Build GRCh38
rs1036439379 Chr.10: 68432023-68432023 on Build GRCh38
rs767438888 Chr.10: 68431989-68431989 on Build GRCh38
rs866790583 Chr.10: 68432173-68432173 on Build GRCh38
rs1046351526 Chr.10: 68432112-68432112 on Build GRCh38
rs746582640 Chr.10: 68432177-68432177 on Build GRCh38
rs746522359 Chr.10: 68431936-68431936 on Build GRCh38
rs777761639 Chr.10: 68431878-68431878 on Build GRCh38
rs773767457 Chr.10: 68432206-68432206 on Build GRCh38
rs370696008 Chr.10: 68432214-68432214 on Build GRCh38
rs568739714 Chr.10: 68431993-68431993 on Build GRCh38
rs770263440 Chr.10: 68432193-68432193 on Build GRCh38
rs531218657 Chr.10: 68432075-68432075 on Build GRCh38
rs144115030 Chr.10: 68432101-68432101 on Build GRCh38
rs762382900 Chr.10: 68431851-68431851 on Build GRCh38

View Full Product Details