Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58012616-58013097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USB1
Entrez Gene ID
79650
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases
C16orf57, HVSL1, Mpn1, PN, hUsb1
UniGene
Hs.408702
Gene Chromosome Location
Chr.16: 57999600-58021623 on Build GRCh38
Transcripts
NM_001195302.1 NM_001204911.1 NM_024598.3 XM_005256144.3 XM_011523328.2 XM_011523329.2 XM_011523330.1

SNP Detail

SNP ID Chromosome Location
rs987378032 Chr.16: 58012825-58012825 on Build GRCh38
rs1028683802 Chr.16: 58012809-58012809 on Build GRCh38
rs772236772 Chr.16: 58012763-58012763 on Build GRCh38
rs867846369 Chr.16: 58012923-58012923 on Build GRCh38
rs763664239 Chr.16: 58012998-58012998 on Build GRCh38
rs532468493 Chr.16: 58012673-58012673 on Build GRCh38
rs945854785 Chr.16: 58012916-58012916 on Build GRCh38
rs997505672 Chr.16: 58012765-58012765 on Build GRCh38
rs141472399 Chr.16: 58012793-58012793 on Build GRCh38
rs927046495 Chr.16: 58012841-58012841 on Build GRCh38
rs773541347 Chr.16: 58012772-58012772 on Build GRCh38
rs889185494 Chr.16: 58012774-58012774 on Build GRCh38
rs1056694345 Chr.16: 58012992-58012992 on Build GRCh38
rs971879232 Chr.16: 58012909-58012909 on Build GRCh38
rs892720541 Chr.16: 58013022-58013022 on Build GRCh38
rs927042902 Chr.16: 58012660-58012660 on Build GRCh38
rs566032187 Chr.16: 58012782-58012782 on Build GRCh38
rs968491278 Chr.16: 58012783-58012783 on Build GRCh38
rs553741946 Chr.16: 58012638-58012638 on Build GRCh38
rs901878286 Chr.16: 58012749-58012749 on Build GRCh38
rs890723646 Chr.16: 58012781-58012790 on Build GRCh38
rs553032924 Chr.16: 58013041-58013041 on Build GRCh38
rs1022512713 Chr.16: 58013061-58013061 on Build GRCh38
rs534392869 Chr.16: 58012964-58012964 on Build GRCh38
rs899257294 Chr.16: 58012634-58012634 on Build GRCh38
rs777298198 Chr.16: 58012869-58012869 on Build GRCh38
rs558868700 Chr.16: 58012906-58012906 on Build GRCh38
rs953405206 Chr.16: 58012824-58012824 on Build GRCh38
rs990391233 Chr.16: 58012908-58012908 on Build GRCh38
rs547894656 Chr.16: 58012682-58012682 on Build GRCh38
rs748411023 Chr.16: 58012693-58012693 on Build GRCh38
rs369096616 Chr.16: 58012646-58012646 on Build GRCh38
rs571040358 Chr.16: 58012907-58012907 on Build GRCh38
rs570362291 Chr.16: 58012848-58012848 on Build GRCh38
rs185762636 Chr.16: 58012728-58012728 on Build GRCh38
rs945907014 Chr.16: 58012744-58012744 on Build GRCh38
rs920671962 Chr.16: 58012922-58012922 on Build GRCh38
rs759901009 Chr.16: 58012876-58012876 on Build GRCh38
rs865900748 Chr.16: 58012713-58012713 on Build GRCh38
rs1041686258 Chr.16: 58012958-58012958 on Build GRCh38
rs981106390 Chr.16: 58012821-58012821 on Build GRCh38
rs1009732763 Chr.16: 58013040-58013040 on Build GRCh38
rs1009755613 Chr.16: 58012751-58012751 on Build GRCh38
rs542058856 Chr.16: 58012867-58012867 on Build GRCh38
rs145819484 Chr.16: 58012672-58012672 on Build GRCh38
rs1045023763 Chr.16: 58012956-58012956 on Build GRCh38
rs949712165 Chr.16: 58012641-58012641 on Build GRCh38
rs768806126 Chr.16: 58012676-58012676 on Build GRCh38
rs892605560 Chr.16: 58012729-58012729 on Build GRCh38
rs1041654960 Chr.16: 58012745-58012745 on Build GRCh38
rs766807078 Chr.16: 58012866-58012866 on Build GRCh38
rs933327915 Chr.16: 58013073-58013073 on Build GRCh38
rs930767665 Chr.16: 58012929-58012929 on Build GRCh38
rs537768544 Chr.16: 58012849-58012849 on Build GRCh38
rs905219006 Chr.16: 58012957-58012957 on Build GRCh38
rs548644328 Chr.16: 58012808-58012808 on Build GRCh38
rs1006816334 Chr.16: 58012781-58012782 on Build GRCh38

View Full Product Details