Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67571055-67571641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTCF
Entrez Gene ID
10664
Gene Name
CCCTC-binding factor
Gene Aliases
MRD21
UniGene
Hs.368367
Gene Chromosome Location
Chr.16: 67562407-67639185 on Build GRCh38
Transcripts
NM_001191022.1 NM_006565.3 XM_005255775.3 XM_017022868.1 XM_017022869.1

SNP Detail

SNP ID Chromosome Location
rs749367209 Chr.16: 67571389-67571389 on Build GRCh38
rs367657072 Chr.16: 67571425-67571425 on Build GRCh38
rs572860237 Chr.16: 67571280-67571281 on Build GRCh38
rs550066499 Chr.16: 67571552-67571552 on Build GRCh38
rs757744624 Chr.16: 67571483-67571483 on Build GRCh38
rs762767712 Chr.16: 67571114-67571114 on Build GRCh38
rs750834324 Chr.16: 67571183-67571183 on Build GRCh38
rs934286512 Chr.16: 67571331-67571331 on Build GRCh38
rs923290706 Chr.16: 67571232-67571232 on Build GRCh38
rs535847824 Chr.16: 67571598-67571598 on Build GRCh38
rs900324526 Chr.16: 67571274-67571274 on Build GRCh38
rs948505375 Chr.16: 67571156-67571156 on Build GRCh38
rs1056501783 Chr.16: 67571574-67571574 on Build GRCh38
rs1035682860 Chr.16: 67571337-67571337 on Build GRCh38
rs949544471 Chr.16: 67571585-67571585 on Build GRCh38
rs527473113 Chr.16: 67571428-67571428 on Build GRCh38
rs761585365 Chr.16: 67571397-67571397 on Build GRCh38
rs983476277 Chr.16: 67571259-67571259 on Build GRCh38
rs867869233 Chr.16: 67571357-67571357 on Build GRCh38
rs1055227975 Chr.16: 67571084-67571084 on Build GRCh38
rs112443468 Chr.16: 67571603-67571603 on Build GRCh38
rs570703747 Chr.16: 67571436-67571436 on Build GRCh38
rs900369637 Chr.16: 67571393-67571393 on Build GRCh38
rs552447148 Chr.16: 67571429-67571429 on Build GRCh38
rs184162604 Chr.16: 67571149-67571149 on Build GRCh38
rs1000014428 Chr.16: 67571535-67571535 on Build GRCh38
rs1045385730 Chr.16: 67571131-67571131 on Build GRCh38
rs111484512 Chr.16: 67571258-67571258 on Build GRCh38
rs531937072 Chr.16: 67571208-67571208 on Build GRCh38
rs991062194 Chr.16: 67571561-67571561 on Build GRCh38
rs778024448 Chr.16: 67571371-67571371 on Build GRCh38
rs961026478 Chr.16: 67571353-67571353 on Build GRCh38
rs75129385 Chr.16: 67571092-67571092 on Build GRCh38
rs1017434338 Chr.16: 67571378-67571378 on Build GRCh38
rs948489309 Chr.16: 67571299-67571299 on Build GRCh38
rs553812422 Chr.16: 67571618-67571618 on Build GRCh38
rs147176456 Chr.16: 67571328-67571328 on Build GRCh38
rs890341319 Chr.16: 67571227-67571227 on Build GRCh38
rs1023786283 Chr.16: 67571372-67571372 on Build GRCh38
rs902525103 Chr.16: 67571312-67571312 on Build GRCh38
rs1044206835 Chr.16: 67571478-67571478 on Build GRCh38
rs944619636 Chr.16: 67571235-67571235 on Build GRCh38
rs902557208 Chr.16: 67571534-67571534 on Build GRCh38
rs982309433 Chr.16: 67571586-67571586 on Build GRCh38
rs966717419 Chr.16: 67571096-67571096 on Build GRCh38
rs537785706 Chr.16: 67571531-67571531 on Build GRCh38
rs928969893 Chr.16: 67571414-67571414 on Build GRCh38
rs142075387 Chr.16: 67571584-67571584 on Build GRCh38
rs900355464 Chr.16: 67571455-67571455 on Build GRCh38
rs752975479 Chr.16: 67571175-67571175 on Build GRCh38
rs1011522014 Chr.16: 67571370-67571370 on Build GRCh38
rs557237093 Chr.16: 67571190-67571190 on Build GRCh38
rs775563926 Chr.16: 67571097-67571097 on Build GRCh38
rs920325815 Chr.16: 67571409-67571409 on Build GRCh38
rs1038920684 Chr.16: 67571236-67571236 on Build GRCh38
rs1052781939 Chr.16: 67571352-67571352 on Build GRCh38
rs1031553245 Chr.16: 67571178-67571178 on Build GRCh38
rs763708954 Chr.16: 67571155-67571155 on Build GRCh38
rs934443803 Chr.16: 67571607-67571607 on Build GRCh38
rs961168335 Chr.16: 67571115-67571115 on Build GRCh38
rs188711712 Chr.16: 67571161-67571161 on Build GRCh38
rs946503864 Chr.16: 67571456-67571456 on Build GRCh38
rs896464474 Chr.16: 67571582-67571582 on Build GRCh38
rs962860264 Chr.16: 67571615-67571615 on Build GRCh38
rs1011384125 Chr.16: 67571182-67571182 on Build GRCh38
rs868322282 Chr.16: 67571441-67571441 on Build GRCh38
rs950819067 Chr.16: 67571252-67571252 on Build GRCh38
rs868422564 Chr.16: 67571090-67571090 on Build GRCh38
rs957282387 Chr.16: 67571179-67571179 on Build GRCh38
rs990908290 Chr.16: 67571151-67571151 on Build GRCh38
rs77172747 Chr.16: 67571222-67571222 on Build GRCh38
rs184427773 Chr.16: 67571424-67571424 on Build GRCh38
rs572261657 Chr.16: 67571093-67571093 on Build GRCh38
rs148636444 Chr.16: 67571421-67571421 on Build GRCh38
rs1016238156 Chr.16: 67571495-67571495 on Build GRCh38
rs1038953130 Chr.16: 67571431-67571431 on Build GRCh38
rs193216663 Chr.16: 67571289-67571289 on Build GRCh38
rs1022364860 Chr.16: 67571291-67571291 on Build GRCh38
rs983388425 Chr.16: 67571481-67571481 on Build GRCh38

View Full Product Details