Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46336901-46337491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL17B
Entrez Gene ID
100506084
Gene Name
ADP ribosylation factor like GTPase 17B
Gene Aliases
ARL17, ARL17A
UniGene
Hs.559259
Gene Chromosome Location
Chr.17: 46274184-46361872 on Build GRCh38
Transcripts
NM_001103154.1 XM_005256918.4 XM_011524160.2 XM_011524161.2 XM_011524162.2 XM_011524163.2 XM_011524164.2 XM_011524165.2 XM_011524166.2 XM_011524167.2 NM_014834.4 XM_005257879.4 XM_005257880.4 XM_005257882.4 XM_006722210.3 XM_006722211.3 XM_011525539.2 XM_017025472.1 XM_017025473.1

Gene Symbol
LRRC37A
Entrez Gene ID
9884
Gene Name
leucine rich repeat containing 37A
Gene Aliases
LRRC37
UniGene
Hs.579108
Gene Chromosome Location
Chr.17: 46248044-46339367 on Build GRCh38
Transcripts
NM_001103154.1 XM_005256918.4 XM_011524160.2 XM_011524161.2 XM_011524162.2 XM_011524163.2 XM_011524164.2 XM_011524165.2 XM_011524166.2 XM_011524167.2 NM_014834.4 XM_005257879.4 XM_005257880.4 XM_005257882.4 XM_006722210.3 XM_006722211.3 XM_011525539.2 XM_017025472.1 XM_017025473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750094 c.4944C>T p.L1648L Chr.17: 46337458-46337458 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751812210 Chr.17: 46337467-46337467 on Build GRCh38
rs745932792 Chr.17: 46337348-46337348 on Build GRCh38
rs761676124 Chr.17: 46337414-46337414 on Build GRCh38
rs144016818 Chr.17: 46337455-46337455 on Build GRCh38
rs377002486 Chr.17: 46337454-46337454 on Build GRCh38
rs753324403 Chr.17: 46337307-46337307 on Build GRCh38
rs982392063 Chr.17: 46337049-46337049 on Build GRCh38
rs766028191 Chr.17: 46337448-46337448 on Build GRCh38
rs760258114 Chr.17: 46337440-46337440 on Build GRCh38
rs928201491 Chr.17: 46337071-46337071 on Build GRCh38
rs773058377 Chr.17: 46337433-46337433 on Build GRCh38
rs373292921 Chr.17: 46337450-46337450 on Build GRCh38
rs1976643 Chr.17: 46337318-46337318 on Build GRCh38
rs376136920 Chr.17: 46337404-46337404 on Build GRCh38
rs781402511 Chr.17: 46337344-46337344 on Build GRCh38
rs915017300 Chr.17: 46337362-46337362 on Build GRCh38
rs747430425 Chr.17: 46337335-46337335 on Build GRCh38
rs768665208 Chr.17: 46337407-46337407 on Build GRCh38
rs273545 Chr.17: 46337471-46337471 on Build GRCh38
rs273544 Chr.17: 46337241-46337241 on Build GRCh38
rs935033939 Chr.17: 46337228-46337228 on Build GRCh38
rs770991485 Chr.17: 46337336-46337336 on Build GRCh38
rs553824507 Chr.17: 46337323-46337323 on Build GRCh38
rs779550399 Chr.17: 46337284-46337284 on Build GRCh38
rs370152403 Chr.17: 46337420-46337420 on Build GRCh38
rs775371624 Chr.17: 46337395-46337395 on Build GRCh38
rs969636759 Chr.17: 46336943-46336943 on Build GRCh38
rs773979273 Chr.17: 46337408-46337408 on Build GRCh38
rs769924360 Chr.17: 46337354-46337354 on Build GRCh38
rs1052517390 Chr.17: 46337256-46337256 on Build GRCh38
rs916710891 Chr.17: 46336936-46336936 on Build GRCh38

View Full Product Details