Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46555306-46555862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL17A
Entrez Gene ID
51326
Gene Name
ADP ribosylation factor like GTPase 17A
Gene Aliases
ARF1P2, ARL17P1
UniGene
Hs.559259
Gene Chromosome Location
Chr.17: 46500213-46579781 on Build GRCh38
Transcripts
NM_001113738.1 NM_001288811.1 NM_001288812.1 NM_001288813.1 NM_016632.2 XM_005257439.4 XM_011524875.2 XM_011524876.2 XM_011524877.2 XM_011524878.2 XM_017024729.1 NM_001006607.2 XM_011524841.2 XM_011524842.2 XM_011524843.2 XM_011524844.2 XM_011524846.2 XM_011524848.2

Gene Symbol
LRRC37A2
Entrez Gene ID
474170
Gene Name
leucine rich repeat containing 37 member A2
Gene Aliases
LRRC37
UniGene
Hs.646568
Gene Chromosome Location
Chr.17: 46512657-46556747 on Build GRCh38
Transcripts
NM_001113738.1 NM_001288811.1 NM_001288812.1 NM_001288813.1 NM_016632.2 XM_005257439.4 XM_011524875.2 XM_011524876.2 XM_011524877.2 XM_011524878.2 XM_017024729.1 NM_001006607.2 XM_011524841.2 XM_011524842.2 XM_011524843.2 XM_011524844.2 XM_011524846.2 XM_011524848.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430266 c.4957A>C p.M1653L Chr.17: 46555327-46555327 on Build GRCh38
4699068 c.4997C>T p.A1666V Chr.17: 46555367-46555367 on Build GRCh38
5828549 c.5102_5103delAG p.? Chr.17: 46555578-46555580 on Build GRCh38
4822538 c.4944C>G p.L1648L Chr.17: 46555314-46555314 on Build GRCh38
4594534 c.4998A>G p.A1666A Chr.17: 46555368-46555368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544848062 Chr.17: 46555699-46555699 on Build GRCh38
rs145809511 Chr.17: 46555648-46555648 on Build GRCh38
rs772353375 Chr.17: 46555330-46555330 on Build GRCh38
rs779098686 Chr.17: 46555382-46555382 on Build GRCh38
rs553421218 Chr.17: 46555463-46555463 on Build GRCh38
rs756821507 Chr.17: 46555378-46555378 on Build GRCh38
rs780695661 Chr.17: 46555380-46555380 on Build GRCh38
rs765273414 Chr.17: 46555413-46555413 on Build GRCh38
rs757969549 Chr.17: 46555369-46555369 on Build GRCh38
rs762876067 Chr.17: 46555423-46555423 on Build GRCh38
rs1055925695 Chr.17: 46555826-46555826 on Build GRCh38
rs747159494 Chr.17: 46555521-46555521 on Build GRCh38
rs199645771 Chr.17: 46555363-46555363 on Build GRCh38
rs751067064 Chr.17: 46555374-46555374 on Build GRCh38
rs201353569 Chr.17: 46555439-46555439 on Build GRCh38
rs11898 Chr.17: 46555368-46555368 on Build GRCh38
rs536035466 Chr.17: 46555544-46555544 on Build GRCh38
rs539595179 Chr.17: 46555410-46555410 on Build GRCh38
rs573510352 Chr.17: 46555538-46555538 on Build GRCh38
rs775120595 Chr.17: 46555420-46555420 on Build GRCh38
rs746004658 Chr.17: 46555548-46555548 on Build GRCh38
rs374337863 Chr.17: 46555390-46555390 on Build GRCh38
rs778294207 Chr.17: 46555518-46555518 on Build GRCh38
rs748455312 Chr.17: 46555513-46555513 on Build GRCh38
rs753877922 Chr.17: 46555345-46555345 on Build GRCh38
rs200731781 Chr.17: 46555399-46555399 on Build GRCh38
rs920633366 Chr.17: 46555718-46555718 on Build GRCh38
rs879841755 Chr.17: 46555666-46555666 on Build GRCh38
rs775558227 Chr.17: 46555558-46555558 on Build GRCh38
rs759542938 Chr.17: 46555353-46555353 on Build GRCh38
rs747069888 Chr.17: 46555397-46555397 on Build GRCh38
rs202076405 Chr.17: 46555687-46555687 on Build GRCh38
rs772152770 Chr.17: 46555383-46555383 on Build GRCh38
rs769618483 Chr.17: 46555549-46555549 on Build GRCh38
rs563139158 Chr.17: 46555793-46555793 on Build GRCh38
rs769237527 Chr.17: 46555381-46555381 on Build GRCh38
rs756909541 Chr.17: 46555446-46555446 on Build GRCh38
rs755418288 Chr.17: 46555470-46555470 on Build GRCh38
rs749972523 Chr.17: 46555469-46555469 on Build GRCh38
rs930628900 Chr.17: 46555804-46555804 on Build GRCh38
rs779514096 Chr.17: 46555483-46555483 on Build GRCh38
rs776852324 Chr.17: 46555402-46555402 on Build GRCh38
rs939929373 Chr.17: 46555590-46555590 on Build GRCh38
rs758889287 Chr.17: 46555514-46555514 on Build GRCh38
rs760906063 Chr.17: 46555339-46555339 on Build GRCh38
rs1038673830 Chr.17: 46555704-46555704 on Build GRCh38
rs773412579 Chr.17: 46555336-46555336 on Build GRCh38
rs781130172 Chr.17: 46555628-46555630 on Build GRCh38
rs777466030 Chr.17: 46555371-46555371 on Build GRCh38
rs372877369 Chr.17: 46555327-46555327 on Build GRCh38

View Full Product Details