Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 54961516-54962084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX11
Entrez Gene ID
1353
Gene Name
COX11, cytochrome c oxidase copper chaperone
Gene Aliases
COX11P
UniGene
Hs.591171
Gene Chromosome Location
Chr.17: 54951898-54971024 on Build GRCh38
Transcripts
NM_001162861.2 NM_001162862.2 NM_004375.4 NR_027941.2 NR_027942.2 NR_135677.1 XM_011524342.2 XM_017024192.1 XM_017024193.1 XM_017024194.1 XM_017024195.1 XM_017024196.1 NM_001321174.1 NM_001321175.1 NM_001321176.1 NM_005486.2 XM_017024002.1

Gene Symbol
TOM1L1
Entrez Gene ID
10040
Gene Name
target of myb1 like 1 membrane trafficking protein
Gene Aliases
SRCASM
UniGene
Hs.153504
Gene Chromosome Location
Chr.17: 54900691-54961967 on Build GRCh38
Transcripts
NM_001162861.2 NM_001162862.2 NM_004375.4 NR_027941.2 NR_027942.2 NR_135677.1 XM_011524342.2 XM_017024192.1 XM_017024193.1 XM_017024194.1 XM_017024195.1 XM_017024196.1 NM_001321174.1 NM_001321175.1 NM_001321176.1 NM_005486.2 XM_017024002.1

SNP Detail

SNP ID Chromosome Location
rs961440449 Chr.17: 54961942-54961942 on Build GRCh38
rs943128041 Chr.17: 54961688-54961688 on Build GRCh38
rs946932452 Chr.17: 54962003-54962003 on Build GRCh38
rs138242114 Chr.17: 54961554-54961554 on Build GRCh38
rs557077088 Chr.17: 54961631-54961631 on Build GRCh38
rs4829 Chr.17: 54961954-54961954 on Build GRCh38
rs796176576 Chr.17: 54961975-54961975 on Build GRCh38
rs1044263137 Chr.17: 54962054-54962054 on Build GRCh38
rs552337861 Chr.17: 54961634-54961634 on Build GRCh38
rs1052767400 Chr.17: 54961805-54961805 on Build GRCh38
rs934239279 Chr.17: 54961591-54961591 on Build GRCh38
rs565458605 Chr.17: 54961914-54961914 on Build GRCh38
rs774925637 Chr.17: 54961665-54961665 on Build GRCh38
rs985704647 Chr.17: 54961965-54961965 on Build GRCh38
rs1005785190 Chr.17: 54961929-54961929 on Build GRCh38
rs188170223 Chr.17: 54961745-54961745 on Build GRCh38
rs750795242 Chr.17: 54961788-54961788 on Build GRCh38
rs972744025 Chr.17: 54961944-54961944 on Build GRCh38
rs1014138709 Chr.17: 54961893-54961893 on Build GRCh38
rs183704823 Chr.17: 54961743-54961743 on Build GRCh38
rs1039976486 Chr.17: 54961937-54961938 on Build GRCh38
rs911391010 Chr.17: 54961975-54961976 on Build GRCh38
rs55829839 Chr.17: 54961793-54961793 on Build GRCh38
rs544229306 Chr.17: 54962052-54962052 on Build GRCh38
rs755430014 Chr.17: 54962036-54962036 on Build GRCh38
rs530926770 Chr.17: 54961795-54961795 on Build GRCh38
rs575437449 Chr.17: 54961677-54961677 on Build GRCh38
rs3214380 Chr.17: 54961965-54961965 on Build GRCh38
rs969888782 Chr.17: 54961927-54961927 on Build GRCh38
rs16955429 Chr.17: 54961738-54961738 on Build GRCh38
rs895769422 Chr.17: 54961841-54961841 on Build GRCh38
rs1050357819 Chr.17: 54961655-54961655 on Build GRCh38
rs181029321 Chr.17: 54961729-54961729 on Build GRCh38
rs998544820 Chr.17: 54961789-54961789 on Build GRCh38
rs764735419 Chr.17: 54961555-54961555 on Build GRCh38
rs955818760 Chr.17: 54961963-54961964 on Build GRCh38

View Full Product Details