Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43360699-43361244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20
Entrez Gene ID
991
Gene Name
cell division cycle 20
Gene Aliases
CDC20A, bA276H19.3, p55CDC
UniGene
Hs.524947
Gene Chromosome Location
Chr.1: 43358955-43363203 on Build GRCh38
Transcripts
NM_001255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846741 c.911C>T p.T304I Chr.1: 43360795-43360795 on Build GRCh38
4455947 c.968A>G p.H323R Chr.1: 43360852-43360852 on Build GRCh38
1185439 c.965G>A p.R322Q Chr.1: 43360849-43360849 on Build GRCh38
6006535 c.1027G>T p.G343C Chr.1: 43360911-43360911 on Build GRCh38
3733995 c.1061A>T p.H354L Chr.1: 43360945-43360945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201462108 Chr.1: 43360968-43360968 on Build GRCh38
rs746546243 Chr.1: 43361163-43361164 on Build GRCh38
rs761743390 Chr.1: 43361136-43361136 on Build GRCh38
rs776517495 Chr.1: 43360979-43360979 on Build GRCh38
rs574307989 Chr.1: 43361030-43361030 on Build GRCh38
rs766010708 Chr.1: 43361004-43361004 on Build GRCh38
rs759422114 Chr.1: 43360791-43360791 on Build GRCh38
rs766199335 Chr.1: 43361215-43361218 on Build GRCh38
rs769715038 Chr.1: 43361146-43361146 on Build GRCh38
rs991296026 Chr.1: 43360740-43360740 on Build GRCh38
rs146864157 Chr.1: 43361102-43361102 on Build GRCh38
rs911860124 Chr.1: 43361116-43361116 on Build GRCh38
rs755438046 Chr.1: 43360852-43360852 on Build GRCh38
rs370835580 Chr.1: 43360942-43360942 on Build GRCh38
rs774688858 Chr.1: 43360907-43360907 on Build GRCh38
rs45507193 Chr.1: 43361091-43361091 on Build GRCh38
rs1022835451 Chr.1: 43361057-43361057 on Build GRCh38
rs765936509 Chr.1: 43360958-43360958 on Build GRCh38
rs781752713 Chr.1: 43360735-43360735 on Build GRCh38
rs199542353 Chr.1: 43360897-43360897 on Build GRCh38
rs373135328 Chr.1: 43360913-43360913 on Build GRCh38
rs181732244 Chr.1: 43360995-43360995 on Build GRCh38
rs749335066 Chr.1: 43360767-43360767 on Build GRCh38
rs779426473 Chr.1: 43361213-43361213 on Build GRCh38
rs774403959 Chr.1: 43360789-43360789 on Build GRCh38
rs960391482 Chr.1: 43360961-43360961 on Build GRCh38
rs552437721 Chr.1: 43360952-43360952 on Build GRCh38
rs1042846422 Chr.1: 43360974-43360974 on Build GRCh38
rs751237829 Chr.1: 43361170-43361170 on Build GRCh38
rs963433918 Chr.1: 43360787-43360787 on Build GRCh38
rs536319846 Chr.1: 43361039-43361039 on Build GRCh38
rs758000204 Chr.1: 43361000-43361000 on Build GRCh38
rs780282020 Chr.1: 43361122-43361122 on Build GRCh38
rs143739422 Chr.1: 43361222-43361222 on Build GRCh38
rs772205711 Chr.1: 43361117-43361117 on Build GRCh38
rs140305765 Chr.1: 43360849-43360849 on Build GRCh38
rs778470330 Chr.1: 43361011-43361011 on Build GRCh38
rs767199661 Chr.1: 43360815-43360815 on Build GRCh38
rs781415109 Chr.1: 43360874-43360874 on Build GRCh38
rs533181943 Chr.1: 43361048-43361048 on Build GRCh38
rs963218155 Chr.1: 43361083-43361083 on Build GRCh38
rs893937639 Chr.1: 43360962-43360962 on Build GRCh38
rs1035736308 Chr.1: 43361016-43361016 on Build GRCh38
rs879241526 Chr.1: 43361190-43361190 on Build GRCh38
rs374986622 Chr.1: 43360991-43360991 on Build GRCh38
rs112630800 Chr.1: 43360770-43360770 on Build GRCh38
rs768882314 Chr.1: 43360729-43360729 on Build GRCh38
rs757475680 Chr.1: 43361079-43361079 on Build GRCh38
rs761176631 Chr.1: 43361180-43361180 on Build GRCh38
rs200047932 Chr.1: 43360912-43360912 on Build GRCh38
rs1026424195 Chr.1: 43361094-43361094 on Build GRCh38
rs748600009 Chr.1: 43360755-43360755 on Build GRCh38
rs11555249 Chr.1: 43361166-43361166 on Build GRCh38
rs371199053 Chr.1: 43361071-43361071 on Build GRCh38
rs780975492 Chr.1: 43361029-43361029 on Build GRCh38
rs756518356 Chr.1: 43361009-43361009 on Build GRCh38
rs756714435 Chr.1: 43360881-43360881 on Build GRCh38
rs748491001 Chr.1: 43360880-43360880 on Build GRCh38
rs368676792 Chr.1: 43361148-43361148 on Build GRCh38
rs754379858 Chr.1: 43361012-43361012 on Build GRCh38
rs764546476 Chr.1: 43361181-43361181 on Build GRCh38
rs775560252 Chr.1: 43360818-43360818 on Build GRCh38
rs549920906 Chr.1: 43360758-43360758 on Build GRCh38
rs374006557 Chr.1: 43361110-43361110 on Build GRCh38
rs776617856 Chr.1: 43361132-43361132 on Build GRCh38
rs753367774 Chr.1: 43360831-43360831 on Build GRCh38
rs915711344 Chr.1: 43360760-43360760 on Build GRCh38
rs1801456 Chr.1: 43361123-43361123 on Build GRCh38
rs756714014 Chr.1: 43360845-43360845 on Build GRCh38
rs754185453 Chr.1: 43361189-43361189 on Build GRCh38
rs765991493 Chr.1: 43361167-43361167 on Build GRCh38
rs760534440 Chr.1: 43360821-43360821 on Build GRCh38
rs372597302 Chr.1: 43360848-43360848 on Build GRCh38
rs973553120 Chr.1: 43360796-43360796 on Build GRCh38
rs531794887 Chr.1: 43361128-43361128 on Build GRCh38
rs749421572 Chr.1: 43360902-43360902 on Build GRCh38
rs763960946 Chr.1: 43360830-43360830 on Build GRCh38
rs895745676 Chr.1: 43361025-43361025 on Build GRCh38
rs554304518 Chr.1: 43361007-43361007 on Build GRCh38
rs377262124 Chr.1: 43360928-43360928 on Build GRCh38
rs762489157 Chr.1: 43361153-43361153 on Build GRCh38
rs867593106 Chr.1: 43360836-43360836 on Build GRCh38
rs749882808 Chr.1: 43360999-43360999 on Build GRCh38
rs764048744 Chr.1: 43360970-43360970 on Build GRCh38
rs757848486 Chr.1: 43361197-43361197 on Build GRCh38

View Full Product Details