Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8369113-8369391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
UniGene
Hs.9613
Gene Chromosome Location
Chr.19: 8364127-8374375 on Build GRCh38
Transcripts
NM_001039667.2 NM_139314.2 NR_104213.1 XM_005272484.3 XM_005272485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4404924 c.621G>C p.Q207H Chr.19: 8369292-8369292 on Build GRCh38
6409702 c.619C>A p.Q207K Chr.19: 8369290-8369290 on Build GRCh38
1397754 c.629C>T p.P210L Chr.19: 8369300-8369300 on Build GRCh38
3541589 c.607G>A p.E203K Chr.19: 8369278-8369278 on Build GRCh38
6599655 c.584G>A p.G195E Chr.19: 8369255-8369255 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756335494 Chr.19: 8369288-8369288 on Build GRCh38
rs200977507 Chr.19: 8369213-8369213 on Build GRCh38
rs759309735 Chr.19: 8369342-8369343 on Build GRCh38
rs765119888 Chr.19: 8369360-8369361 on Build GRCh38
rs768362135 Chr.19: 8369218-8369218 on Build GRCh38
rs372986082 Chr.19: 8369251-8369251 on Build GRCh38
rs778125140 Chr.19: 8369175-8369175 on Build GRCh38
rs559356885 Chr.19: 8369241-8369241 on Build GRCh38
rs377342328 Chr.19: 8369169-8369169 on Build GRCh38
rs34600793 Chr.19: 8369216-8369216 on Build GRCh38
rs746388842 Chr.19: 8369319-8369319 on Build GRCh38
rs754257420 Chr.19: 8369304-8369304 on Build GRCh38
rs530029296 Chr.19: 8369168-8369168 on Build GRCh38
rs776638002 Chr.19: 8369222-8369222 on Build GRCh38
rs1038147222 Chr.19: 8369201-8369201 on Build GRCh38
rs949403681 Chr.19: 8369146-8369146 on Build GRCh38
rs192019697 Chr.19: 8369153-8369153 on Build GRCh38
rs779272884 Chr.19: 8369188-8369188 on Build GRCh38
rs202038040 Chr.19: 8369342-8369342 on Build GRCh38
rs746450280 Chr.19: 8369344-8369344 on Build GRCh38
rs77439987 Chr.19: 8369178-8369178 on Build GRCh38
rs768484549 Chr.19: 8369346-8369346 on Build GRCh38
rs761691527 Chr.19: 8369223-8369223 on Build GRCh38
rs587777517 Chr.19: 8369320-8369320 on Build GRCh38
rs77938377 Chr.19: 8369239-8369239 on Build GRCh38
rs1006366989 Chr.19: 8369300-8369300 on Build GRCh38
rs73925321 Chr.19: 8369167-8369167 on Build GRCh38
rs752715503 Chr.19: 8369276-8369276 on Build GRCh38
rs1003241110 Chr.19: 8369244-8369244 on Build GRCh38
rs771449485 Chr.19: 8369204-8369204 on Build GRCh38
rs776303264 Chr.19: 8369203-8369204 on Build GRCh38
rs955895416 Chr.19: 8369163-8369163 on Build GRCh38
rs368972947 Chr.19: 8369166-8369167 on Build GRCh38
rs767518522 Chr.19: 8369266-8369266 on Build GRCh38
rs200552010 Chr.19: 8369252-8369252 on Build GRCh38
rs776402882 Chr.19: 8369361-8369361 on Build GRCh38
rs376530345 Chr.19: 8369189-8369189 on Build GRCh38
rs751129667 Chr.19: 8369165-8369166 on Build GRCh38
rs765085157 Chr.19: 8369170-8369170 on Build GRCh38
rs779514049 Chr.19: 8369313-8369313 on Build GRCh38
rs866955749 Chr.19: 8369249-8369249 on Build GRCh38
rs35571542 Chr.19: 8369301-8369301 on Build GRCh38
rs573092895 Chr.19: 8369365-8369365 on Build GRCh38
rs777541240 Chr.19: 8369173-8369173 on Build GRCh38
rs766049937 Chr.19: 8369174-8369174 on Build GRCh38
rs781518361 Chr.19: 8369172-8369172 on Build GRCh38
rs997627653 Chr.19: 8369232-8369232 on Build GRCh38
rs76800055 Chr.19: 8369171-8369171 on Build GRCh38
rs974775681 Chr.19: 8369352-8369352 on Build GRCh38
rs757849242 Chr.19: 8369183-8369183 on Build GRCh38
rs758886725 Chr.19: 8369336-8369336 on Build GRCh38

View Full Product Details