Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18442650-18443161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL
Entrez Gene ID
8178
Gene Name
elongation factor for RNA polymerase II
Gene Aliases
C19orf17, ELL1, MEN, PPP1R68
UniGene
Hs.515260
Gene Chromosome Location
Chr.19: 18442663-18522129 on Build GRCh38
Transcripts
NM_006532.3 XM_017027335.1 XM_017027336.1 XM_017027337.1

SNP Detail

SNP ID Chromosome Location
rs986014507 Chr.19: 18442736-18442736 on Build GRCh38
rs866712436 Chr.19: 18442774-18442778 on Build GRCh38
rs888989133 Chr.19: 18443139-18443139 on Build GRCh38
rs397797978 Chr.19: 18442775-18442778 on Build GRCh38
rs1030725358 Chr.19: 18443000-18443001 on Build GRCh38
rs918690954 Chr.19: 18442760-18442760 on Build GRCh38
rs1029602986 Chr.19: 18442906-18442906 on Build GRCh38
rs1032861380 Chr.19: 18442700-18442700 on Build GRCh38
rs965385598 Chr.19: 18442743-18442743 on Build GRCh38
rs554499415 Chr.19: 18442798-18442798 on Build GRCh38
rs576420861 Chr.19: 18442813-18442813 on Build GRCh38
rs940774394 Chr.19: 18443080-18443080 on Build GRCh38
rs67126664 Chr.19: 18442771-18442774 on Build GRCh38
rs1040367195 Chr.19: 18442827-18442827 on Build GRCh38
rs902130879 Chr.19: 18442875-18442876 on Build GRCh38
rs969740727 Chr.19: 18443037-18443037 on Build GRCh38
rs992086541 Chr.19: 18442675-18442675 on Build GRCh38
rs996423085 Chr.19: 18442879-18442879 on Build GRCh38
rs1000459716 Chr.19: 18442697-18442697 on Build GRCh38
rs919777370 Chr.19: 18443067-18443067 on Build GRCh38
rs397709999 Chr.19: 18442884-18442884 on Build GRCh38
rs940661562 Chr.19: 18442801-18442801 on Build GRCh38
rs576203537 Chr.19: 18443114-18443114 on Build GRCh38
rs949780925 Chr.19: 18443008-18443008 on Build GRCh38
rs1000622069 Chr.19: 18442984-18442986 on Build GRCh38
rs901829381 Chr.19: 18443112-18443112 on Build GRCh38
rs189831493 Chr.19: 18442939-18442939 on Build GRCh38
rs907998979 Chr.19: 18443079-18443079 on Build GRCh38
rs148304841 Chr.19: 18442701-18442701 on Build GRCh38
rs11539528 Chr.19: 18442875-18442875 on Build GRCh38
rs1032769011 Chr.19: 18443138-18443138 on Build GRCh38
rs749142621 Chr.19: 18443001-18443003 on Build GRCh38
rs545516421 Chr.19: 18442863-18442863 on Build GRCh38
rs1001069768 Chr.19: 18443130-18443130 on Build GRCh38
rs1040798494 Chr.19: 18443111-18443111 on Build GRCh38
rs930095831 Chr.19: 18442778-18442778 on Build GRCh38
rs904000857 Chr.19: 18442972-18442972 on Build GRCh38
rs999960104 Chr.19: 18443135-18443135 on Build GRCh38
rs1056097761 Chr.19: 18442982-18442982 on Build GRCh38
rs542872794 Chr.19: 18442947-18442947 on Build GRCh38
rs557279313 Chr.19: 18442737-18442737 on Build GRCh38
rs776608000 Chr.19: 18443003-18443003 on Build GRCh38
rs981037534 Chr.19: 18442705-18442705 on Build GRCh38
rs556342454 Chr.19: 18443113-18443113 on Build GRCh38
rs1041446560 Chr.19: 18443033-18443033 on Build GRCh38
rs984848343 Chr.19: 18443070-18443070 on Build GRCh38
rs980685621 Chr.19: 18443042-18443042 on Build GRCh38
rs574701108 Chr.19: 18442676-18442676 on Build GRCh38
rs895475694 Chr.19: 18443000-18443000 on Build GRCh38
rs910057903 Chr.19: 18442874-18442874 on Build GRCh38
rs953264514 Chr.19: 18442720-18442720 on Build GRCh38
rs11319408 Chr.19: 18442876-18442876 on Build GRCh38
rs984668598 Chr.19: 18442810-18442810 on Build GRCh38
rs931108513 Chr.19: 18443069-18443069 on Build GRCh38
rs976850588 Chr.19: 18442751-18442751 on Build GRCh38

View Full Product Details