Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35358443-35359015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FFAR3
Entrez Gene ID
2865
Gene Name
free fatty acid receptor 3
Gene Aliases
FFA3R, GPR41, GPR42
UniGene
Hs.248055
Gene Chromosome Location
Chr.19: 35358106-35360491 on Build GRCh38
Transcripts
NM_005304.3 XM_011526858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
711988 c.87C>G p.L29L Chr.19: 35358977-35358977 on Build GRCh38
711989 c.87C>A p.L29L Chr.19: 35358977-35358977 on Build GRCh38
118374 c.96C>T p.N32N Chr.19: 35358986-35358986 on Build GRCh38
4436398 c.91C>G p.L31V Chr.19: 35358981-35358981 on Build GRCh38
5023069 c.105C>T p.A35A Chr.19: 35358995-35358995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369874702 Chr.19: 35358534-35358534 on Build GRCh38
rs773387604 Chr.19: 35358961-35358961 on Build GRCh38
rs760859202 Chr.19: 35358986-35358986 on Build GRCh38
rs1033477351 Chr.19: 35358525-35358525 on Build GRCh38
rs779424995 Chr.19: 35358959-35358959 on Build GRCh38
rs769651219 Chr.19: 35358908-35358908 on Build GRCh38
rs62109580 Chr.19: 35358764-35358764 on Build GRCh38
rs866504748 Chr.19: 35358679-35358679 on Build GRCh38
rs381099 Chr.19: 35358508-35358508 on Build GRCh38
rs551288253 Chr.19: 35358922-35358922 on Build GRCh38
rs770543372 Chr.19: 35358899-35358899 on Build GRCh38
rs375854528 Chr.19: 35358526-35358526 on Build GRCh38
rs776932133 Chr.19: 35358991-35358991 on Build GRCh38
rs530535002 Chr.19: 35358954-35358954 on Build GRCh38
rs768690809 Chr.19: 35358980-35358980 on Build GRCh38
rs758828825 Chr.19: 35358938-35358938 on Build GRCh38
rs775431489 Chr.19: 35358909-35358909 on Build GRCh38
rs962803434 Chr.19: 35358468-35358468 on Build GRCh38
rs573573596 Chr.19: 35358789-35358789 on Build GRCh38
rs558280055 Chr.19: 35358539-35358539 on Build GRCh38
rs753942882 Chr.19: 35358859-35358859 on Build GRCh38
rs371169601 Chr.19: 35358947-35358947 on Build GRCh38
rs59198030 Chr.19: 35358661-35358661 on Build GRCh38
rs753999232 Chr.19: 35358918-35358918 on Build GRCh38
rs778101022 Chr.19: 35358892-35358892 on Build GRCh38
rs753004610 Chr.19: 35358878-35358878 on Build GRCh38
rs146397125 Chr.19: 35358911-35358911 on Build GRCh38
rs543374650 Chr.19: 35358570-35358570 on Build GRCh38
rs765679497 Chr.19: 35358923-35358923 on Build GRCh38
rs746733456 Chr.19: 35358896-35358896 on Build GRCh38
rs550566313 Chr.19: 35358995-35358995 on Build GRCh38
rs772161262 Chr.19: 35358865-35358866 on Build GRCh38
rs761024959 Chr.19: 35358912-35358912 on Build GRCh38
rs755459537 Chr.19: 35358868-35358868 on Build GRCh38
rs368567016 Chr.19: 35358939-35358939 on Build GRCh38
rs760973410 Chr.19: 35358848-35358848 on Build GRCh38
rs913842356 Chr.19: 35358566-35358566 on Build GRCh38
rs943096444 Chr.19: 35358775-35358775 on Build GRCh38
rs751901197 Chr.19: 35358946-35358946 on Build GRCh38
rs879920889 Chr.19: 35358631-35358632 on Build GRCh38
rs766657837 Chr.19: 35358914-35358914 on Build GRCh38
rs779432262 Chr.19: 35358870-35358870 on Build GRCh38
rs749286356 Chr.19: 35358973-35358973 on Build GRCh38
rs10412120 Chr.19: 35358545-35358545 on Build GRCh38
rs1039204431 Chr.19: 35358786-35358786 on Build GRCh38
rs542544927 Chr.19: 35358836-35358836 on Build GRCh38
rs745335617 Chr.19: 35358906-35358906 on Build GRCh38
rs539350244 Chr.19: 35358569-35358569 on Build GRCh38
rs753309549 Chr.19: 35358928-35358928 on Build GRCh38
rs369303830 Chr.19: 35358788-35358788 on Build GRCh38
rs368520877 Chr.19: 35358840-35358840 on Build GRCh38
rs780730507 Chr.19: 35358905-35358905 on Build GRCh38
rs528333217 Chr.19: 35358853-35358853 on Build GRCh38
rs4806132 Chr.19: 35358981-35358981 on Build GRCh38
rs758735425 Chr.19: 35358884-35358884 on Build GRCh38
rs924962012 Chr.19: 35358672-35358672 on Build GRCh38

View Full Product Details