Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49791924-49792481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A1
Entrez Gene ID
160
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Gene Aliases
ADTAA, AP2-ALPHA, CLAPA1
UniGene
Hs.467125
Gene Chromosome Location
Chr.19: 49766923-49807114 on Build GRCh38
Transcripts
NM_014203.2 NM_130787.2 XM_011526556.2 XM_011526557.2 NR_106857.1

Gene Symbol
MIR6799
Entrez Gene ID
102465479
Gene Name
microRNA 6799
Gene Aliases
hsa-mir-6799
UniGene
Gene Chromosome Location
Chr.19: 49791866-49791934 on Build GRCh38
Transcripts
NM_014203.2 NM_130787.2 XM_011526556.2 XM_011526557.2 NR_106857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
440001 c.599A>G p.H200R Chr.19: 49792060-49792060 on Build GRCh38
1729209 c.527A>C p.Y176S Chr.19: 49791988-49791988 on Build GRCh38
4766394 c.600C>A p.H200Q Chr.19: 49792061-49792061 on Build GRCh38
5726733 c.538C>T p.P180S Chr.19: 49791999-49791999 on Build GRCh38
4498313 c.517C>T p.L173F Chr.19: 49791978-49791978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964620382 Chr.19: 49792037-49792037 on Build GRCh38
rs777106067 Chr.19: 49792013-49792013 on Build GRCh38
rs577561148 Chr.19: 49792177-49792177 on Build GRCh38
rs549028335 Chr.19: 49792226-49792226 on Build GRCh38
rs755160434 Chr.19: 49792062-49792062 on Build GRCh38
rs1028595422 Chr.19: 49792389-49792389 on Build GRCh38
rs542491126 Chr.19: 49792192-49792192 on Build GRCh38
rs756472720 Chr.19: 49792070-49792070 on Build GRCh38
rs550362429 Chr.19: 49792344-49792344 on Build GRCh38
rs185433793 Chr.19: 49792448-49792448 on Build GRCh38
rs546622160 Chr.19: 49792251-49792251 on Build GRCh38
rs145035752 Chr.19: 49792169-49792169 on Build GRCh38
rs573160315 Chr.19: 49792158-49792158 on Build GRCh38
rs560661854 Chr.19: 49792269-49792269 on Build GRCh38
rs748685860 Chr.19: 49791984-49791984 on Build GRCh38
rs372089995 Chr.19: 49792182-49792182 on Build GRCh38
rs995898316 Chr.19: 49792313-49792313 on Build GRCh38
rs745545796 Chr.19: 49792282-49792318 on Build GRCh38
rs770483282 Chr.19: 49791986-49791986 on Build GRCh38
rs571134797 Chr.19: 49792076-49792076 on Build GRCh38
rs571196252 Chr.19: 49792292-49792292 on Build GRCh38
rs780941148 Chr.19: 49791949-49791949 on Build GRCh38
rs753579080 Chr.19: 49792435-49792435 on Build GRCh38
rs538502181 Chr.19: 49792303-49792303 on Build GRCh38
rs990781416 Chr.19: 49792085-49792085 on Build GRCh38
rs538441143 Chr.19: 49792077-49792077 on Build GRCh38
rs769677283 Chr.19: 49791963-49791963 on Build GRCh38
rs763435834 Chr.19: 49792036-49792036 on Build GRCh38
rs770836220 Chr.19: 49792090-49792090 on Build GRCh38
rs940110395 Chr.19: 49792179-49792179 on Build GRCh38
rs950559503 Chr.19: 49792081-49792081 on Build GRCh38
rs536301631 Chr.19: 49792104-49792104 on Build GRCh38
rs766467556 Chr.19: 49792041-49792041 on Build GRCh38
rs534627681 Chr.19: 49791995-49791995 on Build GRCh38
rs560755614 Chr.19: 49792232-49792232 on Build GRCh38
rs898507309 Chr.19: 49792307-49792307 on Build GRCh38
rs1008452601 Chr.19: 49792420-49792420 on Build GRCh38
rs544811638 Chr.19: 49792189-49792189 on Build GRCh38
rs759251973 Chr.19: 49791997-49791997 on Build GRCh38
rs563001648 Chr.19: 49792004-49792004 on Build GRCh38
rs546856843 Chr.19: 49792031-49792031 on Build GRCh38
rs4802626 Chr.19: 49792451-49792451 on Build GRCh38
rs147212058 Chr.19: 49792198-49792198 on Build GRCh38
rs575091062 Chr.19: 49792165-49792165 on Build GRCh38
rs916532126 Chr.19: 49792452-49792452 on Build GRCh38
rs368241524 Chr.19: 49791953-49791953 on Build GRCh38
rs575342456 Chr.19: 49792089-49792089 on Build GRCh38
rs376270224 Chr.19: 49792085-49792085 on Build GRCh38
rs568684368 Chr.19: 49792446-49792446 on Build GRCh38
rs908641922 Chr.19: 49792397-49792397 on Build GRCh38
rs762300764 Chr.19: 49792027-49792027 on Build GRCh38
rs528067270 Chr.19: 49792239-49792239 on Build GRCh38
rs773220772 Chr.19: 49791965-49791965 on Build GRCh38
rs941413262 Chr.19: 49792406-49792406 on Build GRCh38
rs890037422 Chr.19: 49792418-49792418 on Build GRCh38
rs536365394 Chr.19: 49792447-49792447 on Build GRCh38
rs966846496 Chr.19: 49792443-49792443 on Build GRCh38
rs765613912 Chr.19: 49792028-49792028 on Build GRCh38
rs573184747 Chr.19: 49792450-49792450 on Build GRCh38
rs554689934 Chr.19: 49792135-49792135 on Build GRCh38
rs1046207303 Chr.19: 49792108-49792108 on Build GRCh38
rs1056740321 Chr.19: 49792457-49792457 on Build GRCh38
rs370723855 Chr.19: 49792061-49792061 on Build GRCh38
rs1019798879 Chr.19: 49792421-49792421 on Build GRCh38
rs540608090 Chr.19: 49792163-49792163 on Build GRCh38

View Full Product Details