Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57499438-57500005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTCFL
Entrez Gene ID
140690
Gene Name
CCCTC-binding factor like
Gene Aliases
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
UniGene
Hs.131543
Gene Chromosome Location
Chr.20: 57495965-57525652 on Build GRCh38
Transcripts
NM_001269040.1 NM_001269041.1 NM_001269042.1 NM_001269043.1 NM_001269044.1 NM_001269046.1 NM_001269049.1 NM_001269054.1 NM_080618.3 NR_072975.1

SNP Detail

SNP ID Chromosome Location
rs568145475 Chr.20: 57499781-57499781 on Build GRCh38
rs1000874804 Chr.20: 57499689-57499689 on Build GRCh38
rs1032638685 Chr.20: 57499699-57499699 on Build GRCh38
rs754374891 Chr.20: 57499715-57499715 on Build GRCh38
rs76016304 Chr.20: 57499735-57499735 on Build GRCh38
rs184860475 Chr.20: 57499681-57499681 on Build GRCh38
rs2209885 Chr.20: 57499851-57499851 on Build GRCh38
rs181742704 Chr.20: 57499808-57499808 on Build GRCh38
rs112558007 Chr.20: 57499842-57499842 on Build GRCh38
rs374645135 Chr.20: 57499542-57499542 on Build GRCh38
rs1003897255 Chr.20: 57499812-57499812 on Build GRCh38
rs540281665 Chr.20: 57499976-57499976 on Build GRCh38
rs890506888 Chr.20: 57499523-57499523 on Build GRCh38
rs909432387 Chr.20: 57499767-57499767 on Build GRCh38
rs965228230 Chr.20: 57499552-57499552 on Build GRCh38
rs926508359 Chr.20: 57499718-57499718 on Build GRCh38
rs952413424 Chr.20: 57499657-57499657 on Build GRCh38
rs79683305 Chr.20: 57499761-57499761 on Build GRCh38
rs926363783 Chr.20: 57499840-57499840 on Build GRCh38
rs981893352 Chr.20: 57499832-57499832 on Build GRCh38
rs534299496 Chr.20: 57499810-57499810 on Build GRCh38
rs759650983 Chr.20: 57499779-57499779 on Build GRCh38
rs1009037592 Chr.20: 57499528-57499528 on Build GRCh38
rs73625048 Chr.20: 57499892-57499893 on Build GRCh38
rs569170131 Chr.20: 57499724-57499724 on Build GRCh38
rs537640546 Chr.20: 57499732-57499732 on Build GRCh38
rs940827947 Chr.20: 57499468-57499468 on Build GRCh38
rs527934333 Chr.20: 57499717-57499717 on Build GRCh38
rs1039520181 Chr.20: 57499802-57499802 on Build GRCh38
rs970369851 Chr.20: 57499831-57499831 on Build GRCh38
rs1015294940 Chr.20: 57499821-57499821 on Build GRCh38
rs1028317715 Chr.20: 57499629-57499629 on Build GRCh38
rs539461477 Chr.20: 57499834-57499834 on Build GRCh38
rs74182725 Chr.20: 57499894-57499895 on Build GRCh38
rs1005169378 Chr.20: 57499962-57499962 on Build GRCh38
rs60693208 Chr.20: 57499480-57499480 on Build GRCh38
rs72553291 Chr.20: 57499895-57499896 on Build GRCh38
rs544466022 Chr.20: 57499852-57499852 on Build GRCh38
rs772392752 Chr.20: 57499569-57499569 on Build GRCh38
rs893676710 Chr.20: 57499763-57499763 on Build GRCh38
rs1042272571 Chr.20: 57499875-57499875 on Build GRCh38
rs773522000 Chr.20: 57499616-57499616 on Build GRCh38
rs995027335 Chr.20: 57499956-57499956 on Build GRCh38
rs900707697 Chr.20: 57499806-57499806 on Build GRCh38
rs945387583 Chr.20: 57499764-57499764 on Build GRCh38
rs930941010 Chr.20: 57499748-57499748 on Build GRCh38
rs965236635 Chr.20: 57499723-57499723 on Build GRCh38
rs544839976 Chr.20: 57499479-57499479 on Build GRCh38
rs10659599 Chr.20: 57499893-57499894 on Build GRCh38
rs908302778 Chr.20: 57499711-57499711 on Build GRCh38
rs765533477 Chr.20: 57499556-57499560 on Build GRCh38
rs7267256 Chr.20: 57499954-57499954 on Build GRCh38
rs886590303 Chr.20: 57499958-57499958 on Build GRCh38
rs760965118 Chr.20: 57499651-57499651 on Build GRCh38
rs892484272 Chr.20: 57499843-57499843 on Build GRCh38
rs758655279 Chr.20: 57499945-57499945 on Build GRCh38
rs73915846 Chr.20: 57499957-57499957 on Build GRCh38
rs944705158 Chr.20: 57499889-57499889 on Build GRCh38
rs1013613665 Chr.20: 57499546-57499546 on Build GRCh38
rs1017795300 Chr.20: 57499529-57499529 on Build GRCh38
rs905282629 Chr.20: 57499835-57499835 on Build GRCh38
rs983818143 Chr.20: 57499757-57499757 on Build GRCh38
rs530169319 Chr.20: 57499520-57499520 on Build GRCh38
rs771344600 Chr.20: 57499635-57499635 on Build GRCh38
rs560085217 Chr.20: 57499647-57499647 on Build GRCh38
rs149038306 Chr.20: 57499556-57499556 on Build GRCh38
rs558983489 Chr.20: 57499841-57499841 on Build GRCh38
rs746130704 Chr.20: 57499714-57499714 on Build GRCh38
rs1042341786 Chr.20: 57499777-57499777 on Build GRCh38
rs575739714 Chr.20: 57499466-57499466 on Build GRCh38
rs189283177 Chr.20: 57499807-57499807 on Build GRCh38
rs911926722 Chr.20: 57499883-57499883 on Build GRCh38
rs973713395 Chr.20: 57499620-57499620 on Build GRCh38
rs1019576276 Chr.20: 57499927-57499927 on Build GRCh38

View Full Product Details