Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52743455-52743869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVMER34-1
Entrez Gene ID
100288413
Gene Name
endogenous retrovirus group MER34 member 1
Gene Aliases
-
UniGene
Hs.363087
Gene Chromosome Location
Chr.4: 52743517-52751640 on Build GRCh38
Transcripts
NM_001242690.1 NM_024534.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5452563 c.1670A>C p.Q557P Chr.4: 52743851-52743851 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761337707 Chr.4: 52743576-52743576 on Build GRCh38
rs556955728 Chr.4: 52743835-52743835 on Build GRCh38
rs1027765501 Chr.4: 52743546-52743546 on Build GRCh38
rs762064302 Chr.4: 52743756-52743756 on Build GRCh38
rs113024322 Chr.4: 52743714-52743714 on Build GRCh38
rs148889780 Chr.4: 52743581-52743581 on Build GRCh38
rs1037305541 Chr.4: 52743629-52743629 on Build GRCh38
rs890322893 Chr.4: 52743489-52743489 on Build GRCh38
rs1031441005 Chr.4: 52743505-52743505 on Build GRCh38
rs1049250547 Chr.4: 52743689-52743689 on Build GRCh38
rs534372544 Chr.4: 52743709-52743709 on Build GRCh38
rs997409828 Chr.4: 52743498-52743498 on Build GRCh38
rs918723780 Chr.4: 52743660-52743660 on Build GRCh38
rs997357593 Chr.4: 52743809-52743809 on Build GRCh38
rs776814220 Chr.4: 52743723-52743723 on Build GRCh38
rs774165054 Chr.4: 52743553-52743553 on Build GRCh38
rs914663257 Chr.4: 52743567-52743567 on Build GRCh38
rs142679241 Chr.4: 52743757-52743757 on Build GRCh38
rs537086615 Chr.4: 52743817-52743817 on Build GRCh38
rs931510056 Chr.4: 52743678-52743678 on Build GRCh38
rs944535248 Chr.4: 52743523-52743523 on Build GRCh38
rs74336454 Chr.4: 52743537-52743537 on Build GRCh38
rs750752030 Chr.4: 52743830-52743830 on Build GRCh38
rs530956173 Chr.4: 52743527-52743527 on Build GRCh38
rs67541691 Chr.4: 52743602-52743602 on Build GRCh38
rs990247986 Chr.4: 52743772-52743772 on Build GRCh38
rs887533505 Chr.4: 52743702-52743702 on Build GRCh38
rs980092711 Chr.4: 52743590-52743590 on Build GRCh38
rs567982867 Chr.4: 52743551-52743551 on Build GRCh38
rs1013607681 Chr.4: 52743583-52743583 on Build GRCh38
rs957486192 Chr.4: 52743767-52743767 on Build GRCh38
rs13119273 Chr.4: 52743615-52743615 on Build GRCh38

View Full Product Details