Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 248065-248614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20C
Entrez Gene ID
56975
Gene Name
family with sequence similarity 20 member C
Gene Aliases
DMP-4, DMP4, GEF-CK, RNS
UniGene
Hs.134742
Gene Chromosome Location
Chr.7: 192939-260774 on Build GRCh38
Transcripts
NM_020223.3 XM_017012450.1 XM_017012451.1 XM_017012452.1 XM_017012455.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7268673 c.976G>T p.V326F Chr.7: 248334-248334 on Build GRCh38
4639225 c.996G>A p.R332R Chr.7: 248354-248354 on Build GRCh38
4155713 c.962T>A p.L321Q Chr.7: 248320-248320 on Build GRCh38
4748269 c.1040A>G p.K347R Chr.7: 248398-248398 on Build GRCh38
3663131 c.990C>T p.A330A Chr.7: 248348-248348 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953743954 Chr.7: 248303-248303 on Build GRCh38
rs752044491 Chr.7: 248417-248417 on Build GRCh38
rs930775345 Chr.7: 248338-248338 on Build GRCh38
rs1049668174 Chr.7: 248457-248457 on Build GRCh38
rs1009315107 Chr.7: 248148-248148 on Build GRCh38
rs781207320 Chr.7: 248447-248447 on Build GRCh38
rs147477230 Chr.7: 248257-248257 on Build GRCh38
rs1039529929 Chr.7: 248385-248385 on Build GRCh38
rs1000619262 Chr.7: 248456-248456 on Build GRCh38
rs868408490 Chr.7: 248331-248331 on Build GRCh38
rs527843022 Chr.7: 248392-248392 on Build GRCh38
rs545071136 Chr.7: 248302-248302 on Build GRCh38
rs905304724 Chr.7: 248124-248124 on Build GRCh38
rs149565814 Chr.7: 248136-248136 on Build GRCh38
rs765745848 Chr.7: 248379-248379 on Build GRCh38
rs553736166 Chr.7: 248243-248243 on Build GRCh38
rs930871751 Chr.7: 248236-248236 on Build GRCh38
rs983451162 Chr.7: 248349-248349 on Build GRCh38
rs768470051 Chr.7: 248275-248275 on Build GRCh38
rs797044462 Chr.7: 248340-248340 on Build GRCh38
rs374190041 Chr.7: 248299-248299 on Build GRCh38
rs1052777968 Chr.7: 248482-248482 on Build GRCh38
rs191875159 Chr.7: 248105-248105 on Build GRCh38
rs926834990 Chr.7: 248358-248358 on Build GRCh38
rs747888335 Chr.7: 248478-248478 on Build GRCh38
rs986834654 Chr.7: 248307-248307 on Build GRCh38
rs185225273 Chr.7: 248453-248453 on Build GRCh38
rs896333890 Chr.7: 248138-248138 on Build GRCh38
rs1002275389 Chr.7: 248133-248133 on Build GRCh38
rs796051851 Chr.7: 248312-248312 on Build GRCh38
rs533696074 Chr.7: 248531-248531 on Build GRCh38
rs551453368 Chr.7: 248498-248498 on Build GRCh38
rs1030556586 Chr.7: 248191-248191 on Build GRCh38
rs528467859 Chr.7: 248246-248246 on Build GRCh38
rs759219400 Chr.7: 248348-248348 on Build GRCh38
rs576724109 Chr.7: 248276-248276 on Build GRCh38
rs930672213 Chr.7: 248104-248104 on Build GRCh38
rs1011302830 Chr.7: 248561-248561 on Build GRCh38
rs879064067 Chr.7: 248189-248189 on Build GRCh38
rs961109722 Chr.7: 248327-248327 on Build GRCh38
rs567812491 Chr.7: 248450-248450 on Build GRCh38
rs972225643 Chr.7: 248214-248214 on Build GRCh38
rs568839598 Chr.7: 248462-248462 on Build GRCh38
rs369431616 Chr.7: 248267-248267 on Build GRCh38
rs535062850 Chr.7: 248155-248155 on Build GRCh38
rs772843483 Chr.7: 248291-248291 on Build GRCh38
rs557906175 Chr.7: 248207-248207 on Build GRCh38
rs898041914 Chr.7: 248101-248101 on Build GRCh38
rs148750123 Chr.7: 248203-248203 on Build GRCh38
rs967930301 Chr.7: 248178-248178 on Build GRCh38
rs144201916 Chr.7: 248192-248192 on Build GRCh38
rs571366916 Chr.7: 248506-248506 on Build GRCh38
rs986579013 Chr.7: 248195-248195 on Build GRCh38
rs755845471 Chr.7: 248438-248438 on Build GRCh38
rs562650430 Chr.7: 248454-248454 on Build GRCh38
rs961142701 Chr.7: 248479-248479 on Build GRCh38
rs1007929551 Chr.7: 248476-248476 on Build GRCh38
rs979095370 Chr.7: 248179-248179 on Build GRCh38
rs190152406 Chr.7: 248483-248483 on Build GRCh38
rs1019400429 Chr.7: 248310-248310 on Build GRCh38
rs527902877 Chr.7: 248093-248093 on Build GRCh38
rs138754098 Chr.7: 248092-248092 on Build GRCh38
rs377615599 Chr.7: 248384-248384 on Build GRCh38
rs531623625 Chr.7: 248298-248298 on Build GRCh38
rs919545275 Chr.7: 248235-248235 on Build GRCh38
rs556060643 Chr.7: 248570-248570 on Build GRCh38
rs377142273 Chr.7: 248463-248463 on Build GRCh38
rs1047792035 Chr.7: 248091-248091 on Build GRCh38
rs145542456 Chr.7: 248535-248535 on Build GRCh38
rs547974510 Chr.7: 248442-248442 on Build GRCh38
rs759012718 Chr.7: 248391-248391 on Build GRCh38
rs538819921 Chr.7: 248376-248376 on Build GRCh38
rs536526217 Chr.7: 248537-248537 on Build GRCh38
rs1026706693 Chr.7: 248580-248580 on Build GRCh38
rs867143631 Chr.7: 248408-248408 on Build GRCh38
rs530415232 Chr.7: 248451-248451 on Build GRCh38
rs140130809 Chr.7: 248271-248271 on Build GRCh38
rs1051908097 Chr.7: 248448-248448 on Build GRCh38
rs142310673 Chr.7: 248208-248208 on Build GRCh38
rs918029688 Chr.7: 248443-248443 on Build GRCh38
rs747060498 Chr.7: 248245-248245 on Build GRCh38
rs540429942 Chr.7: 248211-248211 on Build GRCh38
rs753579879 Chr.7: 248342-248342 on Build GRCh38
rs1020867132 Chr.7: 248177-248177 on Build GRCh38
rs182397396 Chr.7: 248588-248588 on Build GRCh38
rs555342295 Chr.7: 248165-248165 on Build GRCh38
rs972256876 Chr.7: 248329-248329 on Build GRCh38
rs998463877 Chr.7: 248199-248199 on Build GRCh38

View Full Product Details