Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 967287-967798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX19
Entrez Gene ID
90639
Gene Name
COX19, cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.121593
Gene Chromosome Location
Chr.7: 964850-975599 on Build GRCh38
Transcripts
NM_001031617.2

SNP Detail

SNP ID Chromosome Location
rs531433970 Chr.7: 967377-967377 on Build GRCh38
rs957799470 Chr.7: 967546-967546 on Build GRCh38
rs966438144 Chr.7: 967729-967729 on Build GRCh38
rs897238810 Chr.7: 967772-967772 on Build GRCh38
rs116884794 Chr.7: 967333-967333 on Build GRCh38
rs906389191 Chr.7: 967412-967412 on Build GRCh38
rs73257955 Chr.7: 967378-967378 on Build GRCh38
rs778401642 Chr.7: 967733-967733 on Build GRCh38
rs888509958 Chr.7: 967600-967600 on Build GRCh38
rs938882576 Chr.7: 967559-967559 on Build GRCh38
rs941424875 Chr.7: 967743-967743 on Build GRCh38
rs563790561 Chr.7: 967738-967738 on Build GRCh38
rs973664872 Chr.7: 967708-967708 on Build GRCh38
rs1030240095 Chr.7: 967526-967526 on Build GRCh38
rs954443110 Chr.7: 967336-967336 on Build GRCh38
rs891628573 Chr.7: 967614-967614 on Build GRCh38
rs941468637 Chr.7: 967602-967602 on Build GRCh38
rs142440946 Chr.7: 967458-967458 on Build GRCh38
rs536526218 Chr.7: 967571-967571 on Build GRCh38
rs562688857 Chr.7: 967353-967353 on Build GRCh38
rs982195513 Chr.7: 967731-967731 on Build GRCh38
rs879181657 Chr.7: 967545-967545 on Build GRCh38
rs1029835775 Chr.7: 967635-967635 on Build GRCh38
rs372639959 Chr.7: 967625-967625 on Build GRCh38
rs141727809 Chr.7: 967525-967525 on Build GRCh38
rs1050550338 Chr.7: 967583-967583 on Build GRCh38
rs556231698 Chr.7: 967572-967572 on Build GRCh38
rs1006987434 Chr.7: 967690-967690 on Build GRCh38
rs144685456 Chr.7: 967475-967475 on Build GRCh38
rs1050886395 Chr.7: 967727-967727 on Build GRCh38
rs1019389417 Chr.7: 967524-967524 on Build GRCh38
rs1054967765 Chr.7: 967586-967586 on Build GRCh38
rs771666657 Chr.7: 967397-967397 on Build GRCh38
rs1002288075 Chr.7: 967423-967423 on Build GRCh38
rs530065786 Chr.7: 967723-967723 on Build GRCh38
rs572177566 Chr.7: 967655-967655 on Build GRCh38
rs369446565 Chr.7: 967567-967567 on Build GRCh38
rs1019459723 Chr.7: 967347-967347 on Build GRCh38
rs527789405 Chr.7: 967395-967395 on Build GRCh38
rs1046644387 Chr.7: 967410-967410 on Build GRCh38
rs561146902 Chr.7: 967400-967400 on Build GRCh38
rs75326730 Chr.7: 967489-967489 on Build GRCh38
rs760553729 Chr.7: 967362-967362 on Build GRCh38
rs1034004909 Chr.7: 967639-967639 on Build GRCh38
rs940908903 Chr.7: 967430-967430 on Build GRCh38
rs908041594 Chr.7: 967673-967673 on Build GRCh38
rs910211047 Chr.7: 967371-967371 on Build GRCh38
rs375740426 Chr.7: 967575-967575 on Build GRCh38
rs115997437 Chr.7: 967530-967530 on Build GRCh38
rs571319841 Chr.7: 967534-967534 on Build GRCh38
rs1001585873 Chr.7: 967630-967630 on Build GRCh38
rs960092682 Chr.7: 967651-967651 on Build GRCh38
rs890558766 Chr.7: 967595-967595 on Build GRCh38
rs968752896 Chr.7: 967533-967533 on Build GRCh38
rs3735675 Chr.7: 967747-967747 on Build GRCh38
rs996277551 Chr.7: 967459-967459 on Build GRCh38
rs993023675 Chr.7: 967562-967562 on Build GRCh38
rs990624581 Chr.7: 967737-967737 on Build GRCh38
rs543043272 Chr.7: 967428-967428 on Build GRCh38
rs886712891 Chr.7: 967406-967406 on Build GRCh38
rs1008453350 Chr.7: 967491-967491 on Build GRCh38
rs542574946 Chr.7: 967334-967334 on Build GRCh38
rs890038898 Chr.7: 967658-967658 on Build GRCh38
rs189163485 Chr.7: 967622-967622 on Build GRCh38
rs151023526 Chr.7: 967380-967380 on Build GRCh38
rs1046350754 Chr.7: 967568-967568 on Build GRCh38
rs937929563 Chr.7: 967736-967736 on Build GRCh38
rs1021464643 Chr.7: 967623-967623 on Build GRCh38
rs913594795 Chr.7: 967552-967552 on Build GRCh38
rs145402608 Chr.7: 967739-967739 on Build GRCh38
rs752703058 Chr.7: 967388-967388 on Build GRCh38
rs1008971418 Chr.7: 967598-967598 on Build GRCh38
rs540197178 Chr.7: 967306-967306 on Build GRCh38
rs931946007 Chr.7: 967579-967579 on Build GRCh38
rs748797061 Chr.7: 967716-967716 on Build GRCh38
rs757529772 Chr.7: 967492-967492 on Build GRCh38
rs150492081 Chr.7: 967705-967705 on Build GRCh38
rs908033015 Chr.7: 967569-967569 on Build GRCh38
rs117593139 Chr.7: 967543-967543 on Build GRCh38
rs988460255 Chr.7: 967364-967364 on Build GRCh38
rs945269114 Chr.7: 967553-967553 on Build GRCh38
rs989267795 Chr.7: 967720-967720 on Build GRCh38
rs73669928 Chr.7: 967573-967573 on Build GRCh38
rs553768820 Chr.7: 967547-967547 on Build GRCh38
rs989520765 Chr.7: 967551-967551 on Build GRCh38
rs541140805 Chr.7: 967695-967695 on Build GRCh38
rs572135622 Chr.7: 967726-967726 on Build GRCh38
rs1005927322 Chr.7: 967467-967467 on Build GRCh38
rs1037428458 Chr.7: 967604-967604 on Build GRCh38
rs553807536 Chr.7: 967321-967321 on Build GRCh38
rs1037737392 Chr.7: 967446-967449 on Build GRCh38
rs78461900 Chr.7: 967397-967397 on Build GRCh38

View Full Product Details